Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144316844-144317107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT1
Entrez Gene ID
8694
Gene Name
diacylglycerol O-acyltransferase 1
Gene Aliases
ARAT, ARGP1, DGAT, DIAR7
UniGene
Hs.521954
Gene Chromosome Location
Chr.8: 144314583-144326919 on Build GRCh38
Transcripts
NM_012079.5 XM_011517356.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2871053 c.1290G>A p.A430A Chr.8: 144316874-144316874 on Build GRCh38
1097202 c.1280G>A p.R427H Chr.8: 144316884-144316884 on Build GRCh38
3834272 c.1232C>T p.S411L Chr.8: 144317038-144317038 on Build GRCh38
2871058 c.1168T>C p.Y390H Chr.8: 144317102-144317102 on Build GRCh38
2871054 c.1270C>T p.R424* Chr.8: 144316894-144316894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781948192 Chr.8: 144317063-144317063 on Build GRCh38
rs573410614 Chr.8: 144317038-144317038 on Build GRCh38
rs782385704 Chr.8: 144317052-144317053 on Build GRCh38
rs782146939 Chr.8: 144316979-144316979 on Build GRCh38
rs527868304 Chr.8: 144317004-144317004 on Build GRCh38
rs782255407 Chr.8: 144317052-144317052 on Build GRCh38
rs782542784 Chr.8: 144316945-144316945 on Build GRCh38
rs782394462 Chr.8: 144316924-144316924 on Build GRCh38
rs782136994 Chr.8: 144317028-144317028 on Build GRCh38
rs782230110 Chr.8: 144316989-144316989 on Build GRCh38
rs782164877 Chr.8: 144317086-144317086 on Build GRCh38
rs782560002 Chr.8: 144316930-144316930 on Build GRCh38
rs782323808 Chr.8: 144317060-144317060 on Build GRCh38
rs375843442 Chr.8: 144317084-144317084 on Build GRCh38
rs782381938 Chr.8: 144316882-144316882 on Build GRCh38
rs782003054 Chr.8: 144316974-144316974 on Build GRCh38
rs782367223 Chr.8: 144316973-144316973 on Build GRCh38
rs782678457 Chr.8: 144317055-144317055 on Build GRCh38
rs782595918 Chr.8: 144316961-144316961 on Build GRCh38
rs201304578 Chr.8: 144316868-144316868 on Build GRCh38
rs782590742 Chr.8: 144317037-144317037 on Build GRCh38
rs376578086 Chr.8: 144316934-144316934 on Build GRCh38
rs782480746 Chr.8: 144317046-144317046 on Build GRCh38
rs782186271 Chr.8: 144316951-144316951 on Build GRCh38
rs782179106 Chr.8: 144317056-144317056 on Build GRCh38
rs782738491 Chr.8: 144317014-144317014 on Build GRCh38
rs376419190 Chr.8: 144316943-144316943 on Build GRCh38
rs782354909 Chr.8: 144316885-144316885 on Build GRCh38
rs199880017 Chr.8: 144316929-144316929 on Build GRCh38
rs193169779 Chr.8: 144317083-144317083 on Build GRCh38
rs1138517 Chr.8: 144317034-144317034 on Build GRCh38
rs782420041 Chr.8: 144316990-144316990 on Build GRCh38
rs576833495 Chr.8: 144316935-144316935 on Build GRCh38
rs377582872 Chr.8: 144317054-144317054 on Build GRCh38
rs372051069 Chr.8: 144316904-144316904 on Build GRCh38
rs374096688 Chr.8: 144316941-144316941 on Build GRCh38
rs369528418 Chr.8: 144316939-144316939 on Build GRCh38
rs142347230 Chr.8: 144317025-144317025 on Build GRCh38
rs147655123 Chr.8: 144316884-144316884 on Build GRCh38
rs781911802 Chr.8: 144317031-144317031 on Build GRCh38
rs936786241 Chr.8: 144316910-144316910 on Build GRCh38
rs368966034 Chr.8: 144316965-144316965 on Build GRCh38
rs1040694143 Chr.8: 144316942-144316942 on Build GRCh38
rs782328686 Chr.8: 144316957-144316957 on Build GRCh38
rs782744860 Chr.8: 144316902-144316902 on Build GRCh38
rs782420668 Chr.8: 144316894-144316894 on Build GRCh38
rs782641412 Chr.8: 144317050-144317050 on Build GRCh38
rs201815397 Chr.8: 144316865-144316865 on Build GRCh38
rs782210757 Chr.8: 144317005-144317005 on Build GRCh38
rs782284944 Chr.8: 144316893-144316893 on Build GRCh38
rs369376240 Chr.8: 144316869-144316869 on Build GRCh38
rs782689512 Chr.8: 144316947-144316947 on Build GRCh38
rs782058254 Chr.8: 144317005-144317005 on Build GRCh38
rs782091446 Chr.8: 144317074-144317074 on Build GRCh38
rs782802659 Chr.8: 144317030-144317030 on Build GRCh38
rs918197083 Chr.8: 144316866-144316866 on Build GRCh38
rs545199725 Chr.8: 144316970-144316970 on Build GRCh38
rs782116263 Chr.8: 144316913-144316913 on Build GRCh38
rs781922801 Chr.8: 144316901-144316901 on Build GRCh38
rs376029962 Chr.8: 144316991-144316991 on Build GRCh38

View Full Product Details