Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27355324-27355597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
UniGene
Hs.194694
Gene Chromosome Location
Chr.1: 27355179-27368037 on Build GRCh38
Transcripts
NM_001297609.1 NM_004672.4 XM_017002771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6707392 c.3832C>T p.R1278* Chr.1: 27355426-27355426 on Build GRCh38
4700364 c.3793G>A p.A1265T Chr.1: 27355441-27355441 on Build GRCh38
4700363 c.3817G>A p.A1273T Chr.1: 27355441-27355441 on Build GRCh38
4700365 c.187G>A p.G63S Chr.1: 27355441-27355441 on Build GRCh38
6707393 c.3808C>T p.R1270* Chr.1: 27355426-27355426 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs551405268 Chr.1: 27355518-27355518 on Build GRCh38
rs908050361 Chr.1: 27355442-27355442 on Build GRCh38
rs762759891 Chr.1: 27355504-27355504 on Build GRCh38
rs201154544 Chr.1: 27355361-27355361 on Build GRCh38
rs773239731 Chr.1: 27355359-27355359 on Build GRCh38
rs750139985 Chr.1: 27355388-27355388 on Build GRCh38
rs187573626 Chr.1: 27355533-27355533 on Build GRCh38
rs748479770 Chr.1: 27355429-27355429 on Build GRCh38
rs755460439 Chr.1: 27355521-27355521 on Build GRCh38
rs761482425 Chr.1: 27355479-27355479 on Build GRCh38
rs869092970 Chr.1: 27355492-27355492 on Build GRCh38
rs760605085 Chr.1: 27355473-27355473 on Build GRCh38
rs748347648 Chr.1: 27355345-27355345 on Build GRCh38
rs781559146 Chr.1: 27355426-27355426 on Build GRCh38
rs770013342 Chr.1: 27355435-27355435 on Build GRCh38
rs1038652927 Chr.1: 27355476-27355476 on Build GRCh38
rs779767545 Chr.1: 27355403-27355403 on Build GRCh38
rs141435319 Chr.1: 27355450-27355450 on Build GRCh38
rs775398297 Chr.1: 27355362-27355362 on Build GRCh38
rs530633142 Chr.1: 27355548-27355548 on Build GRCh38
rs765172051 Chr.1: 27355484-27355484 on Build GRCh38
rs753589818 Chr.1: 27355372-27355372 on Build GRCh38
rs763889833 Chr.1: 27355371-27355371 on Build GRCh38
rs370280703 Chr.1: 27355401-27355401 on Build GRCh38
rs756885689 Chr.1: 27355376-27355376 on Build GRCh38
rs766984232 Chr.1: 27355519-27355520 on Build GRCh38
rs776448014 Chr.1: 27355478-27355478 on Build GRCh38
rs774421165 Chr.1: 27355467-27355467 on Build GRCh38
rs749573279 Chr.1: 27355453-27355453 on Build GRCh38
rs374778272 Chr.1: 27355493-27355493 on Build GRCh38
rs770997298 Chr.1: 27355464-27355464 on Build GRCh38
rs764829142 Chr.1: 27355378-27355378 on Build GRCh38
rs544318017 Chr.1: 27355425-27355425 on Build GRCh38
rs768501911 Chr.1: 27355474-27355474 on Build GRCh38
rs917702880 Chr.1: 27355423-27355423 on Build GRCh38
rs760585492 Chr.1: 27355366-27355366 on Build GRCh38
rs1020919674 Chr.1: 27355568-27355568 on Build GRCh38
rs747511532 Chr.1: 27355415-27355415 on Build GRCh38
rs940888969 Chr.1: 27355452-27355452 on Build GRCh38
rs765918928 Chr.1: 27355506-27355506 on Build GRCh38
rs376953864 Chr.1: 27355527-27355527 on Build GRCh38
rs1009485205 Chr.1: 27355567-27355575 on Build GRCh38
rs769915729 Chr.1: 27355356-27355356 on Build GRCh38

View Full Product Details