Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47009743-47009972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF1
Entrez Gene ID
266977
Gene Name
adhesion G protein-coupled receptor F1
Gene Aliases
GPR110, KPG_012, PGR19, hGPCR36
UniGene
Hs.733762
Gene Chromosome Location
Chr.6: 46997703-47042363 on Build GRCh38
Transcripts
NM_153840.3 XM_011514469.1 XM_017010735.1 XM_017010736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7142511 c.1503C>T p.V501V Chr.6: 47009932-47009932 on Build GRCh38
6673441 c.976_977insT p.S326fs*32 Chr.6: 47009867-47009868 on Build GRCh38
6673440 c.933G>A p.T311T Chr.6: 47009911-47009911 on Build GRCh38
6673444 c.886G>A p.V296I Chr.6: 47009958-47009958 on Build GRCh38
7165107 c.1680G>A p.T560T Chr.6: 47009755-47009755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748514839 Chr.6: 47009800-47009800 on Build GRCh38
rs778795668 Chr.6: 47009782-47009782 on Build GRCh38
rs987850677 Chr.6: 47009867-47009868 on Build GRCh38
rs760162530 Chr.6: 47009839-47009839 on Build GRCh38
rs778241405 Chr.6: 47009927-47009927 on Build GRCh38
rs571038697 Chr.6: 47009911-47009911 on Build GRCh38
rs757111776 Chr.6: 47009778-47009778 on Build GRCh38
rs748429245 Chr.6: 47009912-47009912 on Build GRCh38
rs751093145 Chr.6: 47009800-47009801 on Build GRCh38
rs771370107 Chr.6: 47009830-47009830 on Build GRCh38
rs763503923 Chr.6: 47009813-47009813 on Build GRCh38
rs757148706 Chr.6: 47009829-47009830 on Build GRCh38
rs374050541 Chr.6: 47009832-47009832 on Build GRCh38
rs751642301 Chr.6: 47009902-47009902 on Build GRCh38
rs533646624 Chr.6: 47009865-47009865 on Build GRCh38
rs763798898 Chr.6: 47009858-47009858 on Build GRCh38
rs750253635 Chr.6: 47009886-47009886 on Build GRCh38
rs762675226 Chr.6: 47009785-47009786 on Build GRCh38
rs970706004 Chr.6: 47009812-47009812 on Build GRCh38
rs753529424 Chr.6: 47009860-47009860 on Build GRCh38
rs953051403 Chr.6: 47009915-47009915 on Build GRCh38
rs1005111013 Chr.6: 47009844-47009844 on Build GRCh38
rs112946460 Chr.6: 47009799-47009799 on Build GRCh38
rs184466568 Chr.6: 47009884-47009884 on Build GRCh38
rs202087361 Chr.6: 47009908-47009908 on Build GRCh38
rs139252619 Chr.6: 47009789-47009789 on Build GRCh38
rs758292658 Chr.6: 47009900-47009900 on Build GRCh38
rs770162717 Chr.6: 47009926-47009926 on Build GRCh38
rs749617565 Chr.6: 47009930-47009930 on Build GRCh38
rs78642422 Chr.6: 47009888-47009888 on Build GRCh38
rs570355288 Chr.6: 47009901-47009901 on Build GRCh38
rs747061232 Chr.6: 47009796-47009796 on Build GRCh38
rs770322285 Chr.6: 47009803-47009803 on Build GRCh38
rs763538186 Chr.6: 47009799-47009800 on Build GRCh38
rs771458223 Chr.6: 47009939-47009939 on Build GRCh38
rs987193234 Chr.6: 47009936-47009936 on Build GRCh38
rs748565869 Chr.6: 47009805-47009805 on Build GRCh38
rs913626076 Chr.6: 47009883-47009883 on Build GRCh38
rs193250765 Chr.6: 47009861-47009861 on Build GRCh38
rs775007924 Chr.6: 47009836-47009836 on Build GRCh38
rs775805794 Chr.6: 47009850-47009850 on Build GRCh38
rs142552580 Chr.6: 47009792-47009792 on Build GRCh38
rs780145105 Chr.6: 47009793-47009793 on Build GRCh38
rs926280442 Chr.6: 47009933-47009933 on Build GRCh38

View Full Product Details