Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1108544-1108791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
UniGene
Hs.408708
Gene Chromosome Location
Chr.19: 1107634-1174283 on Build GRCh38
Transcripts
NM_001100122.1 NM_014963.2 XM_011527804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7327157 c.3446-10C>T p.? Chr.19: 1108714-1108714 on Build GRCh38
5687590 c.3665A>T p.Q1222L Chr.19: 1108784-1108784 on Build GRCh38
4650437 c.3693C>T p.C1231C Chr.19: 1108682-1108682 on Build GRCh38
6377972 c.3453G>T p.K1151N Chr.19: 1108697-1108697 on Build GRCh38
5744615 c.3740T>C p.L1247P Chr.19: 1108581-1108581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021819735 Chr.19: 1108588-1108596 on Build GRCh38
rs752295232 Chr.19: 1108730-1108730 on Build GRCh38
rs573603595 Chr.19: 1108714-1108714 on Build GRCh38
rs900338992 Chr.19: 1108664-1108664 on Build GRCh38
rs544160705 Chr.19: 1108569-1108569 on Build GRCh38
rs1048460671 Chr.19: 1108619-1108619 on Build GRCh38
rs941841795 Chr.19: 1108644-1108644 on Build GRCh38
rs373031411 Chr.19: 1108667-1108667 on Build GRCh38
rs747334665 Chr.19: 1108691-1108691 on Build GRCh38
rs1030676604 Chr.19: 1108675-1108675 on Build GRCh38
rs780249690 Chr.19: 1108751-1108751 on Build GRCh38
rs968838666 Chr.19: 1108588-1108588 on Build GRCh38
rs777043340 Chr.19: 1108707-1108707 on Build GRCh38
rs767137863 Chr.19: 1108719-1108719 on Build GRCh38
rs748540310 Chr.19: 1108761-1108761 on Build GRCh38
rs755985588 Chr.19: 1108642-1108642 on Build GRCh38
rs376128681 Chr.19: 1108768-1108768 on Build GRCh38
rs1047423153 Chr.19: 1108732-1108732 on Build GRCh38
rs765560127 Chr.19: 1108728-1108728 on Build GRCh38
rs757222589 Chr.19: 1108741-1108741 on Build GRCh38
rs541518951 Chr.19: 1108694-1108694 on Build GRCh38
rs185699102 Chr.19: 1108756-1108756 on Build GRCh38
rs753726220 Chr.19: 1108735-1108735 on Build GRCh38
rs769008081 Chr.19: 1108700-1108700 on Build GRCh38
rs765769914 Chr.19: 1108710-1108710 on Build GRCh38
rs980276495 Chr.19: 1108589-1108589 on Build GRCh38
rs763967934 Chr.19: 1108733-1108733 on Build GRCh38
rs781500675 Chr.19: 1108757-1108757 on Build GRCh38
rs772290379 Chr.19: 1108676-1108676 on Build GRCh38
rs918574639 Chr.19: 1108616-1108616 on Build GRCh38
rs938903485 Chr.19: 1108604-1108604 on Build GRCh38
rs979831464 Chr.19: 1108735-1108736 on Build GRCh38
rs757314058 Chr.19: 1108652-1108652 on Build GRCh38
rs574425907 Chr.19: 1108640-1108640 on Build GRCh38
rs576784181 Chr.19: 1108561-1108561 on Build GRCh38
rs564379739 Chr.19: 1108755-1108755 on Build GRCh38
rs761001783 Chr.19: 1108717-1108717 on Build GRCh38
rs373135662 Chr.19: 1108743-1108743 on Build GRCh38
rs750461485 Chr.19: 1108746-1108746 on Build GRCh38
rs559953905 Chr.19: 1108740-1108740 on Build GRCh38
rs758988648 Chr.19: 1108716-1108716 on Build GRCh38
rs955915778 Chr.19: 1108568-1108568 on Build GRCh38
rs760470589 Chr.19: 1108731-1108731 on Build GRCh38
rs981618143 Chr.19: 1108587-1108588 on Build GRCh38
rs745967139 Chr.19: 1108673-1108673 on Build GRCh38
rs562451114 Chr.19: 1108633-1108633 on Build GRCh38
rs968471754 Chr.19: 1108718-1108718 on Build GRCh38
rs117088896 Chr.19: 1108747-1108747 on Build GRCh38
rs775591320 Chr.19: 1108678-1108678 on Build GRCh38
rs777786299 Chr.19: 1108643-1108643 on Build GRCh38
rs747280041 Chr.19: 1108769-1108770 on Build GRCh38
rs929980963 Chr.19: 1108618-1108618 on Build GRCh38
rs927543112 Chr.19: 1108601-1108601 on Build GRCh38
rs147525230 Chr.19: 1108766-1108766 on Build GRCh38
rs1038852162 Chr.19: 1108645-1108645 on Build GRCh38
rs993492164 Chr.19: 1108607-1108607 on Build GRCh38
rs762281393 Chr.19: 1108709-1108709 on Build GRCh38

View Full Product Details