Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 31484528-31484769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD7A
Entrez Gene ID
161725
Gene Name
OTU deubiquitinase 7A
Gene Aliases
C15orf16, C16ORF15, CEZANNE2, OTUD7
UniGene
Hs.355236
Gene Chromosome Location
Chr.15: 31483126-31655339 on Build GRCh38
Transcripts
NM_130901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
960421 c.1418C>T p.S473L Chr.15: 31484657-31484657 on Build GRCh38
700189 c.1419G>T p.S473S Chr.15: 31484656-31484656 on Build GRCh38
4053979 c.1525C>T p.R509C Chr.15: 31484550-31484550 on Build GRCh38
960422 c.1353G>A p.A451A Chr.15: 31484722-31484722 on Build GRCh38
6736225 c.1528G>A p.A510T Chr.15: 31484547-31484547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146885561 Chr.15: 31484548-31484548 on Build GRCh38
rs377185847 Chr.15: 31484698-31484698 on Build GRCh38
rs778782844 Chr.15: 31484636-31484636 on Build GRCh38
rs148643263 Chr.15: 31484611-31484611 on Build GRCh38
rs750201132 Chr.15: 31484746-31484746 on Build GRCh38
rs373899339 Chr.15: 31484694-31484694 on Build GRCh38
rs771670942 Chr.15: 31484585-31484590 on Build GRCh38
rs76704217 Chr.15: 31484634-31484634 on Build GRCh38
rs755681800 Chr.15: 31484621-31484621 on Build GRCh38
rs779533771 Chr.15: 31484593-31484601 on Build GRCh38
rs775372326 Chr.15: 31484575-31484575 on Build GRCh38
rs573163595 Chr.15: 31484649-31484649 on Build GRCh38
rs201934783 Chr.15: 31484731-31484731 on Build GRCh38
rs745866687 Chr.15: 31484571-31484571 on Build GRCh38
rs778727714 Chr.15: 31484745-31484745 on Build GRCh38
rs552445976 Chr.15: 31484644-31484644 on Build GRCh38
rs763725467 Chr.15: 31484735-31484735 on Build GRCh38
rs777504137 Chr.15: 31484552-31484552 on Build GRCh38
rs765651339 Chr.15: 31484598-31484598 on Build GRCh38
rs753401930 Chr.15: 31484736-31484736 on Build GRCh38
rs770011556 Chr.15: 31484689-31484689 on Build GRCh38
rs747699554 Chr.15: 31484550-31484550 on Build GRCh38
rs762093336 Chr.15: 31484685-31484685 on Build GRCh38
rs760827166 Chr.15: 31484582-31484582 on Build GRCh38
rs1036676805 Chr.15: 31484617-31484617 on Build GRCh38
rs768716767 Chr.15: 31484584-31484584 on Build GRCh38
rs149179005 Chr.15: 31484662-31484662 on Build GRCh38
rs768826447 Chr.15: 31484676-31484676 on Build GRCh38
rs371211492 Chr.15: 31484608-31484608 on Build GRCh38
rs745670428 Chr.15: 31484643-31484643 on Build GRCh38
rs958638372 Chr.15: 31484718-31484718 on Build GRCh38
rs776947158 Chr.15: 31484590-31484590 on Build GRCh38
rs763330565 Chr.15: 31484695-31484695 on Build GRCh38
rs996273208 Chr.15: 31484578-31484578 on Build GRCh38
rs776708470 Chr.15: 31484677-31484677 on Build GRCh38
rs745486596 Chr.15: 31484584-31484586 on Build GRCh38
rs772137593 Chr.15: 31484572-31484572 on Build GRCh38
rs777260675 Chr.15: 31484624-31484624 on Build GRCh38
rs377421686 Chr.15: 31484599-31484599 on Build GRCh38
rs753711730 Chr.15: 31484558-31484578 on Build GRCh38
rs756910731 Chr.15: 31484744-31484744 on Build GRCh38
rs770898697 Chr.15: 31484570-31484570 on Build GRCh38
rs752148499 Chr.15: 31484708-31484708 on Build GRCh38
rs756964120 Chr.15: 31484560-31484562 on Build GRCh38
rs746449396 Chr.15: 31484603-31484611 on Build GRCh38
rs747026174 Chr.15: 31484674-31484674 on Build GRCh38
rs760089268 Chr.15: 31484722-31484722 on Build GRCh38
rs139774758 Chr.15: 31484560-31484560 on Build GRCh38
rs752101108 Chr.15: 31484612-31484612 on Build GRCh38
rs778669864 Chr.15: 31484578-31484580 on Build GRCh38
rs762148134 Chr.15: 31484593-31484593 on Build GRCh38
rs755665876 Chr.15: 31484551-31484551 on Build GRCh38
rs753602457 Chr.15: 31484628-31484628 on Build GRCh38

View Full Product Details