Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39267385-39267636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536784.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021323.1 XM_017021324.1 XM_017021325.1 XM_017021326.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844283 c.45C>T p.L15L Chr.14: 39267504-39267504 on Build GRCh38
955688 c.51G>T p.E17D Chr.14: 39267510-39267510 on Build GRCh38
339611 c.49G>T p.E17* Chr.14: 39267508-39267508 on Build GRCh38
4844284 c.45C>T p.L15L Chr.14: 39267504-39267504 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777798508 Chr.14: 39267572-39267572 on Build GRCh38
rs748086345 Chr.14: 39267428-39267428 on Build GRCh38
rs751260971 Chr.14: 39267555-39267555 on Build GRCh38
rs7140561 Chr.14: 39267476-39267476 on Build GRCh38
rs761064170 Chr.14: 39267409-39267409 on Build GRCh38
rs757396862 Chr.14: 39267414-39267415 on Build GRCh38
rs770344379 Chr.14: 39267530-39267530 on Build GRCh38
rs773541449 Chr.14: 39267535-39267535 on Build GRCh38
rs372657433 Chr.14: 39267416-39267416 on Build GRCh38
rs1048329668 Chr.14: 39267575-39267575 on Build GRCh38
rs762013883 Chr.14: 39267528-39267528 on Build GRCh38
rs772438716 Chr.14: 39267443-39267443 on Build GRCh38
rs573151225 Chr.14: 39267615-39267615 on Build GRCh38
rs17855895 Chr.14: 39267490-39267490 on Build GRCh38
rs774485080 Chr.14: 39267435-39267435 on Build GRCh38
rs763145286 Chr.14: 39267468-39267468 on Build GRCh38
rs752179503 Chr.14: 39267497-39267497 on Build GRCh38
rs1015680877 Chr.14: 39267614-39267614 on Build GRCh38
rs932459866 Chr.14: 39267546-39267546 on Build GRCh38
rs781018209 Chr.14: 39267487-39267487 on Build GRCh38
rs772241512 Chr.14: 39267500-39267500 on Build GRCh38
rs745869917 Chr.14: 39267442-39267442 on Build GRCh38
rs762372630 Chr.14: 39267470-39267470 on Build GRCh38
rs764722518 Chr.14: 39267410-39267410 on Build GRCh38
rs376780966 Chr.14: 39267417-39267417 on Build GRCh38
rs756761883 Chr.14: 39267413-39267413 on Build GRCh38
rs755817221 Chr.14: 39267566-39267566 on Build GRCh38
rs777515592 Chr.14: 39267493-39267493 on Build GRCh38
rs368948446 Chr.14: 39267423-39267423 on Build GRCh38
rs758037941 Chr.14: 39267418-39267418 on Build GRCh38
rs202175054 Chr.14: 39267460-39267460 on Build GRCh38
rs1007542706 Chr.14: 39267570-39267570 on Build GRCh38
rs557160691 Chr.14: 39267429-39267429 on Build GRCh38
rs764716632 Chr.14: 39267472-39267472 on Build GRCh38
rs747215161 Chr.14: 39267516-39267516 on Build GRCh38
rs762155247 Chr.14: 39267537-39267537 on Build GRCh38
rs377313525 Chr.14: 39267436-39267436 on Build GRCh38
rs754333482 Chr.14: 39267411-39267411 on Build GRCh38
rs746353205 Chr.14: 39267608-39267608 on Build GRCh38
rs754807691 Chr.14: 39267481-39267481 on Build GRCh38
rs759175723 Chr.14: 39267556-39267556 on Build GRCh38
rs1052256828 Chr.14: 39267539-39267539 on Build GRCh38
rs376664088 Chr.14: 39267544-39267544 on Build GRCh38
rs113934008 Chr.14: 39267551-39267551 on Build GRCh38
rs753798108 Chr.14: 39267574-39267574 on Build GRCh38
rs757843486 Chr.14: 39267475-39267475 on Build GRCh38
rs779833957 Chr.14: 39267494-39267495 on Build GRCh38
rs780467159 Chr.14: 39267501-39267501 on Build GRCh38
rs767011478 Chr.14: 39267557-39267557 on Build GRCh38
rs751171248 Chr.14: 39267480-39267480 on Build GRCh38
rs781021576 Chr.14: 39267424-39267424 on Build GRCh38
rs777045235 Chr.14: 39267518-39267518 on Build GRCh38
rs34909359 Chr.14: 39267489-39267489 on Build GRCh38
rs761155808 Chr.14: 39267459-39267459 on Build GRCh38
rs752551102 Chr.14: 39267558-39267558 on Build GRCh38
rs369514422 Chr.14: 39267548-39267548 on Build GRCh38
rs775808300 Chr.14: 39267456-39267456 on Build GRCh38
rs889127210 Chr.14: 39267540-39267540 on Build GRCh38
rs542282969 Chr.14: 39267616-39267616 on Build GRCh38
rs751537355 Chr.14: 39267546-39267547 on Build GRCh38
rs1003923861 Chr.14: 39267612-39267612 on Build GRCh38
rs942243918 Chr.14: 39267486-39267486 on Build GRCh38
rs779611509 Chr.14: 39267421-39267421 on Build GRCh38
rs560005506 Chr.14: 39267579-39267579 on Build GRCh38

View Full Product Details