Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55143442-55143715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20B
Entrez Gene ID
166979
Gene Name
cell division cycle 20B
Gene Aliases
G6VTS76519
UniGene
Hs.669184
Gene Chromosome Location
Chr.5: 55112971-55173177 on Build GRCh38
Transcripts
NM_001145734.2 NM_001170402.1 NM_152623.2 XM_011543218.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422854 c.433G>T p.A145S Chr.5: 55143566-55143566 on Build GRCh38
5632424 c.357C>G p.L119L Chr.5: 55143579-55143579 on Build GRCh38
4546060 c.391G>A p.G131R Chr.5: 55143608-55143608 on Build GRCh38
6422855 c.433G>T p.A145S Chr.5: 55143566-55143566 on Build GRCh38
5001473 c.298C>T p.R100C Chr.5: 55143638-55143638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200031751 Chr.5: 55143671-55143671 on Build GRCh38
rs891457221 Chr.5: 55143551-55143551 on Build GRCh38
rs746251568 Chr.5: 55143508-55143508 on Build GRCh38
rs537555928 Chr.5: 55143674-55143674 on Build GRCh38
rs140643849 Chr.5: 55143514-55143514 on Build GRCh38
rs777690639 Chr.5: 55143535-55143535 on Build GRCh38
rs754389124 Chr.5: 55143485-55143485 on Build GRCh38
rs962872440 Chr.5: 55143651-55143651 on Build GRCh38
rs113527284 Chr.5: 55143658-55143658 on Build GRCh38
rs746754909 Chr.5: 55143543-55143543 on Build GRCh38
rs781197249 Chr.5: 55143673-55143673 on Build GRCh38
rs191396 Chr.5: 55143685-55143685 on Build GRCh38
rs148160140 Chr.5: 55143571-55143571 on Build GRCh38
rs764625402 Chr.5: 55143647-55143647 on Build GRCh38
rs199820748 Chr.5: 55143511-55143511 on Build GRCh38
rs370579774 Chr.5: 55143676-55143676 on Build GRCh38
rs963187466 Chr.5: 55143602-55143602 on Build GRCh38
rs752303866 Chr.5: 55143654-55143654 on Build GRCh38
rs578175738 Chr.5: 55143648-55143651 on Build GRCh38
rs776135130 Chr.5: 55143628-55143628 on Build GRCh38
rs775678600 Chr.5: 55143567-55143567 on Build GRCh38
rs753494760 Chr.5: 55143482-55143482 on Build GRCh38
rs1021580 Chr.5: 55143638-55143638 on Build GRCh38
rs751554659 Chr.5: 55143662-55143662 on Build GRCh38
rs373422364 Chr.5: 55143553-55143553 on Build GRCh38
rs936904047 Chr.5: 55143518-55143518 on Build GRCh38
rs746154788 Chr.5: 55143557-55143557 on Build GRCh38
rs762396033 Chr.5: 55143656-55143656 on Build GRCh38
rs769101869 Chr.5: 55143569-55143569 on Build GRCh38
rs781503724 Chr.5: 55143555-55143555 on Build GRCh38
rs145816473 Chr.5: 55143522-55143522 on Build GRCh38
rs762101309 Chr.5: 55143593-55143593 on Build GRCh38
rs769754299 Chr.5: 55143566-55143566 on Build GRCh38
rs765976968 Chr.5: 55143470-55143470 on Build GRCh38
rs767883088 Chr.5: 55143619-55143619 on Build GRCh38
rs756024821 Chr.5: 55143525-55143525 on Build GRCh38

View Full Product Details