Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138459850-138460123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT3
Entrez Gene ID
83850
Gene Name
extended synaptotagmin 3
Gene Aliases
CHR3SYT, E-Syt3, FAM62C
UniGene
Hs.477711
Gene Chromosome Location
Chr.3: 138434573-138479942 on Build GRCh38
Transcripts
NM_001322831.1 NM_001322834.1 NM_031913.4 NR_136409.1 XM_017007307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3123941 c.654G>T p.Q218H Chr.3: 138459950-138459950 on Build GRCh38
4853814 c.730C>T p.Q244* Chr.3: 138460026-138460026 on Build GRCh38
1419434 c.712G>A p.V238M Chr.3: 138460008-138460008 on Build GRCh38
3746609 c.699C>T p.P233P Chr.3: 138459995-138459995 on Build GRCh38
6360545 c.731A>G p.Q244R Chr.3: 138460027-138460027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369447242 Chr.3: 138460053-138460053 on Build GRCh38
rs774608302 Chr.3: 138459999-138459999 on Build GRCh38
rs200379323 Chr.3: 138460034-138460034 on Build GRCh38
rs566430854 Chr.3: 138460007-138460007 on Build GRCh38
rs767744617 Chr.3: 138459902-138459902 on Build GRCh38
rs547266807 Chr.3: 138459872-138459872 on Build GRCh38
rs754188163 Chr.3: 138460027-138460027 on Build GRCh38
rs766409704 Chr.3: 138459994-138459994 on Build GRCh38
rs747919238 Chr.3: 138459961-138459961 on Build GRCh38
rs774379141 Chr.3: 138460062-138460062 on Build GRCh38
rs757917247 Chr.3: 138459930-138459930 on Build GRCh38
rs200676435 Chr.3: 138460003-138460003 on Build GRCh38
rs369097151 Chr.3: 138459886-138459886 on Build GRCh38
rs772755808 Chr.3: 138460042-138460042 on Build GRCh38
rs773289775 Chr.3: 138459977-138459977 on Build GRCh38
rs767564947 Chr.3: 138460070-138460070 on Build GRCh38
rs753049610 Chr.3: 138459904-138459904 on Build GRCh38
rs147568505 Chr.3: 138459957-138459957 on Build GRCh38
rs34165332 Chr.3: 138459942-138459943 on Build GRCh38
rs769593339 Chr.3: 138459969-138459969 on Build GRCh38
rs556894650 Chr.3: 138460094-138460094 on Build GRCh38
rs761014130 Chr.3: 138460076-138460076 on Build GRCh38
rs373151618 Chr.3: 138460078-138460078 on Build GRCh38
rs746092634 Chr.3: 138459939-138459939 on Build GRCh38
rs561184666 Chr.3: 138459885-138459885 on Build GRCh38
rs866981946 Chr.3: 138460002-138460002 on Build GRCh38
rs534760132 Chr.3: 138460066-138460066 on Build GRCh38
rs529902700 Chr.3: 138459915-138459915 on Build GRCh38
rs377114164 Chr.3: 138460081-138460081 on Build GRCh38
rs774997752 Chr.3: 138460046-138460046 on Build GRCh38
rs554963409 Chr.3: 138459950-138459950 on Build GRCh38
rs17857138 Chr.3: 138460033-138460033 on Build GRCh38
rs62282904 Chr.3: 138459960-138459960 on Build GRCh38
rs764384371 Chr.3: 138460020-138460020 on Build GRCh38
rs887686356 Chr.3: 138459982-138459982 on Build GRCh38
rs756437673 Chr.3: 138460014-138460014 on Build GRCh38
rs775587089 Chr.3: 138460072-138460072 on Build GRCh38
rs767333420 Chr.3: 138459932-138459933 on Build GRCh38
rs369827819 Chr.3: 138459923-138459923 on Build GRCh38
rs955840428 Chr.3: 138460084-138460084 on Build GRCh38
rs746551985 Chr.3: 138460035-138460035 on Build GRCh38
rs746518686 Chr.3: 138459943-138459943 on Build GRCh38
rs144720494 Chr.3: 138460049-138460049 on Build GRCh38
rs535008689 Chr.3: 138459910-138459910 on Build GRCh38
rs762930087 Chr.3: 138459991-138459991 on Build GRCh38
rs752923089 Chr.3: 138460008-138460008 on Build GRCh38
rs749056558 Chr.3: 138460058-138460058 on Build GRCh38
rs771037756 Chr.3: 138460059-138460059 on Build GRCh38
rs769493506 Chr.3: 138460052-138460052 on Build GRCh38
rs552234007 Chr.3: 138460045-138460045 on Build GRCh38
rs781734471 Chr.3: 138459895-138459895 on Build GRCh38

View Full Product Details