Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 4711860-4712085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJAP1
Entrez Gene ID
55966
Gene Name
adherens junctions associated protein 1
Gene Aliases
MOT8, SHREW-1, SHREW1
UniGene
Hs.12473
Gene Chromosome Location
Chr.1: 4654775-4792518 on Build GRCh38
Transcripts
NM_001042478.1 NM_018836.3 XM_011541786.2 XM_011541787.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4660687 c.65G>T p.G22V Chr.1: 4711935-4711935 on Build GRCh38
6127031 c.164G>C p.R55P Chr.1: 4712034-4712034 on Build GRCh38
6127032 c.133G>T p.G45C Chr.1: 4712003-4712003 on Build GRCh38
6063760 c.179C>T p.P60L Chr.1: 4712049-4712049 on Build GRCh38
5352851 c.184C>T p.R62W Chr.1: 4712054-4712054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750399307 Chr.1: 4711908-4711908 on Build GRCh38
rs763904972 Chr.1: 4712036-4712036 on Build GRCh38
rs544174805 Chr.1: 4711928-4711928 on Build GRCh38
rs761363510 Chr.1: 4711889-4711889 on Build GRCh38
rs776421160 Chr.1: 4712014-4712014 on Build GRCh38
rs758994527 Chr.1: 4712057-4712057 on Build GRCh38
rs532147012 Chr.1: 4712038-4712038 on Build GRCh38
rs369969783 Chr.1: 4711954-4711954 on Build GRCh38
rs760335322 Chr.1: 4711951-4711951 on Build GRCh38
rs768777082 Chr.1: 4712038-4712046 on Build GRCh38
rs559269335 Chr.1: 4711886-4711887 on Build GRCh38
rs755006298 Chr.1: 4711893-4711893 on Build GRCh38
rs771513181 Chr.1: 4712001-4712001 on Build GRCh38
rs987473812 Chr.1: 4711913-4711913 on Build GRCh38
rs746497364 Chr.1: 4712009-4712009 on Build GRCh38
rs201311316 Chr.1: 4711891-4711891 on Build GRCh38
rs770767997 Chr.1: 4712013-4712013 on Build GRCh38
rs747676999 Chr.1: 4711934-4711934 on Build GRCh38
rs200234894 Chr.1: 4711892-4711892 on Build GRCh38
rs753991951 Chr.1: 4712049-4712049 on Build GRCh38
rs775308984 Chr.1: 4712026-4712026 on Build GRCh38
rs771542278 Chr.1: 4711943-4711943 on Build GRCh38
rs1061968 Chr.1: 4711993-4711993 on Build GRCh38
rs748186917 Chr.1: 4712049-4712050 on Build GRCh38
rs759413505 Chr.1: 4712015-4712015 on Build GRCh38
rs373570588 Chr.1: 4711890-4711890 on Build GRCh38
rs148753165 Chr.1: 4711990-4711990 on Build GRCh38
rs781277874 Chr.1: 4711920-4711920 on Build GRCh38
rs778132143 Chr.1: 4712059-4712059 on Build GRCh38
rs538795214 Chr.1: 4711923-4711923 on Build GRCh38
rs779291510 Chr.1: 4711994-4711994 on Build GRCh38
rs778936687 Chr.1: 4712037-4712038 on Build GRCh38
rs566618227 Chr.1: 4711921-4711921 on Build GRCh38
rs750637126 Chr.1: 4711982-4711982 on Build GRCh38
rs967579755 Chr.1: 4711894-4711894 on Build GRCh38
rs755348555 Chr.1: 4712051-4712051 on Build GRCh38
rs551223535 Chr.1: 4711888-4711888 on Build GRCh38
rs1055255127 Chr.1: 4712011-4712011 on Build GRCh38
rs772763050 Chr.1: 4711949-4711949 on Build GRCh38
rs766652336 Chr.1: 4712043-4712043 on Build GRCh38
rs777384081 Chr.1: 4712003-4712003 on Build GRCh38
rs137881519 Chr.1: 4711902-4711902 on Build GRCh38
rs766099782 Chr.1: 4711959-4711959 on Build GRCh38
rs199654684 Chr.1: 4711976-4711976 on Build GRCh38
rs770127409 Chr.1: 4711886-4711886 on Build GRCh38
rs372616300 Chr.1: 4711987-4711987 on Build GRCh38
rs762904463 Chr.1: 4712033-4712033 on Build GRCh38
rs369982143 Chr.1: 4712030-4712030 on Build GRCh38
rs751652866 Chr.1: 4711897-4711897 on Build GRCh38
rs1021920011 Chr.1: 4712052-4712060 on Build GRCh38
rs746288379 Chr.1: 4711882-4711882 on Build GRCh38
rs757356256 Chr.1: 4711914-4711914 on Build GRCh38
rs749501577 Chr.1: 4711887-4711887 on Build GRCh38
rs765335332 Chr.1: 4711975-4711975 on Build GRCh38
rs779296700 Chr.1: 4712053-4712053 on Build GRCh38
rs146622911 Chr.1: 4712000-4712000 on Build GRCh38
rs138837601 Chr.1: 4711926-4711926 on Build GRCh38
rs780226118 Chr.1: 4711989-4711989 on Build GRCh38
rs774907745 Chr.1: 4711931-4711931 on Build GRCh38
rs775986901 Chr.1: 4711887-4711887 on Build GRCh38
rs868308712 Chr.1: 4711919-4711919 on Build GRCh38
rs752973756 Chr.1: 4712054-4712054 on Build GRCh38
rs376909909 Chr.1: 4712041-4712041 on Build GRCh38
rs776621169 Chr.1: 4711969-4711969 on Build GRCh38
rs149284997 Chr.1: 4711974-4711974 on Build GRCh38
rs975119371 Chr.1: 4711970-4711970 on Build GRCh38
rs769658115 Chr.1: 4712024-4712024 on Build GRCh38

View Full Product Details