Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17793411-17793676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf91
Entrez Gene ID
54149
Gene Name
chromosome 21 open reading frame 91
Gene Aliases
C21orf14, C21orf38, CSSG1, EURL, YG81
UniGene
Hs.293811
Gene Chromosome Location
Chr.21: 17788967-17819386 on Build GRCh38
Transcripts
NM_001100420.1 NM_001100421.1 NM_017447.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5633706 c.883_884insC p.I295fs*>4 Chr.21: 17793425-17793426 on Build GRCh38
3423867 c.808G>A p.E270K Chr.21: 17793501-17793501 on Build GRCh38
5687867 c.819G>C p.L273L Chr.21: 17793490-17793490 on Build GRCh38
1029531 c.888C>T p.N296N Chr.21: 17793421-17793421 on Build GRCh38
1029532 c.807C>T p.I269I Chr.21: 17793502-17793502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17002088 Chr.21: 17793478-17793478 on Build GRCh38
rs375675833 Chr.21: 17793524-17793524 on Build GRCh38
rs367786519 Chr.21: 17793614-17793614 on Build GRCh38
rs201559190 Chr.21: 17793516-17793516 on Build GRCh38
rs371460924 Chr.21: 17793624-17793624 on Build GRCh38
rs761326149 Chr.21: 17793631-17793637 on Build GRCh38
rs769395784 Chr.21: 17793599-17793599 on Build GRCh38
rs776742595 Chr.21: 17793626-17793626 on Build GRCh38
rs772445287 Chr.21: 17793453-17793453 on Build GRCh38
rs780460019 Chr.21: 17793532-17793532 on Build GRCh38
rs779616662 Chr.21: 17793459-17793459 on Build GRCh38
rs776282923 Chr.21: 17793479-17793479 on Build GRCh38
rs535145078 Chr.21: 17793605-17793605 on Build GRCh38
rs765697639 Chr.21: 17793520-17793520 on Build GRCh38
rs770699669 Chr.21: 17793617-17793617 on Build GRCh38
rs773159450 Chr.21: 17793486-17793486 on Build GRCh38
rs752922725 Chr.21: 17793510-17793510 on Build GRCh38
rs779422028 Chr.21: 17793448-17793448 on Build GRCh38
rs75916355 Chr.21: 17793501-17793501 on Build GRCh38
rs376996901 Chr.21: 17793511-17793511 on Build GRCh38
rs530934931 Chr.21: 17793606-17793606 on Build GRCh38
rs764548654 Chr.21: 17793639-17793639 on Build GRCh38
rs76915028 Chr.21: 17793562-17793562 on Build GRCh38
rs902212087 Chr.21: 17793545-17793545 on Build GRCh38
rs757283460 Chr.21: 17793519-17793519 on Build GRCh38
rs373599266 Chr.21: 17793436-17793436 on Build GRCh38
rs2824493 Chr.21: 17793625-17793625 on Build GRCh38
rs761923183 Chr.21: 17793631-17793631 on Build GRCh38
rs200103095 Chr.21: 17793502-17793502 on Build GRCh38
rs370784680 Chr.21: 17793539-17793539 on Build GRCh38
rs374793972 Chr.21: 17793542-17793542 on Build GRCh38
rs550343756 Chr.21: 17793615-17793615 on Build GRCh38
rs746245395 Chr.21: 17793452-17793452 on Build GRCh38
rs747690893 Chr.21: 17793580-17793580 on Build GRCh38
rs764235279 Chr.21: 17793515-17793515 on Build GRCh38
rs746657286 Chr.21: 17793460-17793460 on Build GRCh38
rs769873891 Chr.21: 17793483-17793483 on Build GRCh38
rs377549836 Chr.21: 17793527-17793527 on Build GRCh38
rs551741122 Chr.21: 17793437-17793437 on Build GRCh38
rs750913699 Chr.21: 17793522-17793522 on Build GRCh38
rs757766192 Chr.21: 17793439-17793439 on Build GRCh38

View Full Product Details