Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83819676-83819919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL3
Entrez Gene ID
57188
Gene Name
ADAMTS like 3
Gene Aliases
ADAMTSL-3
UniGene
Hs.459162
Gene Chromosome Location
Chr.15: 83654086-84039842 on Build GRCh38
Transcripts
NM_001301110.1 NM_207517.2 XM_011521822.2 XM_011521823.2 XM_011521824.2 XM_011521825.2 XM_017022434.1 XM_017022435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5832961 c.390C>T p.F130F Chr.15: 83819837-83819837 on Build GRCh38
6875540 c.430C>T p.Q144* Chr.15: 83819877-83819877 on Build GRCh38
6875539 c.430C>T p.Q144* Chr.15: 83819877-83819877 on Build GRCh38
248761 c.457C>T p.R153* Chr.15: 83819904-83819904 on Build GRCh38
5302736 c.458G>A p.R153Q Chr.15: 83819905-83819905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888901939 Chr.15: 83819716-83819720 on Build GRCh38
rs4583200 Chr.15: 83819777-83819777 on Build GRCh38
rs144671764 Chr.15: 83819883-83819883 on Build GRCh38
rs778035458 Chr.15: 83819831-83819831 on Build GRCh38
rs766634485 Chr.15: 83819797-83819797 on Build GRCh38
rs761164415 Chr.15: 83819793-83819793 on Build GRCh38
rs776009651 Chr.15: 83819764-83819765 on Build GRCh38
rs779662187 Chr.15: 83819849-83819849 on Build GRCh38
rs1040105848 Chr.15: 83819717-83819717 on Build GRCh38
rs769675062 Chr.15: 83819847-83819847 on Build GRCh38
rs142393908 Chr.15: 83819825-83819825 on Build GRCh38
rs563672707 Chr.15: 83819728-83819728 on Build GRCh38
rs754391127 Chr.15: 83819800-83819800 on Build GRCh38
rs888025123 Chr.15: 83819751-83819751 on Build GRCh38
rs774420051 Chr.15: 83819881-83819881 on Build GRCh38
rs189771247 Chr.15: 83819791-83819791 on Build GRCh38
rs763658711 Chr.15: 83819828-83819830 on Build GRCh38
rs144806194 Chr.15: 83819803-83819804 on Build GRCh38
rs148209946 Chr.15: 83819776-83819776 on Build GRCh38
rs200762628 Chr.15: 83819770-83819770 on Build GRCh38
rs779056239 Chr.15: 83819818-83819818 on Build GRCh38
rs909433857 Chr.15: 83819792-83819792 on Build GRCh38
rs747201110 Chr.15: 83819839-83819839 on Build GRCh38
rs775537591 Chr.15: 83819775-83819775 on Build GRCh38
rs771681530 Chr.15: 83819891-83819891 on Build GRCh38
rs1037050345 Chr.15: 83819843-83819843 on Build GRCh38
rs888875631 Chr.15: 83819880-83819880 on Build GRCh38
rs768408281 Chr.15: 83819861-83819861 on Build GRCh38
rs376774419 Chr.15: 83819827-83819827 on Build GRCh38
rs181914721 Chr.15: 83819856-83819856 on Build GRCh38
rs746990818 Chr.15: 83819844-83819844 on Build GRCh38
rs761558842 Chr.15: 83819782-83819782 on Build GRCh38
rs992194041 Chr.15: 83819744-83819744 on Build GRCh38
rs897511024 Chr.15: 83819722-83819722 on Build GRCh38
rs755400208 Chr.15: 83819808-83819808 on Build GRCh38
rs4483821 Chr.15: 83819884-83819884 on Build GRCh38
rs944446812 Chr.15: 83819716-83819716 on Build GRCh38
rs933013663 Chr.15: 83819852-83819852 on Build GRCh38

View Full Product Details