Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50482798-50483039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM2
Entrez Gene ID
79924
Gene Name
adrenomedullin 2
Gene Aliases
AM2, dJ579N16.4
UniGene
Hs.449099
Gene Chromosome Location
Chr.22: 50481724-50486440 on Build GRCh38
Transcripts
NM_001253845.1 XM_017028940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479129 c.391G>A p.A131T Chr.22: 50482847-50482847 on Build GRCh38
4639127 c.351G>A p.V117V Chr.22: 50482807-50482807 on Build GRCh38
6595253 c.198G>T p.E66D Chr.22: 50482904-50482904 on Build GRCh38
3708340 c.103A>G p.R35G Chr.22: 50482809-50482809 on Build GRCh38
7354604 c.397C>T p.R133W Chr.22: 50482853-50482853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746081346 Chr.22: 50482951-50482951 on Build GRCh38
rs766321313 Chr.22: 50482908-50482908 on Build GRCh38
rs758981620 Chr.22: 50482893-50482893 on Build GRCh38
rs61732427 Chr.22: 50482954-50482954 on Build GRCh38
rs930463930 Chr.22: 50482859-50482859 on Build GRCh38
rs374496738 Chr.22: 50482849-50482849 on Build GRCh38
rs770576228 Chr.22: 50482997-50482997 on Build GRCh38
rs764410445 Chr.22: 50482928-50482928 on Build GRCh38
rs956348244 Chr.22: 50483015-50483015 on Build GRCh38
rs537686058 Chr.22: 50482992-50482992 on Build GRCh38
rs768112003 Chr.22: 50482911-50482922 on Build GRCh38
rs779222469 Chr.22: 50482947-50482947 on Build GRCh38
rs987647903 Chr.22: 50483017-50483017 on Build GRCh38
rs762543576 Chr.22: 50482878-50482878 on Build GRCh38
rs536506636 Chr.22: 50482854-50482854 on Build GRCh38
rs757445676 Chr.22: 50482941-50482941 on Build GRCh38
rs755497451 Chr.22: 50482824-50482824 on Build GRCh38
rs1040885840 Chr.22: 50482934-50482934 on Build GRCh38
rs753066827 Chr.22: 50482826-50482826 on Build GRCh38
rs555246517 Chr.22: 50482891-50482891 on Build GRCh38
rs756622969 Chr.22: 50482926-50482926 on Build GRCh38
rs1000400122 Chr.22: 50482969-50482969 on Build GRCh38
rs768717029 Chr.22: 50482853-50482853 on Build GRCh38
rs749525166 Chr.22: 50482847-50482847 on Build GRCh38
rs886382051 Chr.22: 50482873-50482873 on Build GRCh38
rs756082631 Chr.22: 50482986-50482986 on Build GRCh38
rs199634529 Chr.22: 50482876-50482876 on Build GRCh38
rs538224252 Chr.22: 50482846-50482846 on Build GRCh38
rs754194801 Chr.22: 50482931-50482931 on Build GRCh38
rs565351807 Chr.22: 50482851-50482851 on Build GRCh38
rs1026984558 Chr.22: 50482978-50482978 on Build GRCh38
rs759054465 Chr.22: 50482975-50482975 on Build GRCh38
rs969307239 Chr.22: 50482957-50482957 on Build GRCh38
rs767012085 Chr.22: 50482912-50482912 on Build GRCh38
rs761792725 Chr.22: 50482863-50482863 on Build GRCh38
rs748822493 Chr.22: 50482844-50482847 on Build GRCh38
rs769729265 Chr.22: 50482955-50482955 on Build GRCh38
rs765846835 Chr.22: 50482818-50482818 on Build GRCh38
rs201372986 Chr.22: 50482845-50482845 on Build GRCh38
rs781622119 Chr.22: 50482825-50482825 on Build GRCh38
rs758606241 Chr.22: 50482953-50482953 on Build GRCh38
rs1056178348 Chr.22: 50482883-50482883 on Build GRCh38
rs1019489500 Chr.22: 50482906-50482906 on Build GRCh38
rs759177930 Chr.22: 50483016-50483016 on Build GRCh38
rs751029879 Chr.22: 50482823-50482823 on Build GRCh38
rs868203547 Chr.22: 50482889-50482889 on Build GRCh38
rs769522164 Chr.22: 50482870-50482870 on Build GRCh38
rs753261161 Chr.22: 50482916-50482916 on Build GRCh38
rs546959903 Chr.22: 50482850-50482850 on Build GRCh38
rs889720378 Chr.22: 50482927-50482927 on Build GRCh38
rs774057338 Chr.22: 50483006-50483006 on Build GRCh38
rs765964730 Chr.22: 50482887-50482887 on Build GRCh38

View Full Product Details