Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178478332-178478605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP7
Entrez Gene ID
51661
Gene Name
FK506 binding protein 7
Gene Aliases
FKBP23, PPIase
UniGene
Hs.410378
Gene Chromosome Location
Chr.2: 178463664-178479878 on Build GRCh38
Transcripts
NM_001135212.1 NM_181342.2 XM_005246638.4 XM_011511348.2 XM_011511349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5860690 c.10delA p.T4fs*32 Chr.2: 178478490-178478490 on Build GRCh38
5736271 c.10A>C p.T4P Chr.2: 178478490-178478490 on Build GRCh38
256965 c.21C>A p.F7L Chr.2: 178478479-178478479 on Build GRCh38
441551 c.74G>C p.R25T Chr.2: 178478426-178478426 on Build GRCh38
418467 c.160G>A p.D54N Chr.2: 178478340-178478340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913055087 Chr.2: 178478567-178478567 on Build GRCh38
rs749083324 Chr.2: 178478532-178478533 on Build GRCh38
rs761931069 Chr.2: 178478401-178478401 on Build GRCh38
rs1041953761 Chr.2: 178478461-178478461 on Build GRCh38
rs777028882 Chr.2: 178478410-178478410 on Build GRCh38
rs576741932 Chr.2: 178478584-178478584 on Build GRCh38
rs769021295 Chr.2: 178478495-178478495 on Build GRCh38
rs765432146 Chr.2: 178478408-178478408 on Build GRCh38
rs553608533 Chr.2: 178478575-178478575 on Build GRCh38
rs961892183 Chr.2: 178478559-178478559 on Build GRCh38
rs188660897 Chr.2: 178478566-178478566 on Build GRCh38
rs376048035 Chr.2: 178478458-178478458 on Build GRCh38
rs775904974 Chr.2: 178478558-178478558 on Build GRCh38
rs141588274 Chr.2: 178478454-178478454 on Build GRCh38
rs537383828 Chr.2: 178478467-178478467 on Build GRCh38
rs758604102 Chr.2: 178478386-178478392 on Build GRCh38
rs763108214 Chr.2: 178478525-178478525 on Build GRCh38
rs992637590 Chr.2: 178478564-178478564 on Build GRCh38
rs779729812 Chr.2: 178478440-178478440 on Build GRCh38
rs754906705 Chr.2: 178478443-178478443 on Build GRCh38
rs781233373 Chr.2: 178478444-178478444 on Build GRCh38
rs369058153 Chr.2: 178478465-178478465 on Build GRCh38
rs748097795 Chr.2: 178478414-178478417 on Build GRCh38
rs556037155 Chr.2: 178478515-178478515 on Build GRCh38
rs780155338 Chr.2: 178478390-178478391 on Build GRCh38
rs753882588 Chr.2: 178478399-178478399 on Build GRCh38
rs766160356 Chr.2: 178478547-178478547 on Build GRCh38
rs574620893 Chr.2: 178478420-178478423 on Build GRCh38
rs776005760 Chr.2: 178478375-178478375 on Build GRCh38
rs770277809 Chr.2: 178478466-178478466 on Build GRCh38
rs761140719 Chr.2: 178478490-178478490 on Build GRCh38
rs759872502 Chr.2: 178478452-178478452 on Build GRCh38
rs777511373 Chr.2: 178478454-178478458 on Build GRCh38
rs761877985 Chr.2: 178478505-178478505 on Build GRCh38
rs570372324 Chr.2: 178478457-178478457 on Build GRCh38
rs1006454145 Chr.2: 178478538-178478538 on Build GRCh38
rs754856174 Chr.2: 178478548-178478548 on Build GRCh38
rs750712715 Chr.2: 178478415-178478415 on Build GRCh38
rs758147503 Chr.2: 178478434-178478434 on Build GRCh38
rs139058251 Chr.2: 178478475-178478475 on Build GRCh38
rs764193393 Chr.2: 178478397-178478397 on Build GRCh38
rs372754221 Chr.2: 178478585-178478585 on Build GRCh38
rs574435335 Chr.2: 178478521-178478521 on Build GRCh38
rs751467891 Chr.2: 178478442-178478442 on Build GRCh38
rs61729158 Chr.2: 178478356-178478356 on Build GRCh38
rs777180461 Chr.2: 178478503-178478503 on Build GRCh38

View Full Product Details