Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53078253-53078498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PODN
Entrez Gene ID
127435
Gene Name
podocan
Gene Aliases
PCAN, SLRR5A
UniGene
Hs.586141
Gene Chromosome Location
Chr.1: 53062052-53085502 on Build GRCh38
Transcripts
NM_153703.4 XM_005270457.3 XM_011540677.2 XM_017000294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426453 c.1121C>T p.A374V Chr.1: 53078487-53078487 on Build GRCh38
4797963 c.1056C>T p.S352S Chr.1: 53078479-53078479 on Build GRCh38
6533699 c.1007C>G p.S336C Chr.1: 53078373-53078373 on Build GRCh38
5822733 c.1096A>C p.K366Q Chr.1: 53078462-53078462 on Build GRCh38
3771775 c.1119C>T p.D373D Chr.1: 53078485-53078485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141453355 Chr.1: 53078446-53078446 on Build GRCh38
rs767340203 Chr.1: 53078425-53078425 on Build GRCh38
rs761572652 Chr.1: 53078423-53078423 on Build GRCh38
rs368209286 Chr.1: 53078416-53078416 on Build GRCh38
rs78516010 Chr.1: 53078436-53078436 on Build GRCh38
rs571754220 Chr.1: 53078286-53078286 on Build GRCh38
rs766374787 Chr.1: 53078430-53078430 on Build GRCh38
rs778210043 Chr.1: 53078447-53078447 on Build GRCh38
rs773744393 Chr.1: 53078317-53078317 on Build GRCh38
rs199677246 Chr.1: 53078412-53078412 on Build GRCh38
rs373726286 Chr.1: 53078435-53078435 on Build GRCh38
rs565662939 Chr.1: 53078398-53078398 on Build GRCh38
rs139549109 Chr.1: 53078393-53078393 on Build GRCh38
rs774910985 Chr.1: 53078327-53078327 on Build GRCh38
rs762409875 Chr.1: 53078415-53078415 on Build GRCh38
rs757792597 Chr.1: 53078468-53078468 on Build GRCh38
rs570086268 Chr.1: 53078355-53078355 on Build GRCh38
rs751297394 Chr.1: 53078329-53078329 on Build GRCh38
rs755277850 Chr.1: 53078359-53078359 on Build GRCh38
rs772022703 Chr.1: 53078330-53078330 on Build GRCh38
rs183323876 Chr.1: 53078479-53078479 on Build GRCh38
rs199647870 Chr.1: 53078350-53078350 on Build GRCh38
rs551163174 Chr.1: 53078340-53078340 on Build GRCh38
rs751339915 Chr.1: 53078339-53078339 on Build GRCh38
rs577667967 Chr.1: 53078474-53078474 on Build GRCh38
rs750120240 Chr.1: 53078428-53078428 on Build GRCh38
rs143780365 Chr.1: 53078480-53078480 on Build GRCh38
rs867530919 Chr.1: 53078397-53078397 on Build GRCh38
rs761637164 Chr.1: 53078341-53078341 on Build GRCh38
rs772490857 Chr.1: 53078315-53078315 on Build GRCh38
rs772466085 Chr.1: 53078377-53078377 on Build GRCh38
rs369991164 Chr.1: 53078342-53078342 on Build GRCh38
rs199897425 Chr.1: 53078434-53078434 on Build GRCh38
rs775002974 Chr.1: 53078414-53078414 on Build GRCh38
rs763687136 Chr.1: 53078331-53078331 on Build GRCh38
rs201582448 Chr.1: 53078373-53078373 on Build GRCh38
rs760706223 Chr.1: 53078429-53078429 on Build GRCh38
rs748844816 Chr.1: 53078475-53078475 on Build GRCh38
rs201267220 Chr.1: 53078467-53078467 on Build GRCh38
rs200704602 Chr.1: 53078378-53078378 on Build GRCh38
rs371534933 Chr.1: 53078354-53078354 on Build GRCh38
rs747842291 Chr.1: 53078481-53078481 on Build GRCh38
rs531008387 Chr.1: 53078314-53078314 on Build GRCh38
rs111734626 Chr.1: 53078363-53078363 on Build GRCh38
rs767405833 Chr.1: 53078343-53078343 on Build GRCh38
rs769285877 Chr.1: 53078322-53078322 on Build GRCh38
rs548566822 Chr.1: 53078309-53078309 on Build GRCh38
rs754005467 Chr.1: 53078433-53078433 on Build GRCh38
rs779157772 Chr.1: 53078369-53078369 on Build GRCh38
rs759023687 Chr.1: 53078318-53078318 on Build GRCh38

View Full Product Details