Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36045503-36045776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SETD4
Entrez Gene ID
54093
Gene Name
SET domain containing 4
Gene Aliases
C21orf18, C21orf27
UniGene
Hs.606200
Gene Chromosome Location
Chr.21: 36034541-36060818 on Build GRCh38
Transcripts
NM_001007259.2 NM_001007261.2 NM_001286752.1 NM_017438.4 NR_040087.2 XM_011529636.1 XM_011529637.1 XM_011529638.2 XM_011529639.1 XM_011529640.2 XM_011529642.1 XM_011529643.1 XM_011529644.1 XM_017028403.1 XM_017028404.1 XM_017028405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030470 c.629A>G p.Y210C Chr.21: 36045679-36045679 on Build GRCh38
478551 c.552T>A p.F184L Chr.21: 36045756-36045756 on Build GRCh38
1030471 c.625G>A p.V209M Chr.21: 36045683-36045683 on Build GRCh38
2844361 c.669G>A p.P223P Chr.21: 36045639-36045639 on Build GRCh38
6775902 c.694C>T p.L232L Chr.21: 36045614-36045614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765199150 Chr.21: 36045559-36045559 on Build GRCh38
rs928294426 Chr.21: 36045689-36045689 on Build GRCh38
rs145056271 Chr.21: 36045624-36045624 on Build GRCh38
rs747373859 Chr.21: 36045745-36045745 on Build GRCh38
rs768902173 Chr.21: 36045753-36045753 on Build GRCh38
rs751127123 Chr.21: 36045698-36045698 on Build GRCh38
rs149120837 Chr.21: 36045693-36045693 on Build GRCh38
rs139822807 Chr.21: 36045619-36045619 on Build GRCh38
rs369543564 Chr.21: 36045674-36045674 on Build GRCh38
rs747647887 Chr.21: 36045609-36045609 on Build GRCh38
rs141006421 Chr.21: 36045558-36045558 on Build GRCh38
rs145926295 Chr.21: 36045683-36045683 on Build GRCh38
rs996891029 Chr.21: 36045594-36045594 on Build GRCh38
rs766389551 Chr.21: 36045755-36045758 on Build GRCh38
rs747277326 Chr.21: 36045639-36045639 on Build GRCh38
rs146739910 Chr.21: 36045706-36045706 on Build GRCh38
rs767120037 Chr.21: 36045702-36045702 on Build GRCh38
rs775795995 Chr.21: 36045626-36045626 on Build GRCh38
rs144764461 Chr.21: 36045591-36045591 on Build GRCh38
rs79501672 Chr.21: 36045699-36045699 on Build GRCh38
rs779495034 Chr.21: 36045719-36045719 on Build GRCh38
rs746227681 Chr.21: 36045623-36045623 on Build GRCh38
rs570642636 Chr.21: 36045666-36045666 on Build GRCh38
rs776742043 Chr.21: 36045661-36045661 on Build GRCh38
rs780780812 Chr.21: 36045608-36045608 on Build GRCh38
rs556542067 Chr.21: 36045746-36045746 on Build GRCh38
rs372107585 Chr.21: 36045589-36045589 on Build GRCh38
rs140282285 Chr.21: 36045710-36045710 on Build GRCh38
rs1043784210 Chr.21: 36045526-36045526 on Build GRCh38
rs1026820893 Chr.21: 36045665-36045665 on Build GRCh38
rs945547829 Chr.21: 36045754-36045754 on Build GRCh38
rs776381987 Chr.21: 36045551-36045551 on Build GRCh38
rs200958203 Chr.21: 36045694-36045694 on Build GRCh38
rs751220141 Chr.21: 36045605-36045605 on Build GRCh38
rs147906191 Chr.21: 36045606-36045606 on Build GRCh38
rs192595505 Chr.21: 36045531-36045531 on Build GRCh38
rs764246654 Chr.21: 36045550-36045550 on Build GRCh38
rs760520258 Chr.21: 36045684-36045685 on Build GRCh38
rs758786045 Chr.21: 36045727-36045727 on Build GRCh38
rs759770207 Chr.21: 36045618-36045618 on Build GRCh38
rs774561873 Chr.21: 36045683-36045684 on Build GRCh38
rs369448213 Chr.21: 36045602-36045602 on Build GRCh38
rs750979056 Chr.21: 36045726-36045726 on Build GRCh38
rs768729980 Chr.21: 36045681-36045682 on Build GRCh38
rs552463390 Chr.21: 36045581-36045581 on Build GRCh38
rs375159015 Chr.21: 36045640-36045640 on Build GRCh38
rs375798093 Chr.21: 36045737-36045737 on Build GRCh38
rs867592947 Chr.21: 36045641-36045641 on Build GRCh38
rs760602339 Chr.21: 36045542-36045542 on Build GRCh38

View Full Product Details