Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32068205-32068478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF125
Entrez Gene ID
54941
Gene Name
ring finger protein 125
Gene Aliases
TNORS, TRAC-1, TRAC1
UniGene
Hs.633703
Gene Chromosome Location
Chr.18: 32018203-32090751 on Build GRCh38
Transcripts
NM_017831.3 XM_011526045.2 XM_011526046.2 XM_011526047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2809775 c.643C>T p.R215* Chr.18: 32068328-32068328 on Build GRCh38
3737863 c.662C>T p.S221L Chr.18: 32068347-32068347 on Build GRCh38
708063 c.658C>T p.R220W Chr.18: 32068343-32068343 on Build GRCh38
5787807 c.682C>T p.H228Y Chr.18: 32068367-32068367 on Build GRCh38
1388421 c.644G>A p.R215Q Chr.18: 32068329-32068329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953335526 Chr.18: 32068233-32068233 on Build GRCh38
rs1005469268 Chr.18: 32068232-32068232 on Build GRCh38
rs899428619 Chr.18: 32068244-32068244 on Build GRCh38
rs918132274 Chr.18: 32068382-32068382 on Build GRCh38
rs755875377 Chr.18: 32068413-32068413 on Build GRCh38
rs776728236 Chr.18: 32068364-32068364 on Build GRCh38
rs767405359 Chr.18: 32068381-32068381 on Build GRCh38
rs747824170 Chr.18: 32068286-32068286 on Build GRCh38
rs773443582 Chr.18: 32068346-32068346 on Build GRCh38
rs572784172 Chr.18: 32068369-32068369 on Build GRCh38
rs202230286 Chr.18: 32068359-32068359 on Build GRCh38
rs990724378 Chr.18: 32068453-32068453 on Build GRCh38
rs374540528 Chr.18: 32068372-32068372 on Build GRCh38
rs147034251 Chr.18: 32068417-32068417 on Build GRCh38
rs200813448 Chr.18: 32068371-32068371 on Build GRCh38
rs770950994 Chr.18: 32068331-32068332 on Build GRCh38
rs748776580 Chr.18: 32068313-32068313 on Build GRCh38
rs778217602 Chr.18: 32068422-32068422 on Build GRCh38
rs968102373 Chr.18: 32068335-32068335 on Build GRCh38
rs897594277 Chr.18: 32068430-32068430 on Build GRCh38
rs948336489 Chr.18: 32068392-32068392 on Build GRCh38
rs377245557 Chr.18: 32068355-32068355 on Build GRCh38
rs191929096 Chr.18: 32068343-32068343 on Build GRCh38
rs779763691 Chr.18: 32068305-32068305 on Build GRCh38
rs779498047 Chr.18: 32068329-32068329 on Build GRCh38
rs750163347 Chr.18: 32068403-32068403 on Build GRCh38
rs750160685 Chr.18: 32068282-32068282 on Build GRCh38
rs757096377 Chr.18: 32068277-32068277 on Build GRCh38
rs751409311 Chr.18: 32068257-32068257 on Build GRCh38
rs985393278 Chr.18: 32068275-32068275 on Build GRCh38
rs754498390 Chr.18: 32068328-32068328 on Build GRCh38
rs766165124 Chr.18: 32068415-32068415 on Build GRCh38
rs1037844930 Chr.18: 32068236-32068236 on Build GRCh38
rs147556999 Chr.18: 32068344-32068344 on Build GRCh38
rs371439734 Chr.18: 32068432-32068432 on Build GRCh38
rs141596575 Chr.18: 32068375-32068375 on Build GRCh38
rs767152794 Chr.18: 32068279-32068279 on Build GRCh38
rs369064654 Chr.18: 32068396-32068396 on Build GRCh38

View Full Product Details