Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219235106-219235365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKZF1
Entrez Gene ID
55139
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
ZNF744
UniGene
Hs.437647
Gene Chromosome Location
Chr.2: 219229757-219236669 on Build GRCh38
Transcripts
NM_001042410.1 NM_001282792.1 NM_018089.2 XM_005246663.2 XM_011511392.2 XM_017004420.1 XM_017004421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3727536 c.1500_1501insGCTTGCC p.A504fs*21 Chr.2: 219235121-219235122 on Build GRCh38
6311658 c.1585G>T p.A529S Chr.2: 219235206-219235206 on Build GRCh38
5632576 c.1642G>T p.G548C Chr.2: 219235263-219235263 on Build GRCh38
4661991 c.1508G>A p.R503Q Chr.2: 219235129-219235129 on Build GRCh38
5861826 c.1654C>T p.R552C Chr.2: 219235275-219235275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755408001 Chr.2: 219235208-219235208 on Build GRCh38
rs765880938 Chr.2: 219235291-219235291 on Build GRCh38
rs1054130197 Chr.2: 219235254-219235254 on Build GRCh38
rs756921185 Chr.2: 219235183-219235183 on Build GRCh38
rs758768090 Chr.2: 219235313-219235313 on Build GRCh38
rs376658598 Chr.2: 219235129-219235129 on Build GRCh38
rs751344224 Chr.2: 219235179-219235179 on Build GRCh38
rs765127798 Chr.2: 219235150-219235150 on Build GRCh38
rs760804275 Chr.2: 219235145-219235145 on Build GRCh38
rs762944656 Chr.2: 219235161-219235161 on Build GRCh38
rs772277823 Chr.2: 219235132-219235132 on Build GRCh38
rs3172168 Chr.2: 219235164-219235164 on Build GRCh38
rs764107165 Chr.2: 219235247-219235247 on Build GRCh38
rs376576937 Chr.2: 219235241-219235241 on Build GRCh38
rs748531607 Chr.2: 219235212-219235212 on Build GRCh38
rs777959728 Chr.2: 219235315-219235315 on Build GRCh38
rs537199833 Chr.2: 219235276-219235276 on Build GRCh38
rs369641231 Chr.2: 219235188-219235188 on Build GRCh38
rs369167050 Chr.2: 219235306-219235306 on Build GRCh38
rs747150991 Chr.2: 219235236-219235236 on Build GRCh38
rs767197234 Chr.2: 219235187-219235187 on Build GRCh38
rs767120087 Chr.2: 219235262-219235262 on Build GRCh38
rs773993147 Chr.2: 219235307-219235308 on Build GRCh38
rs773222072 Chr.2: 219235135-219235135 on Build GRCh38
rs372235404 Chr.2: 219235202-219235202 on Build GRCh38
rs755632549 Chr.2: 219235199-219235199 on Build GRCh38
rs755686215 Chr.2: 219235290-219235290 on Build GRCh38
rs771217809 Chr.2: 219235325-219235325 on Build GRCh38
rs773452240 Chr.2: 219235221-219235221 on Build GRCh38
rs34353965 Chr.2: 219235285-219235286 on Build GRCh38
rs752703809 Chr.2: 219235155-219235155 on Build GRCh38
rs542260959 Chr.2: 219235220-219235220 on Build GRCh38
rs779260347 Chr.2: 219235210-219235210 on Build GRCh38
rs748502240 Chr.2: 219235309-219235309 on Build GRCh38
rs779486619 Chr.2: 219235200-219235200 on Build GRCh38
rs753289721 Chr.2: 219235297-219235297 on Build GRCh38
rs372230672 Chr.2: 219235128-219235128 on Build GRCh38
rs774352425 Chr.2: 219235251-219235251 on Build GRCh38
rs574591851 Chr.2: 219235240-219235240 on Build GRCh38
rs376958703 Chr.2: 219235285-219235285 on Build GRCh38
rs761635123 Chr.2: 219235255-219235255 on Build GRCh38
rs113918917 Chr.2: 219235278-219235278 on Build GRCh38
rs776907359 Chr.2: 219235328-219235328 on Build GRCh38
rs763986651 Chr.2: 219235165-219235165 on Build GRCh38
rs776786551 Chr.2: 219235246-219235246 on Build GRCh38
rs755538310 Chr.2: 219235299-219235299 on Build GRCh38
rs749697933 Chr.2: 219235126-219235126 on Build GRCh38
rs974179618 Chr.2: 219235143-219235143 on Build GRCh38
rs752130991 Chr.2: 219235317-219235317 on Build GRCh38

View Full Product Details