Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42563975-42564248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COASY
Entrez Gene ID
80347
Gene Name
Coenzyme A synthase
Gene Aliases
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2
UniGene
Hs.296422
Gene Chromosome Location
Chr.17: 42562074-42566281 on Build GRCh38
Transcripts
NM_001042529.2 NM_001042532.3 NM_025233.6 XM_006722116.3 XM_011525300.1 XM_017025167.1 XM_017025168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3188941 c.889G>A p.A297T Chr.17: 42564149-42564149 on Build GRCh38
5726486 c.838C>T p.P280S Chr.17: 42564098-42564098 on Build GRCh38
4514156 c.877C>T p.R293C Chr.17: 42564137-42564137 on Build GRCh38
7279294 c.800T>C p.I267T Chr.17: 42564060-42564060 on Build GRCh38
4513131 c.840C>T p.P280P Chr.17: 42564100-42564100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778257471 Chr.17: 42564139-42564139 on Build GRCh38
rs374954180 Chr.17: 42564086-42564086 on Build GRCh38
rs760194932 Chr.17: 42563998-42563998 on Build GRCh38
rs765118088 Chr.17: 42564009-42564009 on Build GRCh38
rs762858854 Chr.17: 42564164-42564164 on Build GRCh38
rs748463979 Chr.17: 42564083-42564083 on Build GRCh38
rs775245472 Chr.17: 42564161-42564161 on Build GRCh38
rs768480206 Chr.17: 42564165-42564165 on Build GRCh38
rs750537316 Chr.17: 42564034-42564034 on Build GRCh38
rs1051067681 Chr.17: 42564224-42564224 on Build GRCh38
rs668799 Chr.17: 42564217-42564217 on Build GRCh38
rs759494047 Chr.17: 42564225-42564225 on Build GRCh38
rs143964295 Chr.17: 42564062-42564062 on Build GRCh38
rs769804661 Chr.17: 42564159-42564159 on Build GRCh38
rs111775750 Chr.17: 42564178-42564178 on Build GRCh38
rs144328390 Chr.17: 42564145-42564145 on Build GRCh38
rs779390522 Chr.17: 42564081-42564081 on Build GRCh38
rs983128235 Chr.17: 42564107-42564107 on Build GRCh38
rs749242869 Chr.17: 42564168-42564168 on Build GRCh38
rs765029731 Chr.17: 42564091-42564091 on Build GRCh38
rs756932678 Chr.17: 42564019-42564019 on Build GRCh38
rs370902320 Chr.17: 42564124-42564124 on Build GRCh38
rs755520740 Chr.17: 42564137-42564137 on Build GRCh38
rs745310397 Chr.17: 42564146-42564146 on Build GRCh38
rs374017503 Chr.17: 42564135-42564135 on Build GRCh38
rs745350501 Chr.17: 42564080-42564080 on Build GRCh38
rs188790503 Chr.17: 42564015-42564015 on Build GRCh38
rs755438696 Chr.17: 42564078-42564078 on Build GRCh38
rs148652390 Chr.17: 42564085-42564085 on Build GRCh38
rs769118653 Chr.17: 42564060-42564061 on Build GRCh38
rs373985260 Chr.17: 42564023-42564023 on Build GRCh38
rs766450381 Chr.17: 42563999-42563999 on Build GRCh38
rs367865615 Chr.17: 42564138-42564138 on Build GRCh38
rs759581809 Chr.17: 42564090-42564090 on Build GRCh38
rs974811976 Chr.17: 42564048-42564048 on Build GRCh38
rs545304443 Chr.17: 42564226-42564226 on Build GRCh38
rs759365582 Chr.17: 42564004-42564004 on Build GRCh38
rs1009150787 Chr.17: 42564066-42564067 on Build GRCh38
rs776578070 Chr.17: 42564000-42564000 on Build GRCh38
rs749842607 Chr.17: 42564074-42564074 on Build GRCh38
rs762699414 Chr.17: 42564116-42564116 on Build GRCh38
rs11539242 Chr.17: 42564064-42564064 on Build GRCh38
rs756298801 Chr.17: 42564125-42564125 on Build GRCh38
rs371155752 Chr.17: 42564084-42564084 on Build GRCh38
rs767753813 Chr.17: 42564121-42564121 on Build GRCh38
rs780099544 Chr.17: 42564140-42564140 on Build GRCh38
rs760947875 Chr.17: 42564175-42564175 on Build GRCh38
rs750228965 Chr.17: 42564215-42564215 on Build GRCh38
rs756141692 Chr.17: 42564038-42564038 on Build GRCh38
rs766451940 Chr.17: 42564181-42564181 on Build GRCh38
rs767662264 Chr.17: 42564033-42564033 on Build GRCh38
rs766549127 Chr.17: 42564132-42564132 on Build GRCh38
rs777151965 Chr.17: 42564156-42564157 on Build GRCh38
rs377744712 Chr.17: 42564046-42564046 on Build GRCh38
rs775363623 Chr.17: 42564098-42564098 on Build GRCh38
rs753302219 Chr.17: 42564227-42564227 on Build GRCh38
rs775969544 Chr.17: 42564088-42564088 on Build GRCh38

View Full Product Details