Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27793677-27793934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSX1
Entrez Gene ID
219409
Gene Name
GS homeobox 1
Gene Aliases
GSH1, Gsh-1
UniGene
Hs.351785
Gene Chromosome Location
Chr.13: 27792643-27793952 on Build GRCh38
Transcripts
NM_145657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676643 c.698C>T p.S233L Chr.13: 27793851-27793851 on Build GRCh38
1366180 c.642C>T p.G214G Chr.13: 27793795-27793795 on Build GRCh38
6996541 c.745T>A p.S249T Chr.13: 27793898-27793898 on Build GRCh38
1724765 c.637_639delGGC p.G218delG Chr.13: 27793790-27793792 on Build GRCh38
1706590 c.571G>A p.V191M Chr.13: 27793724-27793724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752503192 Chr.13: 27793825-27793825 on Build GRCh38
rs751374189 Chr.13: 27793896-27793896 on Build GRCh38
rs375665681 Chr.13: 27793905-27793905 on Build GRCh38
rs867334741 Chr.13: 27793811-27793811 on Build GRCh38
rs761937649 Chr.13: 27793816-27793816 on Build GRCh38
rs372404864 Chr.13: 27793804-27793804 on Build GRCh38
rs752626603 Chr.13: 27793892-27793892 on Build GRCh38
rs201389990 Chr.13: 27793783-27793783 on Build GRCh38
rs770398963 Chr.13: 27793787-27793787 on Build GRCh38
rs771709573 Chr.13: 27793758-27793758 on Build GRCh38
rs764344642 Chr.13: 27793809-27793809 on Build GRCh38
rs745403977 Chr.13: 27793834-27793834 on Build GRCh38
rs750532200 Chr.13: 27793817-27793817 on Build GRCh38
rs749292156 Chr.13: 27793696-27793696 on Build GRCh38
rs765237584 Chr.13: 27793886-27793886 on Build GRCh38
rs1012268095 Chr.13: 27793699-27793699 on Build GRCh38
rs777461016 Chr.13: 27793826-27793826 on Build GRCh38
rs756810752 Chr.13: 27793899-27793899 on Build GRCh38
rs746699454 Chr.13: 27793827-27793827 on Build GRCh38
rs776015477 Chr.13: 27793839-27793839 on Build GRCh38
rs902250546 Chr.13: 27793700-27793700 on Build GRCh38
rs1023617545 Chr.13: 27793722-27793722 on Build GRCh38
rs746498149 Chr.13: 27793785-27793785 on Build GRCh38
rs540409473 Chr.13: 27793904-27793906 on Build GRCh38
rs760103746 Chr.13: 27793799-27793799 on Build GRCh38
rs138515715 Chr.13: 27793912-27793912 on Build GRCh38
rs946524444 Chr.13: 27793873-27793873 on Build GRCh38
rs893270289 Chr.13: 27793729-27793729 on Build GRCh38
rs780853213 Chr.13: 27793901-27793901 on Build GRCh38
rs3742111 Chr.13: 27793819-27793819 on Build GRCh38
rs750485389 Chr.13: 27793882-27793882 on Build GRCh38
rs777495455 Chr.13: 27793894-27793894 on Build GRCh38
rs774623623 Chr.13: 27793858-27793858 on Build GRCh38
rs1025844856 Chr.13: 27793759-27793759 on Build GRCh38
rs570205728 Chr.13: 27793908-27793908 on Build GRCh38
rs962828716 Chr.13: 27793748-27793748 on Build GRCh38
rs777110730 Chr.13: 27793792-27793792 on Build GRCh38
rs776983383 Chr.13: 27793790-27793792 on Build GRCh38
rs780614099 Chr.13: 27793831-27793831 on Build GRCh38
rs200388174 Chr.13: 27793853-27793853 on Build GRCh38
rs770244924 Chr.13: 27793836-27793836 on Build GRCh38
rs758201014 Chr.13: 27793893-27793893 on Build GRCh38
rs770298297 Chr.13: 27793802-27793802 on Build GRCh38
rs371360518 Chr.13: 27793820-27793820 on Build GRCh38
rs376776365 Chr.13: 27793702-27793702 on Build GRCh38
rs1011796165 Chr.13: 27793749-27793749 on Build GRCh38
rs974396445 Chr.13: 27793849-27793849 on Build GRCh38
rs775782496 Chr.13: 27793803-27793803 on Build GRCh38
rs763351283 Chr.13: 27793851-27793851 on Build GRCh38
rs767869458 Chr.13: 27793875-27793875 on Build GRCh38
rs747975037 Chr.13: 27793725-27793725 on Build GRCh38
rs544288982 Chr.13: 27793714-27793714 on Build GRCh38
rs778787212 Chr.13: 27793719-27793719 on Build GRCh38
rs1032259288 Chr.13: 27793703-27793703 on Build GRCh38
rs761870869 Chr.13: 27793860-27793860 on Build GRCh38
rs770470995 Chr.13: 27793830-27793830 on Build GRCh38
rs751775480 Chr.13: 27793810-27793810 on Build GRCh38
rs760871314 Chr.13: 27793883-27793883 on Build GRCh38
rs951627662 Chr.13: 27793769-27793769 on Build GRCh38

View Full Product Details