Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1296056-1296303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
UniGene
Hs.535257
Gene Chromosome Location
Chr.1: 1292384-1307925 on Build GRCh38
Transcripts
NM_030649.2 XM_005244715.2 XM_005244717.3 XM_011540606.2 XM_011540607.1 XM_011540608.2 XM_011540609.2 XM_017000233.1 NR_106784.1

Gene Symbol
MIR6726
Entrez Gene ID
102465434
Gene Name
microRNA 6726
Gene Aliases
hsa-mir-6726
UniGene
Gene Chromosome Location
Chr.1: 1296110-1296170 on Build GRCh38
Transcripts
NM_030649.2 XM_005244715.2 XM_005244717.3 XM_011540606.2 XM_011540607.1 XM_011540608.2 XM_011540609.2 XM_017000233.1 NR_106784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181549 c.1227C>A p.H409Q Chr.1: 1296265-1296265 on Build GRCh38
4465748 c.1273C>G p.L425V Chr.1: 1296219-1296219 on Build GRCh38
4781664 c.1241G>A p.R414Q Chr.1: 1296251-1296251 on Build GRCh38
5664358 c.1367G>T p.R456L Chr.1: 1296251-1296251 on Build GRCh38
4465747 c.1399C>G p.L467V Chr.1: 1296219-1296219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747130451 Chr.1: 1296150-1296155 on Build GRCh38
rs766195951 Chr.1: 1296119-1296119 on Build GRCh38
rs575866601 Chr.1: 1296247-1296247 on Build GRCh38
rs776360480 Chr.1: 1296155-1296155 on Build GRCh38
rs866964637 Chr.1: 1296233-1296233 on Build GRCh38
rs930824923 Chr.1: 1296211-1296211 on Build GRCh38
rs374708743 Chr.1: 1296176-1296176 on Build GRCh38
rs746056774 Chr.1: 1296100-1296100 on Build GRCh38
rs142196716 Chr.1: 1296099-1296099 on Build GRCh38
rs781672997 Chr.1: 1296141-1296141 on Build GRCh38
rs772986954 Chr.1: 1296165-1296165 on Build GRCh38
rs568940219 Chr.1: 1296133-1296133 on Build GRCh38
rs752144252 Chr.1: 1296126-1296126 on Build GRCh38
rs533022989 Chr.1: 1296177-1296177 on Build GRCh38
rs762328501 Chr.1: 1296178-1296178 on Build GRCh38
rs771834099 Chr.1: 1296114-1296114 on Build GRCh38
rs372140455 Chr.1: 1296188-1296188 on Build GRCh38
rs781112703 Chr.1: 1296235-1296235 on Build GRCh38
rs199607738 Chr.1: 1296164-1296164 on Build GRCh38
rs376193058 Chr.1: 1296123-1296123 on Build GRCh38
rs761082675 Chr.1: 1296280-1296280 on Build GRCh38
rs746292944 Chr.1: 1296143-1296143 on Build GRCh38
rs763627196 Chr.1: 1296184-1296184 on Build GRCh38
rs778066925 Chr.1: 1296143-1296143 on Build GRCh38
rs770189542 Chr.1: 1296109-1296109 on Build GRCh38
rs749266601 Chr.1: 1296162-1296162 on Build GRCh38
rs778412580 Chr.1: 1296086-1296086 on Build GRCh38
rs1049278529 Chr.1: 1296212-1296212 on Build GRCh38
rs760340522 Chr.1: 1296116-1296116 on Build GRCh38
rs150828810 Chr.1: 1296142-1296142 on Build GRCh38
rs769656240 Chr.1: 1296242-1296242 on Build GRCh38
rs201281051 Chr.1: 1296204-1296204 on Build GRCh38
rs773994190 Chr.1: 1296263-1296263 on Build GRCh38
rs756557205 Chr.1: 1296201-1296201 on Build GRCh38
rs369338671 Chr.1: 1296125-1296125 on Build GRCh38
rs757717414 Chr.1: 1296095-1296095 on Build GRCh38
rs619608 Chr.1: 1296127-1296127 on Build GRCh38
rs779031901 Chr.1: 1296220-1296220 on Build GRCh38
rs553742772 Chr.1: 1296190-1296190 on Build GRCh38
rs775415413 Chr.1: 1296113-1296113 on Build GRCh38
rs766737326 Chr.1: 1296283-1296283 on Build GRCh38
rs894789044 Chr.1: 1296223-1296223 on Build GRCh38
rs768250680 Chr.1: 1296252-1296252 on Build GRCh38
rs958718762 Chr.1: 1296144-1296144 on Build GRCh38
rs762644301 Chr.1: 1296251-1296251 on Build GRCh38
rs752301730 Chr.1: 1296084-1296084 on Build GRCh38
rs571161059 Chr.1: 1296115-1296115 on Build GRCh38
rs377421242 Chr.1: 1296170-1296170 on Build GRCh38
rs376304551 Chr.1: 1296134-1296134 on Build GRCh38
rs547767704 Chr.1: 1296092-1296092 on Build GRCh38
rs764453255 Chr.1: 1296132-1296132 on Build GRCh38
rs370373247 Chr.1: 1296172-1296172 on Build GRCh38
rs1749951 Chr.1: 1296276-1296276 on Build GRCh38
rs367638759 Chr.1: 1296185-1296185 on Build GRCh38
rs750987038 Chr.1: 1296277-1296277 on Build GRCh38
rs369165944 Chr.1: 1296085-1296085 on Build GRCh38
rs756395788 Chr.1: 1296147-1296147 on Build GRCh38
rs745605879 Chr.1: 1296240-1296240 on Build GRCh38
rs748289109 Chr.1: 1296232-1296232 on Build GRCh38
rs201882356 Chr.1: 1296198-1296198 on Build GRCh38
rs372530591 Chr.1: 1296122-1296122 on Build GRCh38
rs751641071 Chr.1: 1296206-1296206 on Build GRCh38
rs746921654 Chr.1: 1296166-1296166 on Build GRCh38
rs761408156 Chr.1: 1296268-1296268 on Build GRCh38
rs139600717 Chr.1: 1296207-1296207 on Build GRCh38

View Full Product Details