Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39335180-39335429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
UniGene
Hs.5210
Gene Chromosome Location
Chr.19: 39328357-39336086 on Build GRCh38
Transcripts
NM_001301008.1 NM_004877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6151154 c.150G>T p.Q50H Chr.19: 39335261-39335261 on Build GRCh38
3668410 c.107T>C p.V36A Chr.19: 39335304-39335304 on Build GRCh38
5389706 c.127G>A p.V43M Chr.19: 39335284-39335284 on Build GRCh38
3223694 c.137A>G p.E46G Chr.19: 39335274-39335274 on Build GRCh38
3223696 c.118C>T p.R40W Chr.19: 39335293-39335293 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547389726 Chr.19: 39335314-39335314 on Build GRCh38
rs770098630 Chr.19: 39335216-39335216 on Build GRCh38
rs772377570 Chr.19: 39335389-39335389 on Build GRCh38
rs770360059 Chr.19: 39335313-39335319 on Build GRCh38
rs752337215 Chr.19: 39335295-39335295 on Build GRCh38
rs770358455 Chr.19: 39335328-39335328 on Build GRCh38
rs746585997 Chr.19: 39335227-39335227 on Build GRCh38
rs766175992 Chr.19: 39335291-39335291 on Build GRCh38
rs368775222 Chr.19: 39335249-39335249 on Build GRCh38
rs1001796225 Chr.19: 39335374-39335374 on Build GRCh38
rs79466955 Chr.19: 39335398-39335398 on Build GRCh38
rs1014732456 Chr.19: 39335361-39335361 on Build GRCh38
rs1053099346 Chr.19: 39335330-39335330 on Build GRCh38
rs201966850 Chr.19: 39335230-39335230 on Build GRCh38
rs775956817 Chr.19: 39335220-39335220 on Build GRCh38
rs60059725 Chr.19: 39335372-39335372 on Build GRCh38
rs766342568 Chr.19: 39335404-39335404 on Build GRCh38
rs867487329 Chr.19: 39335272-39335272 on Build GRCh38
rs746367279 Chr.19: 39335211-39335211 on Build GRCh38
rs762051379 Chr.19: 39335252-39335252 on Build GRCh38
rs773058228 Chr.19: 39335366-39335366 on Build GRCh38
rs545801689 Chr.19: 39335234-39335234 on Build GRCh38
rs559147372 Chr.19: 39335347-39335347 on Build GRCh38
rs769138372 Chr.19: 39335356-39335356 on Build GRCh38
rs973040883 Chr.19: 39335239-39335239 on Build GRCh38
rs753677461 Chr.19: 39335292-39335292 on Build GRCh38
rs750367633 Chr.19: 39335266-39335266 on Build GRCh38
rs983397989 Chr.19: 39335358-39335358 on Build GRCh38
rs974111319 Chr.19: 39335271-39335271 on Build GRCh38
rs563997238 Chr.19: 39335349-39335349 on Build GRCh38
rs376154512 Chr.19: 39335388-39335388 on Build GRCh38
rs367559060 Chr.19: 39335397-39335397 on Build GRCh38
rs760224044 Chr.19: 39335379-39335379 on Build GRCh38
rs757956102 Chr.19: 39335296-39335296 on Build GRCh38
rs375066591 Chr.19: 39335357-39335357 on Build GRCh38
rs142875775 Chr.19: 39335293-39335293 on Build GRCh38
rs767683009 Chr.19: 39335257-39335257 on Build GRCh38
rs777308826 Chr.19: 39335300-39335300 on Build GRCh38
rs746464216 Chr.19: 39335315-39335315 on Build GRCh38
rs780369417 Chr.19: 39335344-39335344 on Build GRCh38
rs772593024 Chr.19: 39335220-39335221 on Build GRCh38
rs756107664 Chr.19: 39335277-39335277 on Build GRCh38
rs764426032 Chr.19: 39335243-39335243 on Build GRCh38

View Full Product Details