Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 90009213-90009474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBNDD1
Entrez Gene ID
79007
Gene Name
dysbindin domain containing 1
Gene Aliases
-
UniGene
Hs.301394
Gene Chromosome Location
Chr.16: 90004865-90020131 on Build GRCh38
Transcripts
NM_001042610.2 NM_001288708.1 NM_001288709.1 NM_024043.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
385662 c.49G>A p.E17K Chr.16: 90009413-90009413 on Build GRCh38
3378040 c.176A>T p.E59V Chr.16: 90009346-90009346 on Build GRCh38
4063708 c.107C>T p.T36M Chr.16: 90009355-90009355 on Build GRCh38
6290159 c.538+8C>T p.? Chr.16: 90009276-90009276 on Build GRCh38
6290158 c.178+8C>T p.? Chr.16: 90009276-90009276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763898899 Chr.16: 90009366-90009366 on Build GRCh38
rs368511027 Chr.16: 90009364-90009364 on Build GRCh38
rs747619217 Chr.16: 90009244-90009244 on Build GRCh38
rs759266045 Chr.16: 90009257-90009257 on Build GRCh38
rs373444495 Chr.16: 90009269-90009269 on Build GRCh38
rs771885579 Chr.16: 90009355-90009355 on Build GRCh38
rs745510780 Chr.16: 90009416-90009416 on Build GRCh38
rs776009647 Chr.16: 90009243-90009243 on Build GRCh38
rs567073561 Chr.16: 90009295-90009295 on Build GRCh38
rs746864668 Chr.16: 90009371-90009372 on Build GRCh38
rs534187310 Chr.16: 90009339-90009339 on Build GRCh38
rs370975201 Chr.16: 90009261-90009261 on Build GRCh38
rs199893398 Chr.16: 90009342-90009342 on Build GRCh38
rs771803026 Chr.16: 90009417-90009417 on Build GRCh38
rs780447976 Chr.16: 90009390-90009390 on Build GRCh38
rs931459964 Chr.16: 90009284-90009284 on Build GRCh38
rs386794175 Chr.16: 90009250-90009251 on Build GRCh38
rs879250955 Chr.16: 90009391-90009391 on Build GRCh38
rs766458325 Chr.16: 90009455-90009455 on Build GRCh38
rs760338551 Chr.16: 90009425-90009425 on Build GRCh38
rs751826597 Chr.16: 90009392-90009392 on Build GRCh38
rs774151282 Chr.16: 90009254-90009254 on Build GRCh38
rs868848155 Chr.16: 90009422-90009422 on Build GRCh38
rs753995161 Chr.16: 90009372-90009372 on Build GRCh38
rs776645820 Chr.16: 90009426-90009426 on Build GRCh38
rs775337811 Chr.16: 90009361-90009361 on Build GRCh38
rs561043023 Chr.16: 90009323-90009323 on Build GRCh38
rs772735770 Chr.16: 90009340-90009340 on Build GRCh38
rs748742985 Chr.16: 90009341-90009341 on Build GRCh38
rs750726637 Chr.16: 90009448-90009448 on Build GRCh38
rs928584711 Chr.16: 90009260-90009260 on Build GRCh38
rs201676695 Chr.16: 90009256-90009256 on Build GRCh38
rs939853120 Chr.16: 90009333-90009333 on Build GRCh38
rs200913502 Chr.16: 90009313-90009313 on Build GRCh38
rs139586491 Chr.16: 90009403-90009403 on Build GRCh38
rs377278023 Chr.16: 90009389-90009389 on Build GRCh38
rs371183361 Chr.16: 90009322-90009322 on Build GRCh38
rs770421132 Chr.16: 90009405-90009405 on Build GRCh38
rs781618950 Chr.16: 90009397-90009397 on Build GRCh38
rs372581728 Chr.16: 90009375-90009375 on Build GRCh38
rs757629658 Chr.16: 90009236-90009236 on Build GRCh38
rs200024428 Chr.16: 90009312-90009312 on Build GRCh38
rs199610920 Chr.16: 90009250-90009250 on Build GRCh38
rs548121368 Chr.16: 90009275-90009275 on Build GRCh38
rs755527079 Chr.16: 90009396-90009396 on Build GRCh38
rs763208508 Chr.16: 90009450-90009450 on Build GRCh38
rs758725253 Chr.16: 90009319-90009319 on Build GRCh38
rs35368249 Chr.16: 90009294-90009294 on Build GRCh38
rs765396980 Chr.16: 90009446-90009446 on Build GRCh38
rs747040555 Chr.16: 90009335-90009335 on Build GRCh38
rs918712756 Chr.16: 90009277-90009277 on Build GRCh38
rs201427311 Chr.16: 90009406-90009406 on Build GRCh38
rs761928446 Chr.16: 90009374-90009374 on Build GRCh38
rs768568568 Chr.16: 90009443-90009444 on Build GRCh38
rs768845093 Chr.16: 90009368-90009368 on Build GRCh38
rs750671561 Chr.16: 90009376-90009376 on Build GRCh38
rs771727796 Chr.16: 90009281-90009282 on Build GRCh38
rs201885286 Chr.16: 90009242-90009242 on Build GRCh38
rs779907376 Chr.16: 90009293-90009293 on Build GRCh38
rs546664496 Chr.16: 90009354-90009354 on Build GRCh38
rs201800670 Chr.16: 90009419-90009419 on Build GRCh38
rs184812605 Chr.16: 90009251-90009251 on Build GRCh38
rs765972701 Chr.16: 90009252-90009252 on Build GRCh38
rs1011088938 Chr.16: 90009369-90009369 on Build GRCh38
rs764211823 Chr.16: 90009287-90009287 on Build GRCh38

View Full Product Details