Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3624865-3625394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPRG1L
Entrez Gene ID
127262
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
FAM79A, h-mover, mover
UniGene
Hs.20529
Gene Chromosome Location
Chr.1: 3624992-3630131 on Build GRCh38
Transcripts
NM_182752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342110 c.24G>T p.V8V Chr.1: 3625096-3625096 on Build GRCh38
5185261 c.171C>T p.R57R Chr.1: 3625243-3625243 on Build GRCh38
5729444 c.127C>T p.R43C Chr.1: 3625199-3625199 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931005726 Chr.1: 3625185-3625185 on Build GRCh38
rs971705853 Chr.1: 3625362-3625362 on Build GRCh38
rs776347644 Chr.1: 3625324-3625324 on Build GRCh38
rs1023997925 Chr.1: 3625370-3625370 on Build GRCh38
rs758658171 Chr.1: 3625157-3625157 on Build GRCh38
rs867937785 Chr.1: 3625213-3625213 on Build GRCh38
rs572197968 Chr.1: 3625353-3625353 on Build GRCh38
rs928393948 Chr.1: 3625091-3625091 on Build GRCh38
rs536922963 Chr.1: 3624989-3624989 on Build GRCh38
rs999486937 Chr.1: 3625350-3625350 on Build GRCh38
rs1011552662 Chr.1: 3624980-3624980 on Build GRCh38
rs947655348 Chr.1: 3625186-3625186 on Build GRCh38
rs1036102458 Chr.1: 3625077-3625077 on Build GRCh38
rs1030853558 Chr.1: 3624911-3624911 on Build GRCh38
rs901277646 Chr.1: 3625287-3625288 on Build GRCh38
rs907417399 Chr.1: 3624968-3624968 on Build GRCh38
rs965835464 Chr.1: 3625010-3625010 on Build GRCh38
rs879007959 Chr.1: 3625035-3625035 on Build GRCh38
rs981867431 Chr.1: 3625244-3625244 on Build GRCh38
rs998963690 Chr.1: 3625001-3625001 on Build GRCh38
rs975916967 Chr.1: 3625019-3625019 on Build GRCh38
rs931628093 Chr.1: 3624918-3624918 on Build GRCh38
rs1049224448 Chr.1: 3624921-3624921 on Build GRCh38
rs894696879 Chr.1: 3625184-3625184 on Build GRCh38
rs966685700 Chr.1: 3624912-3624912 on Build GRCh38
rs985153922 Chr.1: 3625337-3625337 on Build GRCh38
rs865863085 Chr.1: 3625038-3625038 on Build GRCh38
rs984462881 Chr.1: 3625045-3625045 on Build GRCh38
rs942296156 Chr.1: 3625294-3625294 on Build GRCh38
rs920161429 Chr.1: 3625175-3625175 on Build GRCh38
rs953433013 Chr.1: 3625026-3625026 on Build GRCh38
rs1032258231 Chr.1: 3624939-3624939 on Build GRCh38
rs999558770 Chr.1: 3624934-3624934 on Build GRCh38
rs991025891 Chr.1: 3624986-3624986 on Build GRCh38
rs1044533845 Chr.1: 3624938-3624939 on Build GRCh38
rs973003335 Chr.1: 3625146-3625146 on Build GRCh38
rs867017182 Chr.1: 3625119-3625119 on Build GRCh38
rs957943764 Chr.1: 3624977-3624977 on Build GRCh38
rs902637624 Chr.1: 3625347-3625347 on Build GRCh38
rs909432682 Chr.1: 3625271-3625271 on Build GRCh38
rs535430198 Chr.1: 3625316-3625316 on Build GRCh38
rs1049400693 Chr.1: 3625333-3625333 on Build GRCh38
rs28695195 Chr.1: 3625061-3625061 on Build GRCh38
rs940451449 Chr.1: 3625069-3625069 on Build GRCh38
rs921823603 Chr.1: 3625025-3625025 on Build GRCh38
rs1003069386 Chr.1: 3624973-3624973 on Build GRCh38
rs1012182784 Chr.1: 3625364-3625364 on Build GRCh38
rs1039585306 Chr.1: 3625314-3625314 on Build GRCh38
rs569965835 Chr.1: 3625223-3625223 on Build GRCh38
rs747418795 Chr.1: 3625313-3625313 on Build GRCh38
rs1051846953 Chr.1: 3625325-3625325 on Build GRCh38
rs1043043648 Chr.1: 3625191-3625191 on Build GRCh38
rs938810825 Chr.1: 3625129-3625129 on Build GRCh38
rs534935543 Chr.1: 3625356-3625356 on Build GRCh38
rs908935734 Chr.1: 3624928-3624928 on Build GRCh38
rs188462649 Chr.1: 3625351-3625351 on Build GRCh38
rs1030831415 Chr.1: 3625006-3625006 on Build GRCh38
rs900552573 Chr.1: 3624894-3624894 on Build GRCh38
rs61454555 Chr.1: 3625159-3625159 on Build GRCh38
rs983792738 Chr.1: 3625270-3625270 on Build GRCh38
rs997627813 Chr.1: 3624903-3624903 on Build GRCh38
rs113047510 Chr.1: 3625165-3625165 on Build GRCh38
rs918361248 Chr.1: 3625265-3625265 on Build GRCh38
rs748594911 Chr.1: 3625054-3625054 on Build GRCh38
rs868543091 Chr.1: 3625274-3625274 on Build GRCh38
rs970861576 Chr.1: 3625243-3625243 on Build GRCh38
rs929636991 Chr.1: 3625267-3625267 on Build GRCh38
rs903216951 Chr.1: 3625199-3625199 on Build GRCh38

View Full Product Details