Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55027867-55028410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEUROD4
Entrez Gene ID
58158
Gene Name
neuronal differentiation 4
Gene Aliases
ATH-3, ATH3, Atoh3, MATH-3, MATH3, bHLHa4
UniGene
Hs.591024
Gene Chromosome Location
Chr.12: 55019945-55030017 on Build GRCh38
Transcripts
NM_021191.2

SNP Detail

SNP ID Chromosome Location
rs958457532 Chr.12: 55028361-55028361 on Build GRCh38
rs766863375 Chr.12: 55027941-55027941 on Build GRCh38
rs987048382 Chr.12: 55028080-55028080 on Build GRCh38
rs920330787 Chr.12: 55028119-55028119 on Build GRCh38
rs534286053 Chr.12: 55027896-55027896 on Build GRCh38
rs540606739 Chr.12: 55028171-55028171 on Build GRCh38
rs1039713769 Chr.12: 55028273-55028273 on Build GRCh38
rs911557168 Chr.12: 55028091-55028091 on Build GRCh38
rs35739714 Chr.12: 55028127-55028127 on Build GRCh38
rs1045715694 Chr.12: 55028182-55028182 on Build GRCh38
rs757844496 Chr.12: 55028136-55028136 on Build GRCh38
rs574218302 Chr.12: 55027952-55027952 on Build GRCh38
rs562041625 Chr.12: 55028180-55028180 on Build GRCh38
rs958385576 Chr.12: 55028034-55028034 on Build GRCh38
rs998935801 Chr.12: 55027957-55027957 on Build GRCh38
rs998154997 Chr.12: 55028307-55028307 on Build GRCh38
rs903217870 Chr.12: 55027924-55027924 on Build GRCh38
rs764745465 Chr.12: 55028066-55028066 on Build GRCh38
rs575743713 Chr.12: 55028063-55028063 on Build GRCh38
rs374260081 Chr.12: 55028372-55028372 on Build GRCh38
rs147815707 Chr.12: 55028046-55028046 on Build GRCh38
rs182305032 Chr.12: 55028181-55028181 on Build GRCh38
rs924165563 Chr.12: 55028174-55028174 on Build GRCh38
rs1025976261 Chr.12: 55028325-55028325 on Build GRCh38
rs761174649 Chr.12: 55027926-55027926 on Build GRCh38
rs754689009 Chr.12: 55027987-55027987 on Build GRCh38
rs1045747568 Chr.12: 55027906-55027906 on Build GRCh38
rs1006827154 Chr.12: 55027910-55027910 on Build GRCh38
rs983906837 Chr.12: 55027979-55027979 on Build GRCh38
rs915774668 Chr.12: 55028147-55028147 on Build GRCh38
rs957693856 Chr.12: 55027978-55027978 on Build GRCh38
rs901128076 Chr.12: 55028277-55028277 on Build GRCh38
rs546481769 Chr.12: 55028107-55028107 on Build GRCh38
rs935488913 Chr.12: 55028175-55028175 on Build GRCh38
rs942756583 Chr.12: 55028221-55028221 on Build GRCh38
rs1018666175 Chr.12: 55027929-55027929 on Build GRCh38
rs1055615898 Chr.12: 55028145-55028145 on Build GRCh38
rs1054876674 Chr.12: 55028249-55028249 on Build GRCh38
rs886972473 Chr.12: 55028336-55028336 on Build GRCh38
rs997362744 Chr.12: 55028335-55028335 on Build GRCh38
rs796617312 Chr.12: 55028191-55028191 on Build GRCh38
rs186724353 Chr.12: 55027930-55027930 on Build GRCh38
rs144912274 Chr.12: 55028202-55028202 on Build GRCh38
rs191166307 Chr.12: 55027961-55027961 on Build GRCh38
rs903102286 Chr.12: 55028195-55028195 on Build GRCh38
rs893195216 Chr.12: 55028326-55028326 on Build GRCh38
rs551955420 Chr.12: 55028275-55028275 on Build GRCh38
rs1005373833 Chr.12: 55028344-55028344 on Build GRCh38
rs909621861 Chr.12: 55028009-55028009 on Build GRCh38
rs141464168 Chr.12: 55028108-55028108 on Build GRCh38
rs998474971 Chr.12: 55027958-55027958 on Build GRCh38

View Full Product Details