Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8354275-8354777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RERE
Entrez Gene ID
473
Gene Name
arginine-glutamic acid dipeptide repeats
Gene Aliases
ARG, ARP, ATN1L, DNB1, NEDBEH
UniGene
Hs.463041
Gene Chromosome Location
Chr.1: 8352404-8826226 on Build GRCh38
Transcripts
NM_001042681.1 NM_001042682.1 NM_012102.3 XM_005263464.2 XM_005263466.1 XM_011541510.1 XM_017001358.1 XM_017001359.1

SNP Detail

SNP ID Chromosome Location
rs982335717 Chr.1: 8354700-8354700 on Build GRCh38
rs1009216135 Chr.1: 8354333-8354333 on Build GRCh38
rs796215279 Chr.1: 8354636-8354636 on Build GRCh38
rs140531389 Chr.1: 8354362-8354362 on Build GRCh38
rs759661513 Chr.1: 8354296-8354297 on Build GRCh38
rs777940177 Chr.1: 8354593-8354593 on Build GRCh38
rs912873020 Chr.1: 8354490-8354491 on Build GRCh38
rs772091996 Chr.1: 8354667-8354667 on Build GRCh38
rs926292712 Chr.1: 8354507-8354507 on Build GRCh38
rs1034156354 Chr.1: 8354707-8354707 on Build GRCh38
rs964001346 Chr.1: 8354341-8354341 on Build GRCh38
rs908692732 Chr.1: 8354384-8354384 on Build GRCh38
rs890400606 Chr.1: 8354315-8354315 on Build GRCh38
rs567163209 Chr.1: 8354637-8354637 on Build GRCh38
rs1042611734 Chr.1: 8354544-8354544 on Build GRCh38
rs544255100 Chr.1: 8354523-8354523 on Build GRCh38
rs183903552 Chr.1: 8354686-8354686 on Build GRCh38
rs912923575 Chr.1: 8354738-8354738 on Build GRCh38
rs116520730 Chr.1: 8354383-8354383 on Build GRCh38
rs563286362 Chr.1: 8354479-8354479 on Build GRCh38
rs532921710 Chr.1: 8354621-8354621 on Build GRCh38
rs979918545 Chr.1: 8354690-8354690 on Build GRCh38
rs572834337 Chr.1: 8354296-8354296 on Build GRCh38
rs971019548 Chr.1: 8354483-8354483 on Build GRCh38
rs945627024 Chr.1: 8354498-8354498 on Build GRCh38
rs1033924945 Chr.1: 8354704-8354704 on Build GRCh38
rs1008819359 Chr.1: 8354624-8354624 on Build GRCh38
rs371641759 Chr.1: 8354371-8354371 on Build GRCh38
rs528038787 Chr.1: 8354645-8354645 on Build GRCh38
rs561591495 Chr.1: 8354379-8354379 on Build GRCh38
rs61055963 Chr.1: 8354626-8354626 on Build GRCh38
rs996891709 Chr.1: 8354644-8354644 on Build GRCh38
rs982304676 Chr.1: 8354497-8354497 on Build GRCh38
rs996942337 Chr.1: 8354349-8354349 on Build GRCh38
rs890664551 Chr.1: 8354558-8354558 on Build GRCh38
rs1024224094 Chr.1: 8354672-8354672 on Build GRCh38
rs748394213 Chr.1: 8354370-8354370 on Build GRCh38
rs562457313 Chr.1: 8354611-8354611 on Build GRCh38
rs971263289 Chr.1: 8354689-8354689 on Build GRCh38
rs944860297 Chr.1: 8354572-8354572 on Build GRCh38
rs957366129 Chr.1: 8354708-8354708 on Build GRCh38
rs549076845 Chr.1: 8354685-8354685 on Build GRCh38
rs952856808 Chr.1: 8354676-8354676 on Build GRCh38
rs989129422 Chr.1: 8354726-8354726 on Build GRCh38
rs1019646323 Chr.1: 8354340-8354340 on Build GRCh38
rs551430946 Chr.1: 8354636-8354636 on Build GRCh38
rs935708432 Chr.1: 8354398-8354398 on Build GRCh38
rs756213569 Chr.1: 8354388-8354388 on Build GRCh38
rs983346837 Chr.1: 8354310-8354310 on Build GRCh38
rs189223675 Chr.1: 8354713-8354713 on Build GRCh38
rs945101069 Chr.1: 8354600-8354600 on Build GRCh38
rs971210849 Chr.1: 8354329-8354329 on Build GRCh38
rs754176154 Chr.1: 8354342-8354342 on Build GRCh38
rs979885905 Chr.1: 8354372-8354372 on Build GRCh38
rs74512989 Chr.1: 8354625-8354625 on Build GRCh38
rs915035915 Chr.1: 8354599-8354599 on Build GRCh38

View Full Product Details