Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101746677-101747218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
UniGene
Hs.46850
Gene Chromosome Location
Chr.12: 101745497-101830867 on Build GRCh38
Transcripts
NM_024312.4 XM_011538731.2 XM_017019961.1 XM_017019962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934031 c.3734T>A p.I1245K Chr.12: 101747201-101747201 on Build GRCh38
2063595 c.3757C>T p.R1253* Chr.12: 101747178-101747178 on Build GRCh38
934030 c.3758G>A p.R1253Q Chr.12: 101747177-101747177 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs992409734 Chr.12: 101746751-101746751 on Build GRCh38
rs956686617 Chr.12: 101747109-101747109 on Build GRCh38
rs76448464 Chr.12: 101746756-101746756 on Build GRCh38
rs993885800 Chr.12: 101747013-101747013 on Build GRCh38
rs775800393 Chr.12: 101746835-101746835 on Build GRCh38
rs867058692 Chr.12: 101747101-101747101 on Build GRCh38
rs897476827 Chr.12: 101747030-101747030 on Build GRCh38
rs201636648 Chr.12: 101747159-101747159 on Build GRCh38
rs377647303 Chr.12: 101747011-101747011 on Build GRCh38
rs1033485316 Chr.12: 101747178-101747178 on Build GRCh38
rs768136710 Chr.12: 101746745-101746745 on Build GRCh38
rs886048848 Chr.12: 101747113-101747113 on Build GRCh38
rs545889458 Chr.12: 101747063-101747063 on Build GRCh38
rs587779412 Chr.12: 101747193-101747196 on Build GRCh38
rs532164205 Chr.12: 101746903-101746903 on Build GRCh38
rs1013695344 Chr.12: 101747026-101747026 on Build GRCh38
rs960884391 Chr.12: 101747023-101747023 on Build GRCh38
rs757111810 Chr.12: 101747120-101747120 on Build GRCh38
rs1025770413 Chr.12: 101746739-101746739 on Build GRCh38
rs750399203 Chr.12: 101747125-101747125 on Build GRCh38
rs977690837 Chr.12: 101746919-101746919 on Build GRCh38
rs75757716 Chr.12: 101746841-101746841 on Build GRCh38
rs281865022 Chr.12: 101747191-101747194 on Build GRCh38
rs183430543 Chr.12: 101747107-101747107 on Build GRCh38
rs960827631 Chr.12: 101746745-101746745 on Build GRCh38
rs779965656 Chr.12: 101746897-101746897 on Build GRCh38
rs749653387 Chr.12: 101747180-101747180 on Build GRCh38
rs752135662 Chr.12: 101747162-101747174 on Build GRCh38
rs746295410 Chr.12: 101746879-101746879 on Build GRCh38
rs1007189646 Chr.12: 101746944-101746944 on Build GRCh38
rs552133314 Chr.12: 101747119-101747119 on Build GRCh38
rs143697254 Chr.12: 101747062-101747062 on Build GRCh38
rs866487764 Chr.12: 101747135-101747135 on Build GRCh38
rs769838026 Chr.12: 101747175-101747175 on Build GRCh38
rs546034103 Chr.12: 101746957-101746957 on Build GRCh38
rs562940893 Chr.12: 101746972-101746972 on Build GRCh38
rs886048846 Chr.12: 101746728-101746728 on Build GRCh38
rs1051029556 Chr.12: 101746878-101746878 on Build GRCh38
rs1038057556 Chr.12: 101747009-101747009 on Build GRCh38
rs781082746 Chr.12: 101747146-101747146 on Build GRCh38
rs368459751 Chr.12: 101747126-101747126 on Build GRCh38
rs979470900 Chr.12: 101746826-101746826 on Build GRCh38
rs936182294 Chr.12: 101746953-101746953 on Build GRCh38
rs756564004 Chr.12: 101747039-101747039 on Build GRCh38
rs767059848 Chr.12: 101747160-101747162 on Build GRCh38
rs778120023 Chr.12: 101747177-101747177 on Build GRCh38
rs1050927605 Chr.12: 101747182-101747182 on Build GRCh38
rs913106412 Chr.12: 101746850-101746850 on Build GRCh38
rs76267316 Chr.12: 101747021-101747021 on Build GRCh38
rs993853410 Chr.12: 101746728-101746728 on Build GRCh38
rs780676592 Chr.12: 101747090-101747090 on Build GRCh38
rs887609718 Chr.12: 101746760-101746760 on Build GRCh38
rs79089208 Chr.12: 101747121-101747121 on Build GRCh38
rs886048847 Chr.12: 101746744-101746745 on Build GRCh38
rs1024195463 Chr.12: 101747061-101747061 on Build GRCh38
rs560787679 Chr.12: 101746804-101746804 on Build GRCh38
rs925252982 Chr.12: 101746832-101746832 on Build GRCh38
rs76332781 Chr.12: 101746755-101746755 on Build GRCh38
rs751418309 Chr.12: 101747151-101747160 on Build GRCh38
rs926234798 Chr.12: 101746933-101746933 on Build GRCh38
rs372942733 Chr.12: 101746808-101746808 on Build GRCh38
rs190881677 Chr.12: 101746855-101746855 on Build GRCh38

View Full Product Details