Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101757123-101757623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
UniGene
Hs.46850
Gene Chromosome Location
Chr.12: 101745497-101830867 on Build GRCh38
Transcripts
NM_024312.4 XM_006719593.2 XM_011538731.2 XM_017019961.1 XM_017019962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703810 c.3335+9T>C p.? Chr.12: 101757563-101757563 on Build GRCh38
3670930 c.3377T>A p.I1126N Chr.12: 101757269-101757269 on Build GRCh38
1208433 c.3286C>T p.P1096S Chr.12: 101757621-101757621 on Build GRCh38
6250550 c.3363C>A p.I1121I Chr.12: 101757283-101757283 on Build GRCh38
5561425 c.3431C>A p.P1144H Chr.12: 101757215-101757215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770879064 Chr.12: 101757216-101757216 on Build GRCh38
rs587777914 Chr.12: 101757578-101757579 on Build GRCh38
rs372300988 Chr.12: 101757177-101757177 on Build GRCh38
rs778051529 Chr.12: 101757538-101757538 on Build GRCh38
rs191087950 Chr.12: 101757561-101757561 on Build GRCh38
rs931694294 Chr.12: 101757447-101757447 on Build GRCh38
rs759860576 Chr.12: 101757168-101757168 on Build GRCh38
rs530706212 Chr.12: 101757196-101757196 on Build GRCh38
rs756776207 Chr.12: 101757208-101757208 on Build GRCh38
rs921704035 Chr.12: 101757411-101757411 on Build GRCh38
rs766483004 Chr.12: 101757354-101757355 on Build GRCh38
rs371971179 Chr.12: 101757316-101757316 on Build GRCh38
rs281865013 Chr.12: 101757597-101757597 on Build GRCh38
rs281865015 Chr.12: 101757257-101757258 on Build GRCh38
rs3736476 Chr.12: 101757335-101757335 on Build GRCh38
rs34940801 Chr.12: 101757571-101757571 on Build GRCh38
rs767741989 Chr.12: 101757299-101757299 on Build GRCh38
rs749361169 Chr.12: 101757266-101757266 on Build GRCh38
rs759703222 Chr.12: 101757290-101757290 on Build GRCh38
rs551411643 Chr.12: 101757259-101757259 on Build GRCh38
rs397507562 Chr.12: 101757311-101757311 on Build GRCh38
rs760291071 Chr.12: 101757313-101757314 on Build GRCh38
rs756760913 Chr.12: 101757349-101757349 on Build GRCh38
rs1043915308 Chr.12: 101757400-101757400 on Build GRCh38
rs996615189 Chr.12: 101757478-101757478 on Build GRCh38
rs141927805 Chr.12: 101757558-101757558 on Build GRCh38
rs768634271 Chr.12: 101757258-101757258 on Build GRCh38
rs565505799 Chr.12: 101757197-101757197 on Build GRCh38
rs764653236 Chr.12: 101757317-101757317 on Build GRCh38
rs1039988266 Chr.12: 101757399-101757399 on Build GRCh38
rs915045768 Chr.12: 101757297-101757297 on Build GRCh38
rs892747662 Chr.12: 101757375-101757375 on Build GRCh38
rs148143260 Chr.12: 101757155-101757155 on Build GRCh38
rs281865036 Chr.12: 101757211-101757211 on Build GRCh38
rs755074565 Chr.12: 101757544-101757545 on Build GRCh38
rs563459332 Chr.12: 101757160-101757160 on Build GRCh38
rs34788341 Chr.12: 101757566-101757566 on Build GRCh38
rs563131363 Chr.12: 101757398-101757398 on Build GRCh38
rs201135024 Chr.12: 101757532-101757532 on Build GRCh38
rs372518393 Chr.12: 101757539-101757539 on Build GRCh38
rs281865017 Chr.12: 101757217-101757218 on Build GRCh38
rs761079309 Chr.12: 101757594-101757594 on Build GRCh38
rs758054970 Chr.12: 101757552-101757552 on Build GRCh38
rs373942566 Chr.12: 101757557-101757557 on Build GRCh38
rs1040081337 Chr.12: 101757151-101757151 on Build GRCh38
rs745600783 Chr.12: 101757229-101757230 on Build GRCh38
rs775751653 Chr.12: 101757593-101757593 on Build GRCh38
rs923625439 Chr.12: 101757353-101757353 on Build GRCh38
rs753387393 Chr.12: 101757204-101757204 on Build GRCh38
rs746803948 Chr.12: 101757241-101757241 on Build GRCh38
rs750175714 Chr.12: 101757315-101757316 on Build GRCh38
rs774443381 Chr.12: 101757282-101757282 on Build GRCh38
rs577796531 Chr.12: 101757215-101757215 on Build GRCh38
rs776720992 Chr.12: 101757314-101757315 on Build GRCh38
rs548963882 Chr.12: 101757432-101757432 on Build GRCh38
rs751743725 Chr.12: 101757536-101757537 on Build GRCh38
rs181723747 Chr.12: 101757506-101757506 on Build GRCh38
rs775768061 Chr.12: 101757313-101757313 on Build GRCh38
rs142065232 Chr.12: 101757236-101757236 on Build GRCh38
rs775105620 Chr.12: 101757313-101757324 on Build GRCh38
rs548910758 Chr.12: 101757237-101757237 on Build GRCh38
rs778606499 Chr.12: 101757212-101757212 on Build GRCh38
rs758698837 Chr.12: 101757317-101757318 on Build GRCh38
rs281865016 Chr.12: 101757254-101757254 on Build GRCh38
rs146680824 Chr.12: 101757574-101757574 on Build GRCh38
rs771601013 Chr.12: 101757286-101757288 on Build GRCh38
rs748650433 Chr.12: 101757199-101757200 on Build GRCh38
rs34341883 Chr.12: 101757499-101757500 on Build GRCh38
rs764517026 Chr.12: 101757198-101757198 on Build GRCh38
rs140312213 Chr.12: 101757590-101757590 on Build GRCh38
rs1049705416 Chr.12: 101757517-101757517 on Build GRCh38
rs758156963 Chr.12: 101757231-101757231 on Build GRCh38
rs971386555 Chr.12: 101757217-101757217 on Build GRCh38
rs148602406 Chr.12: 101757232-101757232 on Build GRCh38
rs770952898 Chr.12: 101757267-101757267 on Build GRCh38
rs1049206131 Chr.12: 101757175-101757175 on Build GRCh38
rs528882097 Chr.12: 101757420-101757420 on Build GRCh38
rs187256830 Chr.12: 101757559-101757559 on Build GRCh38
rs281865014 Chr.12: 101757579-101757580 on Build GRCh38
rs797044663 Chr.12: 101757580-101757581 on Build GRCh38

View Full Product Details