Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110143282-110143819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT81
Entrez Gene ID
28981
Gene Name
intraflagellar transport 81
Gene Aliases
CDV-1, CDV-1R, CDV1, CDV1R, DV1
UniGene
Hs.528382
Gene Chromosome Location
Chr.12: 110124335-110218795 on Build GRCh38
Transcripts
NM_001143779.1 NM_014055.3 NM_031473.3 XM_011538232.2 XM_017019217.1 XM_017019218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4038624 c.900A>G p.S300S Chr.12: 110143500-110143500 on Build GRCh38
1298901 c.937G>C p.E313Q Chr.12: 110143537-110143537 on Build GRCh38
5937549 c.844C>T p.P282S Chr.12: 110143444-110143444 on Build GRCh38
4038625 c.900A>G p.S300S Chr.12: 110143500-110143500 on Build GRCh38
934921 c.789G>T p.M263I Chr.12: 110143389-110143389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748641300 Chr.12: 110143494-110143494 on Build GRCh38
rs752285806 Chr.12: 110143531-110143531 on Build GRCh38
rs767043905 Chr.12: 110143413-110143413 on Build GRCh38
rs753762781 Chr.12: 110143586-110143586 on Build GRCh38
rs749119106 Chr.12: 110143429-110143429 on Build GRCh38
rs763261209 Chr.12: 110143363-110143363 on Build GRCh38
rs117092761 Chr.12: 110143503-110143503 on Build GRCh38
rs751150566 Chr.12: 110143380-110143380 on Build GRCh38
rs748149875 Chr.12: 110143605-110143605 on Build GRCh38
rs756050254 Chr.12: 110143425-110143425 on Build GRCh38
rs144191176 Chr.12: 110143752-110143755 on Build GRCh38
rs916643786 Chr.12: 110143655-110143655 on Build GRCh38
rs764142019 Chr.12: 110143572-110143572 on Build GRCh38
rs1008541457 Chr.12: 110143783-110143783 on Build GRCh38
rs373713735 Chr.12: 110143422-110143422 on Build GRCh38
rs936102429 Chr.12: 110143672-110143672 on Build GRCh38
rs929209695 Chr.12: 110143734-110143734 on Build GRCh38
rs1039659067 Chr.12: 110143679-110143679 on Build GRCh38
rs917395217 Chr.12: 110143608-110143608 on Build GRCh38
rs137970540 Chr.12: 110143517-110143517 on Build GRCh38
rs757061250 Chr.12: 110143438-110143438 on Build GRCh38
rs771215887 Chr.12: 110143513-110143513 on Build GRCh38
rs1009052577 Chr.12: 110143398-110143398 on Build GRCh38
rs763317803 Chr.12: 110143510-110143510 on Build GRCh38
rs1004362483 Chr.12: 110143353-110143353 on Build GRCh38
rs765637006 Chr.12: 110143345-110143345 on Build GRCh38
rs768772933 Chr.12: 110143324-110143324 on Build GRCh38
rs781307888 Chr.12: 110143492-110143492 on Build GRCh38
rs1016725201 Chr.12: 110143356-110143356 on Build GRCh38
rs79746168 Chr.12: 110143715-110143715 on Build GRCh38
rs372365248 Chr.12: 110143526-110143526 on Build GRCh38
rs927937565 Chr.12: 110143599-110143599 on Build GRCh38
rs899030764 Chr.12: 110143721-110143724 on Build GRCh38
rs777815051 Chr.12: 110143426-110143426 on Build GRCh38
rs529783964 Chr.12: 110143516-110143516 on Build GRCh38
rs1037504212 Chr.12: 110143705-110143705 on Build GRCh38
rs563332921 Chr.12: 110143708-110143708 on Build GRCh38
rs978407342 Chr.12: 110143582-110143582 on Build GRCh38
rs763536755 Chr.12: 110143330-110143330 on Build GRCh38
rs764579685 Chr.12: 110143589-110143589 on Build GRCh38
rs773534261 Chr.12: 110143357-110143357 on Build GRCh38
rs543715727 Chr.12: 110143311-110143311 on Build GRCh38
rs977993844 Chr.12: 110143625-110143625 on Build GRCh38
rs886976227 Chr.12: 110143344-110143344 on Build GRCh38
rs767562148 Chr.12: 110143462-110143462 on Build GRCh38
rs941427720 Chr.12: 110143686-110143686 on Build GRCh38
rs946274526 Chr.12: 110143615-110143615 on Build GRCh38
rs780428578 Chr.12: 110143447-110143447 on Build GRCh38
rs768675931 Chr.12: 110143486-110143486 on Build GRCh38
rs760215363 Chr.12: 110143569-110143569 on Build GRCh38
rs565209983 Chr.12: 110143664-110143664 on Build GRCh38
rs576969206 Chr.12: 110143385-110143385 on Build GRCh38
rs967358516 Chr.12: 110143557-110143557 on Build GRCh38
rs60059195 Chr.12: 110143755-110143758 on Build GRCh38
rs776017535 Chr.12: 110143499-110143500 on Build GRCh38
rs1047661665 Chr.12: 110143742-110143742 on Build GRCh38
rs747250475 Chr.12: 110143478-110143478 on Build GRCh38
rs926027854 Chr.12: 110143632-110143632 on Build GRCh38
rs558440431 Chr.12: 110143323-110143323 on Build GRCh38
rs867003036 Chr.12: 110143529-110143529 on Build GRCh38
rs761083847 Chr.12: 110143548-110143549 on Build GRCh38
rs773658965 Chr.12: 110143507-110143507 on Build GRCh38
rs368690243 Chr.12: 110143427-110143427 on Build GRCh38
rs1026399689 Chr.12: 110143358-110143361 on Build GRCh38
rs540684366 Chr.12: 110143392-110143392 on Build GRCh38

View Full Product Details