Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23117528-23118058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPE
Entrez Gene ID
1053
Gene Name
CCAAT/enhancer binding protein epsilon
Gene Aliases
C/EBP-epsilon, CRP1
UniGene
Hs.558308
Gene Chromosome Location
Chr.14: 23117306-23119611 on Build GRCh38
Transcripts
NM_001805.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625966 c.717C>T p.Y239Y Chr.14: 23117616-23117616 on Build GRCh38
5603146 c.565C>T p.P189S Chr.14: 23117768-23117768 on Build GRCh38
6625964 c.622C>T p.R208W Chr.14: 23117711-23117711 on Build GRCh38
3495117 c.514C>T p.P172S Chr.14: 23117819-23117819 on Build GRCh38
5560821 c.603C>G p.N201K Chr.14: 23117730-23117730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780403578 Chr.14: 23117753-23117753 on Build GRCh38
rs987222721 Chr.14: 23118018-23118018 on Build GRCh38
rs367795312 Chr.14: 23117622-23117622 on Build GRCh38
rs923787623 Chr.14: 23117638-23117638 on Build GRCh38
rs755654290 Chr.14: 23117561-23117561 on Build GRCh38
rs747524697 Chr.14: 23117680-23117680 on Build GRCh38
rs545257637 Chr.14: 23117833-23117833 on Build GRCh38
rs1044847849 Chr.14: 23117887-23117887 on Build GRCh38
rs753337511 Chr.14: 23117724-23117724 on Build GRCh38
rs780345897 Chr.14: 23117654-23117654 on Build GRCh38
rs943222909 Chr.14: 23117863-23117863 on Build GRCh38
rs749364093 Chr.14: 23117839-23117839 on Build GRCh38
rs775481612 Chr.14: 23117786-23117786 on Build GRCh38
rs751972931 Chr.14: 23117821-23117821 on Build GRCh38
rs373251741 Chr.14: 23117766-23117766 on Build GRCh38
rs778242451 Chr.14: 23117744-23117744 on Build GRCh38
rs779209834 Chr.14: 23117587-23117587 on Build GRCh38
rs374712257 Chr.14: 23117610-23117610 on Build GRCh38
rs757986590 Chr.14: 23117752-23117752 on Build GRCh38
rs902804048 Chr.14: 23117942-23117942 on Build GRCh38
rs755540311 Chr.14: 23117758-23117758 on Build GRCh38
rs201609537 Chr.14: 23117621-23117621 on Build GRCh38
rs772839606 Chr.14: 23117866-23117866 on Build GRCh38
rs777915705 Chr.14: 23117838-23117838 on Build GRCh38
rs141185489 Chr.14: 23117709-23117709 on Build GRCh38
rs746230262 Chr.14: 23117588-23117588 on Build GRCh38
rs954495730 Chr.14: 23117840-23117840 on Build GRCh38
rs201257734 Chr.14: 23117732-23117732 on Build GRCh38
rs750122474 Chr.14: 23117637-23117637 on Build GRCh38
rs768742384 Chr.14: 23117604-23117604 on Build GRCh38
rs770176922 Chr.14: 23117913-23117913 on Build GRCh38
rs748972715 Chr.14: 23117582-23117582 on Build GRCh38
rs375672870 Chr.14: 23117694-23117694 on Build GRCh38
rs778374638 Chr.14: 23117797-23117797 on Build GRCh38
rs369145261 Chr.14: 23117769-23117769 on Build GRCh38
rs1031340948 Chr.14: 23117981-23117981 on Build GRCh38
rs764813413 Chr.14: 23117800-23117800 on Build GRCh38
rs372651475 Chr.14: 23117653-23117653 on Build GRCh38
rs777380202 Chr.14: 23117580-23117580 on Build GRCh38
rs753524435 Chr.14: 23117730-23117730 on Build GRCh38
rs529712973 Chr.14: 23117898-23117898 on Build GRCh38
rs895303867 Chr.14: 23117933-23117933 on Build GRCh38
rs985574262 Chr.14: 23117835-23117835 on Build GRCh38
rs756917368 Chr.14: 23117636-23117636 on Build GRCh38
rs564121394 Chr.14: 23117884-23117884 on Build GRCh38
rs769087828 Chr.14: 23117682-23117682 on Build GRCh38
rs758115639 Chr.14: 23117809-23117809 on Build GRCh38
rs749492170 Chr.14: 23118030-23118030 on Build GRCh38
rs776758085 Chr.14: 23117557-23117558 on Build GRCh38
rs55722931 Chr.14: 23117586-23117586 on Build GRCh38
rs781711989 Chr.14: 23117761-23117761 on Build GRCh38
rs755307869 Chr.14: 23117829-23117829 on Build GRCh38
rs751372334 Chr.14: 23117615-23117615 on Build GRCh38
rs376534358 Chr.14: 23117768-23117768 on Build GRCh38
rs756759095 Chr.14: 23117731-23117731 on Build GRCh38
rs767445685 Chr.14: 23117711-23117711 on Build GRCh38
rs747372846 Chr.14: 23117757-23117757 on Build GRCh38
rs764108892 Chr.14: 23117723-23117723 on Build GRCh38
rs761948458 Chr.14: 23117614-23117614 on Build GRCh38
rs746255618 Chr.14: 23117645-23117645 on Build GRCh38
rs766005459 Chr.14: 23117816-23117816 on Build GRCh38
rs761811363 Chr.14: 23117864-23117864 on Build GRCh38
rs781372006 Chr.14: 23117831-23117831 on Build GRCh38
rs764908366 Chr.14: 23117795-23117796 on Build GRCh38
rs1051640040 Chr.14: 23117603-23117603 on Build GRCh38
rs761813926 Chr.14: 23117722-23117725 on Build GRCh38
rs772501158 Chr.14: 23117594-23117594 on Build GRCh38
rs764166526 Chr.14: 23117792-23117792 on Build GRCh38
rs754942176 Chr.14: 23117552-23117552 on Build GRCh38
rs746160407 Chr.14: 23117891-23117891 on Build GRCh38
rs1022553425 Chr.14: 23117940-23117940 on Build GRCh38
rs991992471 Chr.14: 23117878-23117878 on Build GRCh38
rs544526350 Chr.14: 23117883-23117883 on Build GRCh38
rs756067667 Chr.14: 23117625-23117625 on Build GRCh38
rs998433552 Chr.14: 23117970-23117970 on Build GRCh38
rs965771623 Chr.14: 23117739-23117739 on Build GRCh38
rs147384349 Chr.14: 23117747-23117747 on Build GRCh38
rs761128015 Chr.14: 23117600-23117600 on Build GRCh38
rs906571776 Chr.14: 23117900-23117900 on Build GRCh38
rs765084840 Chr.14: 23117865-23117865 on Build GRCh38
rs2144827 Chr.14: 23118022-23118022 on Build GRCh38
rs775939975 Chr.14: 23117599-23117599 on Build GRCh38
rs948803604 Chr.14: 23117734-23117734 on Build GRCh38
rs769963026 Chr.14: 23117703-23117703 on Build GRCh38
rs889923257 Chr.14: 23117982-23117982 on Build GRCh38
rs964571530 Chr.14: 23118024-23118024 on Build GRCh38
rs763046578 Chr.14: 23117710-23117710 on Build GRCh38
rs749540911 Chr.14: 23117782-23117782 on Build GRCh38
rs372854243 Chr.14: 23117775-23117775 on Build GRCh38
rs998489439 Chr.14: 23117962-23117962 on Build GRCh38
rs746046901 Chr.14: 23117847-23117847 on Build GRCh38
rs137857228 Chr.14: 23117799-23117799 on Build GRCh38
rs954591533 Chr.14: 23118009-23118009 on Build GRCh38
rs766005797 Chr.14: 23117720-23117720 on Build GRCh38
rs771272639 Chr.14: 23117784-23117784 on Build GRCh38
rs147453535 Chr.14: 23117837-23117837 on Build GRCh38
rs777425212 Chr.14: 23117630-23117630 on Build GRCh38
rs539823908 Chr.14: 23117921-23117921 on Build GRCh38
rs978378493 Chr.14: 23117635-23117635 on Build GRCh38
rs760523803 Chr.14: 23117787-23117787 on Build GRCh38
rs767427610 Chr.14: 23117923-23117923 on Build GRCh38
rs768590416 Chr.14: 23117855-23117855 on Build GRCh38
rs939468748 Chr.14: 23117912-23117912 on Build GRCh38
rs914888795 Chr.14: 23117611-23117611 on Build GRCh38
rs1011104096 Chr.14: 23117934-23117934 on Build GRCh38
rs749960753 Chr.14: 23117811-23117819 on Build GRCh38
rs45496295 Chr.14: 23117846-23117846 on Build GRCh38
rs776227686 Chr.14: 23117858-23117858 on Build GRCh38
rs749832978 Chr.14: 23117805-23117805 on Build GRCh38
rs751304427 Chr.14: 23117551-23117551 on Build GRCh38
rs763610496 Chr.14: 23117601-23117601 on Build GRCh38
rs753381359 Chr.14: 23117631-23117631 on Build GRCh38
rs184275935 Chr.14: 23118027-23118027 on Build GRCh38
rs760759078 Chr.14: 23117722-23117722 on Build GRCh38

View Full Product Details