Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92939644-92940150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts
XM_017021262.1 XM_017021263.1 XM_017021264.1

SNP Detail

SNP ID Chromosome Location
rs974408367 Chr.14: 92939729-92939729 on Build GRCh38
rs987101375 Chr.14: 92939747-92939747 on Build GRCh38
rs535258817 Chr.14: 92939699-92939699 on Build GRCh38
rs982600552 Chr.14: 92939877-92939877 on Build GRCh38
rs982548054 Chr.14: 92940073-92940073 on Build GRCh38
rs886301897 Chr.14: 92939694-92939694 on Build GRCh38
rs776246008 Chr.14: 92939880-92939880 on Build GRCh38
rs188946764 Chr.14: 92940070-92940070 on Build GRCh38
rs924754253 Chr.14: 92939750-92939750 on Build GRCh38
rs1031933397 Chr.14: 92939999-92939999 on Build GRCh38
rs955601257 Chr.14: 92939744-92939744 on Build GRCh38
rs893358336 Chr.14: 92940000-92940000 on Build GRCh38
rs999459749 Chr.14: 92939682-92939682 on Build GRCh38
rs948446927 Chr.14: 92939938-92939938 on Build GRCh38
rs7156335 Chr.14: 92939887-92939887 on Build GRCh38
rs761027461 Chr.14: 92939875-92939875 on Build GRCh38
rs995960649 Chr.14: 92939951-92939951 on Build GRCh38
rs955220607 Chr.14: 92939840-92939840 on Build GRCh38
rs934862024 Chr.14: 92939770-92939770 on Build GRCh38
rs941292940 Chr.14: 92939907-92939907 on Build GRCh38
rs907248729 Chr.14: 92939851-92939851 on Build GRCh38
rs573525789 Chr.14: 92940006-92940006 on Build GRCh38
rs1006364195 Chr.14: 92939695-92939695 on Build GRCh38
rs999021074 Chr.14: 92939991-92939991 on Build GRCh38
rs3814829 Chr.14: 92939964-92939964 on Build GRCh38
rs965117004 Chr.14: 92939780-92939780 on Build GRCh38
rs773505523 Chr.14: 92939870-92939870 on Build GRCh38
rs562199667 Chr.14: 92940028-92940028 on Build GRCh38
rs1016034050 Chr.14: 92939720-92939720 on Build GRCh38
rs542419973 Chr.14: 92940027-92940027 on Build GRCh38
rs1028103623 Chr.14: 92939689-92939703 on Build GRCh38
rs980186512 Chr.14: 92940121-92940121 on Build GRCh38
rs990608678 Chr.14: 92939779-92939779 on Build GRCh38
rs938585951 Chr.14: 92939865-92939865 on Build GRCh38
rs1030395810 Chr.14: 92940037-92940037 on Build GRCh38
rs546483457 Chr.14: 92939692-92939692 on Build GRCh38
rs1030041758 Chr.14: 92939739-92939739 on Build GRCh38
rs1046296590 Chr.14: 92939671-92939671 on Build GRCh38
rs74074511 Chr.14: 92939815-92939815 on Build GRCh38
rs956365989 Chr.14: 92940007-92940007 on Build GRCh38
rs148617200 Chr.14: 92939932-92939932 on Build GRCh38
rs111467667 Chr.14: 92939679-92939679 on Build GRCh38
rs970110281 Chr.14: 92940111-92940111 on Build GRCh38
rs928622291 Chr.14: 92940130-92940130 on Build GRCh38
rs570671160 Chr.14: 92939672-92939672 on Build GRCh38
rs577487867 Chr.14: 92939918-92939918 on Build GRCh38
rs964345293 Chr.14: 92939727-92939727 on Build GRCh38
rs950901594 Chr.14: 92940071-92940071 on Build GRCh38
rs752314145 Chr.14: 92939979-92939979 on Build GRCh38
rs372501450 Chr.14: 92939799-92939799 on Build GRCh38
rs566292723 Chr.14: 92939696-92939696 on Build GRCh38
rs367917032 Chr.14: 92939678-92939678 on Build GRCh38
rs537480911 Chr.14: 92939817-92939817 on Build GRCh38
rs61993813 Chr.14: 92939812-92939812 on Build GRCh38
rs760508618 Chr.14: 92939669-92939669 on Build GRCh38
rs764741207 Chr.14: 92939977-92939977 on Build GRCh38
rs954382108 Chr.14: 92939965-92939965 on Build GRCh38
rs1017624558 Chr.14: 92939714-92939714 on Build GRCh38
rs575774971 Chr.14: 92940066-92940066 on Build GRCh38

View Full Product Details