Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84174340-84174835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF1
Entrez Gene ID
123872
Gene Name
dynein axonemal assembly factor 1
Gene Aliases
CILD13, LRRC50, ODA7
UniGene
Hs.310164
Gene Chromosome Location
Chr.16: 84145260-84178761 on Build GRCh38
Transcripts
NM_001318756.1 NM_178452.5 XM_006721129.2 XM_011522853.2 XM_011522854.2 XM_011522855.2 XM_011522857.2 XM_011522858.2 XM_017022918.1 XM_017022919.1 XM_017022920.1 XM_017022921.1 XM_017022922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974350 c.1685C>A p.S562Y Chr.16: 84174709-84174709 on Build GRCh38
1679307 c.1660G>A p.E554K Chr.16: 84174684-84174684 on Build GRCh38
1609723 c.1686T>C p.S562S Chr.16: 84174710-84174710 on Build GRCh38
6555956 c.1647C>T p.D549D Chr.16: 84174671-84174671 on Build GRCh38
148128 c.1698+7G>T p.? Chr.16: 84174729-84174729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774381286 Chr.16: 84174720-84174720 on Build GRCh38
rs370131347 Chr.16: 84174618-84174618 on Build GRCh38
rs761818154 Chr.16: 84174722-84174722 on Build GRCh38
rs972481887 Chr.16: 84174623-84174623 on Build GRCh38
rs764303732 Chr.16: 84174431-84174431 on Build GRCh38
rs995033031 Chr.16: 84174440-84174440 on Build GRCh38
rs1018531059 Chr.16: 84174490-84174490 on Build GRCh38
rs778217102 Chr.16: 84174696-84174696 on Build GRCh38
rs754260073 Chr.16: 84174628-84174628 on Build GRCh38
rs752104162 Chr.16: 84174773-84174773 on Build GRCh38
rs889070412 Chr.16: 84174455-84174455 on Build GRCh38
rs145973397 Chr.16: 84174688-84174688 on Build GRCh38
rs757333074 Chr.16: 84174702-84174702 on Build GRCh38
rs762954791 Chr.16: 84174622-84174622 on Build GRCh38
rs561443632 Chr.16: 84174631-84174631 on Build GRCh38
rs545882696 Chr.16: 84174564-84174564 on Build GRCh38
rs369588547 Chr.16: 84174788-84174788 on Build GRCh38
rs751005065 Chr.16: 84174687-84174689 on Build GRCh38
rs952239308 Chr.16: 84174554-84174554 on Build GRCh38
rs371392611 Chr.16: 84174617-84174617 on Build GRCh38
rs542552230 Chr.16: 84174603-84174603 on Build GRCh38
rs201984126 Chr.16: 84174647-84174647 on Build GRCh38
rs765209942 Chr.16: 84174683-84174685 on Build GRCh38
rs746038599 Chr.16: 84174597-84174597 on Build GRCh38
rs751700362 Chr.16: 84174433-84174433 on Build GRCh38
rs990052423 Chr.16: 84174557-84174557 on Build GRCh38
rs772878634 Chr.16: 84174787-84174787 on Build GRCh38
rs188634781 Chr.16: 84174526-84174526 on Build GRCh38
rs576850509 Chr.16: 84174552-84174552 on Build GRCh38
rs377049642 Chr.16: 84174629-84174629 on Build GRCh38
rs990593941 Chr.16: 84174803-84174803 on Build GRCh38
rs763374253 Chr.16: 84174742-84174743 on Build GRCh38
rs776671436 Chr.16: 84174649-84174649 on Build GRCh38
rs560012159 Chr.16: 84174565-84174565 on Build GRCh38
rs371575198 Chr.16: 84174706-84174706 on Build GRCh38
rs200928272 Chr.16: 84174749-84174749 on Build GRCh38
rs1054964094 Chr.16: 84174410-84174410 on Build GRCh38
rs764423202 Chr.16: 84174772-84174772 on Build GRCh38
rs779727587 Chr.16: 84174709-84174709 on Build GRCh38
rs867181839 Chr.16: 84174453-84174453 on Build GRCh38
rs774002479 Chr.16: 84174669-84174669 on Build GRCh38
rs3803642 Chr.16: 84174538-84174538 on Build GRCh38
rs1008770656 Chr.16: 84174467-84174467 on Build GRCh38
rs767219566 Chr.16: 84174687-84174687 on Build GRCh38
rs192432193 Chr.16: 84174650-84174650 on Build GRCh38
rs117306571 Chr.16: 84174587-84174587 on Build GRCh38
rs3826151 Chr.16: 84174729-84174729 on Build GRCh38
rs550676070 Chr.16: 84174630-84174630 on Build GRCh38
rs139519641 Chr.16: 84174723-84174723 on Build GRCh38
rs3803643 Chr.16: 84174426-84174426 on Build GRCh38
rs1013479766 Chr.16: 84174499-84174499 on Build GRCh38
rs376368762 Chr.16: 84174690-84174690 on Build GRCh38
rs528405428 Chr.16: 84174489-84174489 on Build GRCh38
rs1023143448 Chr.16: 84174584-84174584 on Build GRCh38
rs894741241 Chr.16: 84174460-84174460 on Build GRCh38
rs567337940 Chr.16: 84174452-84174452 on Build GRCh38
rs761366675 Chr.16: 84174674-84174674 on Build GRCh38
rs986998262 Chr.16: 84174616-84174616 on Build GRCh38
rs373640751 Chr.16: 84174708-84174708 on Build GRCh38
rs768743954 Chr.16: 84174717-84174717 on Build GRCh38
rs747117537 Chr.16: 84174699-84174699 on Build GRCh38
rs772771852 Chr.16: 84174725-84174725 on Build GRCh38
rs139889264 Chr.16: 84174782-84174782 on Build GRCh38
rs191387291 Chr.16: 84174434-84174434 on Build GRCh38
rs775341712 Chr.16: 84174620-84174620 on Build GRCh38
rs907407703 Chr.16: 84174666-84174666 on Build GRCh38
rs184127602 Chr.16: 84174515-84174515 on Build GRCh38
rs576928019 Chr.16: 84174689-84174689 on Build GRCh38
rs199514761 Chr.16: 84174627-84174627 on Build GRCh38
rs763765269 Chr.16: 84174624-84174624 on Build GRCh38
rs1024479777 Chr.16: 84174512-84174512 on Build GRCh38
rs533280238 Chr.16: 84174643-84174643 on Build GRCh38
rs186999548 Chr.16: 84174382-84174382 on Build GRCh38
rs749025746 Chr.16: 84174714-84174714 on Build GRCh38
rs775351767 Chr.16: 84174660-84174660 on Build GRCh38
rs370600493 Chr.16: 84174662-84174662 on Build GRCh38
rs763259283 Chr.16: 84174752-84174752 on Build GRCh38
rs745624969 Chr.16: 84174395-84174395 on Build GRCh38
rs371678330 Chr.16: 84174731-84174731 on Build GRCh38
rs751102864 Chr.16: 84174626-84174626 on Build GRCh38
rs746144095 Chr.16: 84174707-84174707 on Build GRCh38
rs764066045 Chr.16: 84174667-84174667 on Build GRCh38
rs112364380 Chr.16: 84174492-84174492 on Build GRCh38
rs115253516 Chr.16: 84174646-84174646 on Build GRCh38
rs1050294964 Chr.16: 84174436-84174436 on Build GRCh38

View Full Product Details