Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89576017-89576550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_017023138.1 XM_017023139.1 XM_017023140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667919 c.153G>A p.Q51Q Chr.16: 89576050-89576050 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755565927 Chr.16: 89576035-89576035 on Build GRCh38
rs74590681 Chr.16: 89576050-89576050 on Build GRCh38
rs903116010 Chr.16: 89576510-89576510 on Build GRCh38
rs977147445 Chr.16: 89576354-89576354 on Build GRCh38
rs967264368 Chr.16: 89576334-89576334 on Build GRCh38
rs563473897 Chr.16: 89576440-89576440 on Build GRCh38
rs1046825207 Chr.16: 89576268-89576268 on Build GRCh38
rs950228353 Chr.16: 89576406-89576406 on Build GRCh38
rs1055975893 Chr.16: 89576164-89576164 on Build GRCh38
rs747038071 Chr.16: 89576080-89576080 on Build GRCh38
rs996343162 Chr.16: 89576315-89576315 on Build GRCh38
rs971829479 Chr.16: 89576173-89576173 on Build GRCh38
rs900788163 Chr.16: 89576314-89576314 on Build GRCh38
rs890766921 Chr.16: 89576082-89576082 on Build GRCh38
rs1018726100 Chr.16: 89576496-89576496 on Build GRCh38
rs77278584 Chr.16: 89576088-89576088 on Build GRCh38
rs964207494 Chr.16: 89576504-89576504 on Build GRCh38
rs555065710 Chr.16: 89576105-89576105 on Build GRCh38
rs893275507 Chr.16: 89576367-89576367 on Build GRCh38
rs943729995 Chr.16: 89576295-89576295 on Build GRCh38
rs1038373369 Chr.16: 89576438-89576438 on Build GRCh38
rs552739508 Chr.16: 89576241-89576241 on Build GRCh38
rs545216071 Chr.16: 89576257-89576257 on Build GRCh38
rs1038775534 Chr.16: 89576258-89576258 on Build GRCh38
rs530745102 Chr.16: 89576481-89576481 on Build GRCh38
rs908836542 Chr.16: 89576415-89576415 on Build GRCh38
rs543899872 Chr.16: 89576485-89576485 on Build GRCh38
rs912294616 Chr.16: 89576270-89576270 on Build GRCh38
rs745599606 Chr.16: 89576056-89576056 on Build GRCh38
rs1033057842 Chr.16: 89576363-89576363 on Build GRCh38
rs763350134 Chr.16: 89576120-89576120 on Build GRCh38
rs958881560 Chr.16: 89576452-89576452 on Build GRCh38
rs982598095 Chr.16: 89576493-89576493 on Build GRCh38
rs957452263 Chr.16: 89576136-89576136 on Build GRCh38
rs573268034 Chr.16: 89576116-89576116 on Build GRCh38
rs935742155 Chr.16: 89576144-89576144 on Build GRCh38
rs975485743 Chr.16: 89576523-89576523 on Build GRCh38
rs545009557 Chr.16: 89576422-89576422 on Build GRCh38
rs1039364205 Chr.16: 89576309-89576309 on Build GRCh38
rs950299533 Chr.16: 89576182-89576182 on Build GRCh38
rs373250085 Chr.16: 89576184-89576184 on Build GRCh38
rs916095560 Chr.16: 89576405-89576405 on Build GRCh38
rs953185122 Chr.16: 89576286-89576286 on Build GRCh38
rs887345852 Chr.16: 89576226-89576226 on Build GRCh38
rs776821630 Chr.16: 89576097-89576097 on Build GRCh38
rs577859195 Chr.16: 89576298-89576298 on Build GRCh38
rs1028842527 Chr.16: 89576285-89576285 on Build GRCh38
rs770099669 Chr.16: 89576112-89576112 on Build GRCh38
rs1018839624 Chr.16: 89576329-89576329 on Build GRCh38
rs911451360 Chr.16: 89576281-89576281 on Build GRCh38
rs1008671018 Chr.16: 89576310-89576310 on Build GRCh38
rs756948433 Chr.16: 89576053-89576053 on Build GRCh38
rs991623940 Chr.16: 89576386-89576386 on Build GRCh38
rs997297775 Chr.16: 89576284-89576284 on Build GRCh38
rs781026528 Chr.16: 89576362-89576362 on Build GRCh38
rs912334950 Chr.16: 89576505-89576509 on Build GRCh38
rs919723856 Chr.16: 89576249-89576249 on Build GRCh38
rs779844549 Chr.16: 89576077-89576077 on Build GRCh38
rs771872095 Chr.16: 89576076-89576076 on Build GRCh38
rs540626768 Chr.16: 89576177-89576177 on Build GRCh38
rs1033135862 Chr.16: 89576122-89576122 on Build GRCh38
rs1049829320 Chr.16: 89576475-89576475 on Build GRCh38
rs1010242183 Chr.16: 89576152-89576152 on Build GRCh38
rs898585811 Chr.16: 89576264-89576264 on Build GRCh38
rs1041048547 Chr.16: 89576089-89576089 on Build GRCh38
rs943800140 Chr.16: 89576513-89576513 on Build GRCh38
rs929726420 Chr.16: 89576256-89576256 on Build GRCh38
rs981590649 Chr.16: 89576409-89576409 on Build GRCh38
rs958649756 Chr.16: 89576236-89576236 on Build GRCh38
rs957146734 Chr.16: 89576375-89576375 on Build GRCh38
rs577281487 Chr.16: 89576194-89576194 on Build GRCh38
rs932763849 Chr.16: 89576463-89576463 on Build GRCh38
rs973958975 Chr.16: 89576468-89576468 on Build GRCh38
rs562174975 Chr.16: 89576492-89576492 on Build GRCh38
rs879506552 Chr.16: 89576480-89576480 on Build GRCh38
rs753398994 Chr.16: 89576052-89576052 on Build GRCh38
rs1020253756 Chr.16: 89576171-89576171 on Build GRCh38
rs907279146 Chr.16: 89576388-89576388 on Build GRCh38
rs187729343 Chr.16: 89576187-89576187 on Build GRCh38

View Full Product Details