Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32668837-32669352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
UniGene
Hs.16003
Gene Chromosome Location
Chr.1: 32651148-32686211 on Build GRCh38
Transcripts
NM_001135255.1 NM_001135256.1 NM_005610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6901944 c.805G>A p.V269I Chr.1: 32669274-32669274 on Build GRCh38
5053406 c.808G>A p.D270N Chr.1: 32669277-32669277 on Build GRCh38
5053405 c.592G>C p.D198H Chr.1: 32668846-32668846 on Build GRCh38
3386089 c.595G>A p.D199N Chr.1: 32668849-32668849 on Build GRCh38
681094 c.847C>T p.P283S Chr.1: 32669316-32669316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200625432 Chr.1: 32668948-32668948 on Build GRCh38
rs201113457 Chr.1: 32668884-32668884 on Build GRCh38
rs751933323 Chr.1: 32669180-32669180 on Build GRCh38
rs371771007 Chr.1: 32669020-32669020 on Build GRCh38
rs762374569 Chr.1: 32669210-32669213 on Build GRCh38
rs1027104603 Chr.1: 32669164-32669164 on Build GRCh38
rs564889648 Chr.1: 32669196-32669196 on Build GRCh38
rs929716749 Chr.1: 32669279-32669279 on Build GRCh38
rs747765068 Chr.1: 32668924-32668924 on Build GRCh38
rs34196891 Chr.1: 32668999-32668999 on Build GRCh38
rs777339760 Chr.1: 32669121-32669121 on Build GRCh38
rs915764036 Chr.1: 32668873-32668873 on Build GRCh38
rs1046851546 Chr.1: 32669303-32669303 on Build GRCh38
rs369281857 Chr.1: 32669163-32669163 on Build GRCh38
rs911166029 Chr.1: 32669185-32669185 on Build GRCh38
rs370455306 Chr.1: 32668934-32668934 on Build GRCh38
rs779417589 Chr.1: 32668870-32668870 on Build GRCh38
rs777286839 Chr.1: 32669211-32669211 on Build GRCh38
rs768158469 Chr.1: 32668906-32668906 on Build GRCh38
rs766731160 Chr.1: 32669174-32669174 on Build GRCh38
rs74064157 Chr.1: 32669321-32669321 on Build GRCh38
rs748908469 Chr.1: 32669212-32669212 on Build GRCh38
rs904474182 Chr.1: 32668903-32668903 on Build GRCh38
rs359955 Chr.1: 32669173-32669173 on Build GRCh38
rs770717103 Chr.1: 32668947-32668947 on Build GRCh38
rs762114793 Chr.1: 32668990-32668990 on Build GRCh38
rs756927602 Chr.1: 32669213-32669213 on Build GRCh38
rs769306288 Chr.1: 32669106-32669106 on Build GRCh38
rs770736989 Chr.1: 32669150-32669150 on Build GRCh38
rs747252061 Chr.1: 32669184-32669185 on Build GRCh38
rs952986508 Chr.1: 32669172-32669172 on Build GRCh38
rs768948867 Chr.1: 32669200-32669201 on Build GRCh38
rs569847693 Chr.1: 32668864-32668864 on Build GRCh38
rs201750273 Chr.1: 32669152-32669152 on Build GRCh38
rs1015225154 Chr.1: 32669179-32669179 on Build GRCh38
rs111859546 Chr.1: 32669213-32669214 on Build GRCh38
rs77147900 Chr.1: 32669015-32669015 on Build GRCh38
rs368708643 Chr.1: 32668945-32668945 on Build GRCh38
rs775532306 Chr.1: 32669226-32669226 on Build GRCh38
rs60231808 Chr.1: 32669222-32669222 on Build GRCh38
rs752080971 Chr.1: 32669028-32669028 on Build GRCh38
rs763229529 Chr.1: 32669285-32669285 on Build GRCh38
rs561809945 Chr.1: 32669270-32669270 on Build GRCh38
rs780746346 Chr.1: 32669052-32669052 on Build GRCh38
rs748969873 Chr.1: 32669148-32669148 on Build GRCh38
rs758931608 Chr.1: 32669214-32669214 on Build GRCh38
rs776980235 Chr.1: 32669201-32669201 on Build GRCh38
rs755580900 Chr.1: 32669207-32669207 on Build GRCh38
rs376744476 Chr.1: 32669160-32669160 on Build GRCh38
rs774054739 Chr.1: 32669151-32669151 on Build GRCh38
rs758800115 Chr.1: 32669013-32669013 on Build GRCh38
rs374282585 Chr.1: 32668923-32668923 on Build GRCh38
rs750877935 Chr.1: 32668872-32668872 on Build GRCh38
rs547545024 Chr.1: 32669251-32669251 on Build GRCh38
rs765716339 Chr.1: 32669213-32669213 on Build GRCh38
rs767318199 Chr.1: 32668881-32668881 on Build GRCh38
rs771962733 Chr.1: 32669155-32669155 on Build GRCh38
rs755495114 Chr.1: 32669189-32669189 on Build GRCh38
rs1052460422 Chr.1: 32668940-32668940 on Build GRCh38
rs780300213 Chr.1: 32669025-32669025 on Build GRCh38
rs764203653 Chr.1: 32668974-32668974 on Build GRCh38
rs376838717 Chr.1: 32668968-32668968 on Build GRCh38
rs575009341 Chr.1: 32668977-32668977 on Build GRCh38
rs771907999 Chr.1: 32669224-32669224 on Build GRCh38
rs755475560 Chr.1: 32669036-32669036 on Build GRCh38
rs767080361 Chr.1: 32669222-32669223 on Build GRCh38
rs777470641 Chr.1: 32669191-32669191 on Build GRCh38
rs536898667 Chr.1: 32668867-32668867 on Build GRCh38
rs757594918 Chr.1: 32668868-32668868 on Build GRCh38
rs751490523 Chr.1: 32669242-32669242 on Build GRCh38
rs576789637 Chr.1: 32668907-32668907 on Build GRCh38
rs867168487 Chr.1: 32668997-32668997 on Build GRCh38
rs201443101 Chr.1: 32668957-32668957 on Build GRCh38
rs772975974 Chr.1: 32668941-32668941 on Build GRCh38
rs778757806 Chr.1: 32669214-32669214 on Build GRCh38
rs747712240 Chr.1: 32669097-32669097 on Build GRCh38
rs765501303 Chr.1: 32668998-32668998 on Build GRCh38
rs11551266 Chr.1: 32669264-32669264 on Build GRCh38
rs7538661 Chr.1: 32669324-32669324 on Build GRCh38
rs760741172 Chr.1: 32668863-32668863 on Build GRCh38
rs746969572 Chr.1: 32669227-32669227 on Build GRCh38
rs760124363 Chr.1: 32669143-32669143 on Build GRCh38
rs758851159 Chr.1: 32668877-32668877 on Build GRCh38

View Full Product Details