Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41017514-41018006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFNL1
Entrez Gene ID
200172
Gene Name
schlafen like 1
Gene Aliases
-
UniGene
Hs.194609
Gene Chromosome Location
Chr.1: 41015597-41023444 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Gene Symbol
SLFNL1-AS1
Entrez Gene ID
100507178
Gene Name
SLFNL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660056
Gene Chromosome Location
Chr.1: 41014590-41043890 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1226660 c.778G>A p.V260M Chr.1: 41017814-41017814 on Build GRCh38
1127014 c.767G>A p.G256D Chr.1: 41017825-41017825 on Build GRCh38
4007984 c.618C>T p.I206I Chr.1: 41017974-41017974 on Build GRCh38
5934422 c.957G>A p.K319K Chr.1: 41017635-41017635 on Build GRCh38
4007983 c.768C>T p.G256G Chr.1: 41017824-41017824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781198806 Chr.1: 41017805-41017805 on Build GRCh38
rs540394421 Chr.1: 41017800-41017800 on Build GRCh38
rs751023739 Chr.1: 41017767-41017767 on Build GRCh38
rs753977265 Chr.1: 41017909-41017909 on Build GRCh38
rs772975519 Chr.1: 41017956-41017956 on Build GRCh38
rs773002577 Chr.1: 41017706-41017706 on Build GRCh38
rs200045066 Chr.1: 41017703-41017703 on Build GRCh38
rs370312980 Chr.1: 41017695-41017695 on Build GRCh38
rs746509813 Chr.1: 41017873-41017873 on Build GRCh38
rs776695336 Chr.1: 41017760-41017760 on Build GRCh38
rs745713064 Chr.1: 41017866-41017866 on Build GRCh38
rs571291332 Chr.1: 41017861-41017861 on Build GRCh38
rs771200715 Chr.1: 41017862-41017862 on Build GRCh38
rs139107002 Chr.1: 41017859-41017859 on Build GRCh38
rs536545640 Chr.1: 41017914-41017914 on Build GRCh38
rs747095718 Chr.1: 41017688-41017688 on Build GRCh38
rs747097546 Chr.1: 41017753-41017753 on Build GRCh38
rs369339564 Chr.1: 41017761-41017761 on Build GRCh38
rs766874421 Chr.1: 41017785-41017785 on Build GRCh38
rs981259360 Chr.1: 41017685-41017685 on Build GRCh38
rs376140034 Chr.1: 41017857-41017857 on Build GRCh38
rs1016489188 Chr.1: 41017737-41017737 on Build GRCh38
rs200423444 Chr.1: 41017802-41017802 on Build GRCh38
rs768735444 Chr.1: 41017937-41017937 on Build GRCh38
rs774550200 Chr.1: 41017896-41017896 on Build GRCh38
rs779998681 Chr.1: 41017931-41017931 on Build GRCh38
rs886324277 Chr.1: 41017728-41017728 on Build GRCh38
rs761321171 Chr.1: 41017882-41017882 on Build GRCh38
rs576391406 Chr.1: 41017647-41017647 on Build GRCh38
rs567621532 Chr.1: 41017913-41017913 on Build GRCh38
rs755520675 Chr.1: 41017804-41017804 on Build GRCh38
rs752468552 Chr.1: 41017745-41017745 on Build GRCh38
rs768685854 Chr.1: 41017758-41017758 on Build GRCh38
rs772476700 Chr.1: 41017681-41017681 on Build GRCh38
rs765145753 Chr.1: 41017614-41017614 on Build GRCh38
rs779189966 Chr.1: 41017582-41017585 on Build GRCh38
rs760360436 Chr.1: 41017791-41017791 on Build GRCh38
rs377572340 Chr.1: 41017962-41017962 on Build GRCh38
rs368020789 Chr.1: 41017853-41017853 on Build GRCh38
rs767190732 Chr.1: 41017744-41017744 on Build GRCh38
rs754495740 Chr.1: 41017740-41017740 on Build GRCh38
rs762830136 Chr.1: 41017719-41017719 on Build GRCh38
rs375640131 Chr.1: 41017625-41017625 on Build GRCh38
rs770879895 Chr.1: 41017771-41017771 on Build GRCh38
rs150541088 Chr.1: 41017865-41017865 on Build GRCh38
rs1023383586 Chr.1: 41017889-41017889 on Build GRCh38
rs770027784 Chr.1: 41017763-41017763 on Build GRCh38
rs780471752 Chr.1: 41017752-41017752 on Build GRCh38
rs942753678 Chr.1: 41017687-41017687 on Build GRCh38
rs759336749 Chr.1: 41017815-41017815 on Build GRCh38
rs906250682 Chr.1: 41017892-41017892 on Build GRCh38
rs746313380 Chr.1: 41017670-41017670 on Build GRCh38
rs74071110 Chr.1: 41017698-41017698 on Build GRCh38
rs773255974 Chr.1: 41017907-41017907 on Build GRCh38
rs77061292 Chr.1: 41017656-41017656 on Build GRCh38
rs775596812 Chr.1: 41017684-41017684 on Build GRCh38
rs759216038 Chr.1: 41017663-41017663 on Build GRCh38
rs146420187 Chr.1: 41017765-41017765 on Build GRCh38
rs761488527 Chr.1: 41017801-41017803 on Build GRCh38
rs150044513 Chr.1: 41017906-41017906 on Build GRCh38
rs970999491 Chr.1: 41017564-41017564 on Build GRCh38
rs758334308 Chr.1: 41017622-41017622 on Build GRCh38
rs750588926 Chr.1: 41017980-41017980 on Build GRCh38
rs964556609 Chr.1: 41017917-41017917 on Build GRCh38
rs763821111 Chr.1: 41017839-41017839 on Build GRCh38
rs779695625 Chr.1: 41017930-41017930 on Build GRCh38
rs112014176 Chr.1: 41017643-41017643 on Build GRCh38
rs750425534 Chr.1: 41017621-41017621 on Build GRCh38
rs141710904 Chr.1: 41017640-41017640 on Build GRCh38
rs768214751 Chr.1: 41017874-41017874 on Build GRCh38
rs376479010 Chr.1: 41017731-41017731 on Build GRCh38
rs751943831 Chr.1: 41017803-41017803 on Build GRCh38
rs138195003 Chr.1: 41017814-41017814 on Build GRCh38
rs182933235 Chr.1: 41017854-41017854 on Build GRCh38
rs752272148 Chr.1: 41017790-41017790 on Build GRCh38
rs957135235 Chr.1: 41017784-41017784 on Build GRCh38
rs763876241 Chr.1: 41017796-41017796 on Build GRCh38
rs554063327 Chr.1: 41017542-41017542 on Build GRCh38
rs761645031 Chr.1: 41017612-41017612 on Build GRCh38
rs753836544 Chr.1: 41017973-41017973 on Build GRCh38
rs767807067 Chr.1: 41017965-41017965 on Build GRCh38
rs751050866 Chr.1: 41017738-41017738 on Build GRCh38
rs760307090 Chr.1: 41017832-41017832 on Build GRCh38
rs762696363 Chr.1: 41017766-41017766 on Build GRCh38
rs756245710 Chr.1: 41017858-41017858 on Build GRCh38
rs758330920 Chr.1: 41017925-41017925 on Build GRCh38
rs750790060 Chr.1: 41017797-41017797 on Build GRCh38
rs769311448 Chr.1: 41017948-41017948 on Build GRCh38
rs41268103 Chr.1: 41017829-41017829 on Build GRCh38
rs980594091 Chr.1: 41017835-41017835 on Build GRCh38
rs879793589 Chr.1: 41017538-41017538 on Build GRCh38
rs556560244 Chr.1: 41017966-41017966 on Build GRCh38
rs762865549 Chr.1: 41017563-41017563 on Build GRCh38
rs758706343 Chr.1: 41017983-41017983 on Build GRCh38
rs759086428 Chr.1: 41017778-41017778 on Build GRCh38
rs776452085 Chr.1: 41017597-41017597 on Build GRCh38
rs779562511 Chr.1: 41017657-41017657 on Build GRCh38
rs142668626 Chr.1: 41017941-41017941 on Build GRCh38
rs768896298 Chr.1: 41017693-41017693 on Build GRCh38
rs778018968 Chr.1: 41017809-41017809 on Build GRCh38
rs772110855 Chr.1: 41017869-41017869 on Build GRCh38
rs569404071 Chr.1: 41017754-41017754 on Build GRCh38
rs761123920 Chr.1: 41017969-41017969 on Build GRCh38
rs764695402 Chr.1: 41017972-41017972 on Build GRCh38
rs776371180 Chr.1: 41017639-41017640 on Build GRCh38
rs139214565 Chr.1: 41017697-41017697 on Build GRCh38
rs202080004 Chr.1: 41017808-41017808 on Build GRCh38
rs372854218 Chr.1: 41017826-41017826 on Build GRCh38
rs761531145 Chr.1: 41017847-41017847 on Build GRCh38
rs949269816 Chr.1: 41017665-41017665 on Build GRCh38
rs886376714 Chr.1: 41017850-41017850 on Build GRCh38
rs775622500 Chr.1: 41017870-41017870 on Build GRCh38
rs748260284 Chr.1: 41017806-41017806 on Build GRCh38
rs373696681 Chr.1: 41017653-41017653 on Build GRCh38
rs752229764 Chr.1: 41017748-41017748 on Build GRCh38
rs767999694 Chr.1: 41017898-41017898 on Build GRCh38
rs151120121 Chr.1: 41017742-41017742 on Build GRCh38
rs746537061 Chr.1: 41017589-41017595 on Build GRCh38
rs369745596 Chr.1: 41017968-41017968 on Build GRCh38
rs201917045 Chr.1: 41017895-41017895 on Build GRCh38
rs141850103 Chr.1: 41017834-41017834 on Build GRCh38
rs370243333 Chr.1: 41017634-41017634 on Build GRCh38
rs753610727 Chr.1: 41017846-41017846 on Build GRCh38
rs746264338 Chr.1: 41017747-41017747 on Build GRCh38
rs780747853 Chr.1: 41017646-41017646 on Build GRCh38
rs143152054 Chr.1: 41017777-41017777 on Build GRCh38
rs751975242 Chr.1: 41017851-41017851 on Build GRCh38
rs372756325 Chr.1: 41017880-41017880 on Build GRCh38
rs956572234 Chr.1: 41017547-41017547 on Build GRCh38
rs755835817 Chr.1: 41017746-41017746 on Build GRCh38
rs757413470 Chr.1: 41017974-41017974 on Build GRCh38
rs982396911 Chr.1: 41017773-41017773 on Build GRCh38
rs746980294 Chr.1: 41017766-41017766 on Build GRCh38
rs963420019 Chr.1: 41017537-41017537 on Build GRCh38
rs758903507 Chr.1: 41017751-41017751 on Build GRCh38
rs956989250 Chr.1: 41017899-41017899 on Build GRCh38
rs766593436 Chr.1: 41017848-41017848 on Build GRCh38
rs745632859 Chr.1: 41017924-41017924 on Build GRCh38
rs768156659 Chr.1: 41017638-41017640 on Build GRCh38
rs769749051 Chr.1: 41017911-41017912 on Build GRCh38
rs749653149 Chr.1: 41017648-41017648 on Build GRCh38
rs982344352 Chr.1: 41017577-41017577 on Build GRCh38
rs374489586 Chr.1: 41017908-41017908 on Build GRCh38

View Full Product Details