Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13933100-13933596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
UniGene
Hs.448497
Gene Chromosome Location
Chr.19: 13931187-13953392 on Build GRCh38
Transcripts
NM_001146254.1 NM_001146255.1 NM_024825.3 XM_006722901.1 XM_006722902.1 XM_006722903.1 XM_011528308.1 XM_011528309.2 XM_011528310.2 XM_011528311.1 XM_011528313.1 XM_011528314.1 XM_011528316.1 XM_017027313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6280611 c.825C>A p.H275Q Chr.19: 13933419-13933419 on Build GRCh38
1611650 c.1072C>T p.R358C Chr.19: 13933172-13933172 on Build GRCh38
4796815 c.995C>T p.A332V Chr.19: 13933249-13933249 on Build GRCh38
5474072 c.941G>A p.R314H Chr.19: 13933303-13933303 on Build GRCh38
4074727 c.925G>A p.G309R Chr.19: 13933319-13933319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369602527 Chr.19: 13933473-13933473 on Build GRCh38
rs1054984938 Chr.19: 13933479-13933479 on Build GRCh38
rs375379231 Chr.19: 13933256-13933256 on Build GRCh38
rs372890896 Chr.19: 13933320-13933320 on Build GRCh38
rs545216208 Chr.19: 13933537-13933537 on Build GRCh38
rs751682655 Chr.19: 13933359-13933359 on Build GRCh38
rs757289801 Chr.19: 13933454-13933454 on Build GRCh38
rs1008133197 Chr.19: 13933238-13933238 on Build GRCh38
rs781015023 Chr.19: 13933398-13933406 on Build GRCh38
rs781430307 Chr.19: 13933372-13933372 on Build GRCh38
rs376362504 Chr.19: 13933169-13933169 on Build GRCh38
rs767820910 Chr.19: 13933239-13933239 on Build GRCh38
rs759960334 Chr.19: 13933257-13933257 on Build GRCh38
rs981363964 Chr.19: 13933499-13933499 on Build GRCh38
rs761340811 Chr.19: 13933281-13933281 on Build GRCh38
rs73515522 Chr.19: 13933574-13933574 on Build GRCh38
rs368240194 Chr.19: 13933315-13933315 on Build GRCh38
rs754594319 Chr.19: 13933385-13933385 on Build GRCh38
rs755008910 Chr.19: 13933235-13933235 on Build GRCh38
rs368368090 Chr.19: 13933215-13933215 on Build GRCh38
rs773385161 Chr.19: 13933249-13933249 on Build GRCh38
rs761936290 Chr.19: 13933160-13933160 on Build GRCh38
rs373561110 Chr.19: 13933230-13933230 on Build GRCh38
rs577780298 Chr.19: 13933518-13933518 on Build GRCh38
rs35418329 Chr.19: 13933538-13933539 on Build GRCh38
rs1020368381 Chr.19: 13933350-13933350 on Build GRCh38
rs753706216 Chr.19: 13933313-13933313 on Build GRCh38
rs951089345 Chr.19: 13933162-13933162 on Build GRCh38
rs567613608 Chr.19: 13933314-13933314 on Build GRCh38
rs745701176 Chr.19: 13933118-13933118 on Build GRCh38
rs193206432 Chr.19: 13933228-13933228 on Build GRCh38
rs903294147 Chr.19: 13933221-13933221 on Build GRCh38
rs982627421 Chr.19: 13933163-13933163 on Build GRCh38
rs746530403 Chr.19: 13933343-13933343 on Build GRCh38
rs549358408 Chr.19: 13933269-13933269 on Build GRCh38
rs1016720706 Chr.19: 13933470-13933470 on Build GRCh38
rs1055752487 Chr.19: 13933180-13933180 on Build GRCh38
rs933916089 Chr.19: 13933446-13933446 on Build GRCh38
rs780337115 Chr.19: 13933384-13933384 on Build GRCh38
rs746295709 Chr.19: 13933328-13933328 on Build GRCh38
rs35714771 Chr.19: 13933420-13933421 on Build GRCh38
rs1010240692 Chr.19: 13933544-13933544 on Build GRCh38
rs781043502 Chr.19: 13933213-13933213 on Build GRCh38
rs934660794 Chr.19: 13933335-13933335 on Build GRCh38
rs757452688 Chr.19: 13933369-13933369 on Build GRCh38
rs981742518 Chr.19: 13933250-13933250 on Build GRCh38
rs755434797 Chr.19: 13933200-13933200 on Build GRCh38
rs756759524 Chr.19: 13933348-13933349 on Build GRCh38
rs997881805 Chr.19: 13933244-13933244 on Build GRCh38
rs750035660 Chr.19: 13933289-13933289 on Build GRCh38
rs924524226 Chr.19: 13933509-13933509 on Build GRCh38
rs572326843 Chr.19: 13933352-13933352 on Build GRCh38
rs1003872448 Chr.19: 13933436-13933436 on Build GRCh38
rs922372621 Chr.19: 13933157-13933157 on Build GRCh38
rs760372644 Chr.19: 13933344-13933344 on Build GRCh38
rs764836559 Chr.19: 13933398-13933398 on Build GRCh38
rs751522801 Chr.19: 13933449-13933449 on Build GRCh38
rs2305778 Chr.19: 13933474-13933474 on Build GRCh38
rs776833225 Chr.19: 13933327-13933327 on Build GRCh38
rs774547846 Chr.19: 13933149-13933149 on Build GRCh38
rs562783628 Chr.19: 13933191-13933191 on Build GRCh38
rs754277907 Chr.19: 13933175-13933175 on Build GRCh38
rs562723161 Chr.19: 13933496-13933496 on Build GRCh38
rs754760460 Chr.19: 13933346-13933346 on Build GRCh38
rs769672864 Chr.19: 13933500-13933501 on Build GRCh38
rs10423549 Chr.19: 13933254-13933254 on Build GRCh38
rs763293497 Chr.19: 13933351-13933351 on Build GRCh38
rs749163395 Chr.19: 13933234-13933234 on Build GRCh38
rs146550820 Chr.19: 13933401-13933401 on Build GRCh38
rs575072620 Chr.19: 13933558-13933558 on Build GRCh38
rs762850419 Chr.19: 13933440-13933440 on Build GRCh38
rs1054859305 Chr.19: 13933304-13933304 on Build GRCh38
rs777481907 Chr.19: 13933301-13933301 on Build GRCh38
rs369579003 Chr.19: 13933493-13933493 on Build GRCh38
rs776063589 Chr.19: 13933507-13933507 on Build GRCh38
rs751151283 Chr.19: 13933193-13933193 on Build GRCh38
rs772919396 Chr.19: 13933337-13933337 on Build GRCh38
rs371819749 Chr.19: 13933391-13933391 on Build GRCh38
rs554074866 Chr.19: 13933348-13933348 on Build GRCh38
rs576049307 Chr.19: 13933459-13933459 on Build GRCh38
rs1055371525 Chr.19: 13933137-13933137 on Build GRCh38
rs772160187 Chr.19: 13933243-13933243 on Build GRCh38
rs765069020 Chr.19: 13933416-13933416 on Build GRCh38
rs202013571 Chr.19: 13933168-13933168 on Build GRCh38
rs571493723 Chr.19: 13933333-13933333 on Build GRCh38
rs752555756 Chr.19: 13933439-13933439 on Build GRCh38
rs560575571 Chr.19: 13933555-13933555 on Build GRCh38
rs746111010 Chr.19: 13933376-13933376 on Build GRCh38
rs200487966 Chr.19: 13933136-13933136 on Build GRCh38
rs201236546 Chr.19: 13933129-13933129 on Build GRCh38
rs755885466 Chr.19: 13933303-13933303 on Build GRCh38
rs773715294 Chr.19: 13933280-13933280 on Build GRCh38
rs777826005 Chr.19: 13933358-13933358 on Build GRCh38
rs767137374 Chr.19: 13933285-13933285 on Build GRCh38
rs144261976 Chr.19: 13933394-13933394 on Build GRCh38
rs769322459 Chr.19: 13933388-13933388 on Build GRCh38
rs758408194 Chr.19: 13933319-13933319 on Build GRCh38
rs1016688224 Chr.19: 13933312-13933312 on Build GRCh38
rs763503040 Chr.19: 13933192-13933192 on Build GRCh38
rs760213002 Chr.19: 13933395-13933395 on Build GRCh38
rs767118816 Chr.19: 13933336-13933336 on Build GRCh38
rs554562507 Chr.19: 13933404-13933404 on Build GRCh38
rs868695775 Chr.19: 13933262-13933264 on Build GRCh38
rs758389798 Chr.19: 13933357-13933357 on Build GRCh38
rs200314809 Chr.19: 13933345-13933345 on Build GRCh38
rs748307018 Chr.19: 13933240-13933240 on Build GRCh38
rs915805552 Chr.19: 13933190-13933190 on Build GRCh38
rs756831541 Chr.19: 13933194-13933194 on Build GRCh38
rs79400921 Chr.19: 13933199-13933199 on Build GRCh38
rs963781438 Chr.19: 13933130-13933130 on Build GRCh38
rs199509128 Chr.19: 13933332-13933332 on Build GRCh38
rs780431360 Chr.19: 13933477-13933477 on Build GRCh38
rs768557278 Chr.19: 13933146-13933146 on Build GRCh38
rs941050885 Chr.19: 13933397-13933397 on Build GRCh38
rs763717991 Chr.19: 13933461-13933463 on Build GRCh38
rs760980835 Chr.19: 13933171-13933171 on Build GRCh38
rs374900850 Chr.19: 13933410-13933410 on Build GRCh38
rs750129577 Chr.19: 13933222-13933222 on Build GRCh38
rs371711931 Chr.19: 13933253-13933253 on Build GRCh38
rs766272609 Chr.19: 13933311-13933311 on Build GRCh38
rs934691511 Chr.19: 13933519-13933519 on Build GRCh38
rs766762709 Chr.19: 13933172-13933172 on Build GRCh38
rs888257358 Chr.19: 13933148-13933148 on Build GRCh38
rs201972395 Chr.19: 13933492-13933492 on Build GRCh38
rs770418095 Chr.19: 13933500-13933500 on Build GRCh38
rs371100559 Chr.19: 13933131-13933131 on Build GRCh38
rs35632054 Chr.19: 13933494-13933495 on Build GRCh38

View Full Product Details