Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49861680-49862185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNKP
Entrez Gene ID
11284
Gene Name
polynucleotide kinase 3'-phosphatase
Gene Aliases
AOA4, EIEE10, MCSZ, PNK
UniGene
Hs.78016
Gene Chromosome Location
Chr.19: 49861203-49867565 on Build GRCh38
Transcripts
NM_007254.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1161287 c.1178A>C p.H393P Chr.19: 49862054-49862054 on Build GRCh38
6751261 c.1238T>C p.L413P Chr.19: 49861832-49861832 on Build GRCh38
4080227 c.1179C>T p.H393H Chr.19: 49862053-49862053 on Build GRCh38
6751251 c.1168G>A p.G390R Chr.19: 49862064-49862064 on Build GRCh38
6231140 c.1285G>A p.A429T Chr.19: 49861785-49861785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764379536 Chr.19: 49861779-49861792 on Build GRCh38
rs373524398 Chr.19: 49861792-49861792 on Build GRCh38
rs769761063 Chr.19: 49861719-49861719 on Build GRCh38
rs776617733 Chr.19: 49861766-49861767 on Build GRCh38
rs193234253 Chr.19: 49862147-49862147 on Build GRCh38
rs768249162 Chr.19: 49861925-49861925 on Build GRCh38
rs769707108 Chr.19: 49861784-49861784 on Build GRCh38
rs1038282650 Chr.19: 49861982-49861982 on Build GRCh38
rs748938228 Chr.19: 49862162-49862162 on Build GRCh38
rs768847609 Chr.19: 49861811-49861812 on Build GRCh38
rs751609401 Chr.19: 49861811-49861811 on Build GRCh38
rs587784366 Chr.19: 49861766-49861775 on Build GRCh38
rs796052861 Chr.19: 49861769-49861770 on Build GRCh38
rs748420867 Chr.19: 49861781-49861781 on Build GRCh38
rs765575612 Chr.19: 49861799-49861799 on Build GRCh38
rs899556646 Chr.19: 49861868-49861868 on Build GRCh38
rs1004562051 Chr.19: 49862158-49862158 on Build GRCh38
rs746413622 Chr.19: 49861786-49861787 on Build GRCh38
rs747698126 Chr.19: 49862041-49862041 on Build GRCh38
rs763250528 Chr.19: 49861871-49861871 on Build GRCh38
rs779474259 Chr.19: 49861922-49861922 on Build GRCh38
rs967589662 Chr.19: 49862072-49862072 on Build GRCh38
rs369003964 Chr.19: 49861813-49861813 on Build GRCh38
rs756362768 Chr.19: 49861892-49861892 on Build GRCh38
rs573674802 Chr.19: 49861972-49861972 on Build GRCh38
rs756988139 Chr.19: 49862163-49862163 on Build GRCh38
rs563918674 Chr.19: 49862046-49862046 on Build GRCh38
rs763782151 Chr.19: 49862036-49862036 on Build GRCh38
rs937360079 Chr.19: 49861936-49861936 on Build GRCh38
rs575803996 Chr.19: 49862067-49862067 on Build GRCh38
rs780736753 Chr.19: 49862032-49862032 on Build GRCh38
rs903644862 Chr.19: 49861942-49861942 on Build GRCh38
rs967919895 Chr.19: 49861740-49861740 on Build GRCh38
rs775222885 Chr.19: 49862009-49862011 on Build GRCh38
rs777641575 Chr.19: 49861818-49861818 on Build GRCh38
rs754921430 Chr.19: 49862088-49862088 on Build GRCh38
rs771186109 Chr.19: 49861999-49861999 on Build GRCh38
rs761732328 Chr.19: 49861708-49861708 on Build GRCh38
rs528514147 Chr.19: 49861910-49861910 on Build GRCh38
rs752004904 Chr.19: 49861876-49861876 on Build GRCh38
rs786205207 Chr.19: 49861847-49861849 on Build GRCh38
rs757518270 Chr.19: 49862111-49862112 on Build GRCh38
rs751871245 Chr.19: 49862038-49862048 on Build GRCh38
rs766203926 Chr.19: 49862082-49862082 on Build GRCh38
rs773745801 Chr.19: 49861732-49861732 on Build GRCh38
rs765068023 Chr.19: 49861768-49861776 on Build GRCh38
rs797045892 Chr.19: 49861767-49861768 on Build GRCh38
rs765192654 Chr.19: 49861995-49861996 on Build GRCh38
rs764305482 Chr.19: 49862018-49862018 on Build GRCh38
rs893076827 Chr.19: 49861774-49861787 on Build GRCh38
rs754158913 Chr.19: 49861904-49861904 on Build GRCh38
rs200470735 Chr.19: 49861746-49861746 on Build GRCh38
rs541840060 Chr.19: 49861796-49861796 on Build GRCh38
rs756416098 Chr.19: 49861815-49861815 on Build GRCh38
rs773452413 Chr.19: 49862144-49862144 on Build GRCh38
rs776536622 Chr.19: 49862124-49862124 on Build GRCh38
rs766816785 Chr.19: 49861874-49861874 on Build GRCh38
rs758168476 Chr.19: 49862077-49862077 on Build GRCh38
rs560951767 Chr.19: 49861901-49861901 on Build GRCh38
rs368832563 Chr.19: 49861900-49861903 on Build GRCh38
rs776097247 Chr.19: 49861920-49861921 on Build GRCh38
rs143390809 Chr.19: 49861777-49861777 on Build GRCh38
rs886054583 Chr.19: 49861836-49861839 on Build GRCh38
rs746555608 Chr.19: 49861924-49861924 on Build GRCh38
rs758965167 Chr.19: 49861755-49861755 on Build GRCh38
rs730882224 Chr.19: 49861819-49861820 on Build GRCh38
rs1055654261 Chr.19: 49861704-49861704 on Build GRCh38
rs768304312 Chr.19: 49861860-49861860 on Build GRCh38
rs926815534 Chr.19: 49862001-49862001 on Build GRCh38
rs950502049 Chr.19: 49861778-49861778 on Build GRCh38
rs760249644 Chr.19: 49861765-49861766 on Build GRCh38
rs770775129 Chr.19: 49861789-49861789 on Build GRCh38
rs761335711 Chr.19: 49862064-49862064 on Build GRCh38
rs778516383 Chr.19: 49862021-49862021 on Build GRCh38
rs772610025 Chr.19: 49862055-49862055 on Build GRCh38
rs772063359 Chr.19: 49861918-49861918 on Build GRCh38
rs752773107 Chr.19: 49862015-49862015 on Build GRCh38
rs765634741 Chr.19: 49861752-49861752 on Build GRCh38
rs760147150 Chr.19: 49861878-49861878 on Build GRCh38
rs991568285 Chr.19: 49861944-49861944 on Build GRCh38
rs376289875 Chr.19: 49862116-49862116 on Build GRCh38
rs746264747 Chr.19: 49862053-49862053 on Build GRCh38
rs769490709 Chr.19: 49862045-49862045 on Build GRCh38
rs1057518102 Chr.19: 49862043-49862043 on Build GRCh38
rs146311500 Chr.19: 49862069-49862069 on Build GRCh38
rs563802371 Chr.19: 49861836-49861836 on Build GRCh38
rs200776490 Chr.19: 49862150-49862150 on Build GRCh38
rs749177501 Chr.19: 49861786-49861786 on Build GRCh38
rs534639835 Chr.19: 49861973-49861973 on Build GRCh38
rs370937493 Chr.19: 49862151-49862151 on Build GRCh38
rs868384150 Chr.19: 49861838-49861838 on Build GRCh38
rs149614720 Chr.19: 49861771-49861771 on Build GRCh38
rs781413617 Chr.19: 49861786-49861786 on Build GRCh38
rs541129252 Chr.19: 49861753-49861753 on Build GRCh38
rs895695725 Chr.19: 49861722-49861722 on Build GRCh38
rs1042831313 Chr.19: 49862105-49862105 on Build GRCh38
rs3739203 Chr.19: 49862113-49862113 on Build GRCh38
rs3739205 Chr.19: 49861891-49861891 on Build GRCh38
rs761976639 Chr.19: 49862136-49862136 on Build GRCh38
rs759892733 Chr.19: 49862009-49862009 on Build GRCh38
rs796052860 Chr.19: 49861775-49861776 on Build GRCh38
rs578207030 Chr.19: 49861766-49861766 on Build GRCh38
rs776421692 Chr.19: 49861706-49861706 on Build GRCh38
rs758087762 Chr.19: 49861775-49861776 on Build GRCh38
rs555353376 Chr.19: 49861970-49861970 on Build GRCh38
rs756665207 Chr.19: 49861789-49861789 on Build GRCh38
rs777211623 Chr.19: 49862161-49862161 on Build GRCh38
rs569815553 Chr.19: 49861958-49861958 on Build GRCh38
rs759158211 Chr.19: 49861734-49861734 on Build GRCh38
rs780500233 Chr.19: 49862092-49862092 on Build GRCh38
rs774130130 Chr.19: 49861847-49861847 on Build GRCh38
rs908966809 Chr.19: 49861991-49861991 on Build GRCh38
rs375663661 Chr.19: 49861939-49861939 on Build GRCh38
rs755670029 Chr.19: 49862094-49862094 on Build GRCh38
rs373766090 Chr.19: 49862074-49862074 on Build GRCh38
rs111470432 Chr.19: 49861954-49861954 on Build GRCh38
rs112635688 Chr.19: 49861701-49861701 on Build GRCh38
rs559532107 Chr.19: 49861758-49861758 on Build GRCh38
rs377485860 Chr.19: 49861809-49861809 on Build GRCh38
rs763027558 Chr.19: 49861864-49861864 on Build GRCh38
rs141938129 Chr.19: 49861853-49861853 on Build GRCh38
rs779991747 Chr.19: 49862024-49862024 on Build GRCh38
rs983583870 Chr.19: 49861981-49861981 on Build GRCh38
rs890894773 Chr.19: 49862031-49862033 on Build GRCh38
rs536614762 Chr.19: 49861720-49861720 on Build GRCh38
rs763228709 Chr.19: 49862145-49862145 on Build GRCh38
rs748929352 Chr.19: 49862104-49862104 on Build GRCh38
rs936791908 Chr.19: 49862111-49862111 on Build GRCh38
rs768124502 Chr.19: 49862030-49862030 on Build GRCh38
rs537361270 Chr.19: 49861969-49861969 on Build GRCh38
rs750240376 Chr.19: 49862023-49862023 on Build GRCh38
rs964341783 Chr.19: 49861754-49861754 on Build GRCh38
rs772557014 Chr.19: 49862112-49862112 on Build GRCh38
rs780655453 Chr.19: 49861790-49861791 on Build GRCh38
rs113879990 Chr.19: 49861947-49861947 on Build GRCh38
rs925952327 Chr.19: 49861934-49861934 on Build GRCh38
rs750802489 Chr.19: 49861803-49861803 on Build GRCh38
rs758836805 Chr.19: 49861807-49861807 on Build GRCh38
rs774790818 Chr.19: 49862002-49862002 on Build GRCh38
rs771791338 Chr.19: 49861717-49861718 on Build GRCh38
rs796052855 Chr.19: 49862087-49862087 on Build GRCh38
rs762820617 Chr.19: 49862013-49862013 on Build GRCh38
rs201743589 Chr.19: 49862149-49862149 on Build GRCh38
rs545237601 Chr.19: 49862027-49862027 on Build GRCh38
rs759000111 Chr.19: 49861925-49861926 on Build GRCh38
rs370351286 Chr.19: 49862020-49862020 on Build GRCh38
rs587784365 Chr.19: 49861800-49861801 on Build GRCh38
rs755614377 Chr.19: 49862159-49862159 on Build GRCh38
rs757600464 Chr.19: 49861750-49861750 on Build GRCh38
rs776788871 Chr.19: 49862068-49862068 on Build GRCh38
rs747464508 Chr.19: 49862115-49862115 on Build GRCh38
rs3739204 Chr.19: 49861948-49861948 on Build GRCh38
rs745782148 Chr.19: 49861913-49861913 on Build GRCh38
rs761417360 Chr.19: 49861715-49861716 on Build GRCh38
rs769293635 Chr.19: 49862117-49862117 on Build GRCh38
rs772675208 Chr.19: 49861758-49861759 on Build GRCh38
rs758986802 Chr.19: 49861793-49861793 on Build GRCh38
rs555356993 Chr.19: 49861737-49861737 on Build GRCh38
rs1033301071 Chr.19: 49862125-49862125 on Build GRCh38
rs748365843 Chr.19: 49861814-49861814 on Build GRCh38
rs775042023 Chr.19: 49861858-49861858 on Build GRCh38
rs1001338139 Chr.19: 49861703-49861703 on Build GRCh38
rs770849181 Chr.19: 49861846-49861846 on Build GRCh38
rs146492255 Chr.19: 49861997-49861997 on Build GRCh38
rs770469920 Chr.19: 49861920-49861920 on Build GRCh38
rs794727126 Chr.19: 49861765-49861765 on Build GRCh38
rs752561267 Chr.19: 49861774-49861785 on Build GRCh38
rs777457079 Chr.19: 49862103-49862103 on Build GRCh38
rs546676673 Chr.19: 49861938-49861938 on Build GRCh38
rs751483948 Chr.19: 49862086-49862086 on Build GRCh38
rs988123606 Chr.19: 49861761-49861761 on Build GRCh38
rs757469639 Chr.19: 49861782-49861783 on Build GRCh38
rs372404688 Chr.19: 49861863-49861863 on Build GRCh38
rs199652836 Chr.19: 49861851-49861851 on Build GRCh38
rs1055603577 Chr.19: 49861804-49861804 on Build GRCh38
rs549069697 Chr.19: 49861859-49861859 on Build GRCh38
rs367695939 Chr.19: 49862006-49862006 on Build GRCh38
rs767107348 Chr.19: 49861795-49861795 on Build GRCh38
rs752622480 Chr.19: 49862154-49862154 on Build GRCh38
rs1010036717 Chr.19: 49861877-49861877 on Build GRCh38
rs751675183 Chr.19: 49861780-49861781 on Build GRCh38
rs752826211 Chr.19: 49861886-49861886 on Build GRCh38
rs765283582 Chr.19: 49862142-49862142 on Build GRCh38
rs776105055 Chr.19: 49862063-49862063 on Build GRCh38
rs983252097 Chr.19: 49861780-49861780 on Build GRCh38
rs747723740 Chr.19: 49862093-49862093 on Build GRCh38
rs557750182 Chr.19: 49861699-49861699 on Build GRCh38
rs765972315 Chr.19: 49861728-49861728 on Build GRCh38
rs768166074 Chr.19: 49862013-49862013 on Build GRCh38
rs1020456127 Chr.19: 49862071-49862071 on Build GRCh38
rs767645983 Chr.19: 49861883-49861883 on Build GRCh38
rs903931173 Chr.19: 49862106-49862106 on Build GRCh38
rs148386689 Chr.19: 49861785-49861785 on Build GRCh38
rs747065246 Chr.19: 49861760-49861760 on Build GRCh38
rs754981151 Chr.19: 49861812-49861812 on Build GRCh38
rs376011384 Chr.19: 49861879-49861879 on Build GRCh38
rs763050200 Chr.19: 49861727-49861727 on Build GRCh38
rs553083451 Chr.19: 49861992-49861992 on Build GRCh38

View Full Product Details