Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57472463-57472959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF772
Entrez Gene ID
400720
Gene Name
zinc finger protein 772
Gene Aliases
-
UniGene
Hs.388810
Gene Chromosome Location
Chr.19: 57464198-57480950 on Build GRCh38
Transcripts
NM_001024596.2 NM_001144068.1 XM_005258943.4 XM_005258944.3 XM_011526980.2 XM_017026813.1

SNP Detail

SNP ID Chromosome Location
rs534299577 Chr.19: 57472672-57472672 on Build GRCh38
rs568467712 Chr.19: 57472670-57472670 on Build GRCh38
rs182918848 Chr.19: 57472806-57472806 on Build GRCh38
rs960251618 Chr.19: 57472797-57472797 on Build GRCh38
rs980412189 Chr.19: 57472492-57472492 on Build GRCh38
rs978296210 Chr.19: 57472730-57472730 on Build GRCh38
rs10407309 Chr.19: 57472529-57472529 on Build GRCh38
rs10406544 Chr.19: 57472695-57472695 on Build GRCh38
rs1022118782 Chr.19: 57472735-57472735 on Build GRCh38
rs191687278 Chr.19: 57472779-57472779 on Build GRCh38
rs980489770 Chr.19: 57472758-57472758 on Build GRCh38
rs1048834062 Chr.19: 57472859-57472859 on Build GRCh38
rs1049284967 Chr.19: 57472599-57472599 on Build GRCh38
rs751280154 Chr.19: 57472809-57472809 on Build GRCh38
rs1025488938 Chr.19: 57472563-57472563 on Build GRCh38
rs929036874 Chr.19: 57472593-57472593 on Build GRCh38
rs769606692 Chr.19: 57472937-57472937 on Build GRCh38
rs1046860818 Chr.19: 57472928-57472928 on Build GRCh38
rs1037137792 Chr.19: 57472554-57472554 on Build GRCh38
rs541098762 Chr.19: 57472834-57472834 on Build GRCh38
rs10406621 Chr.19: 57472886-57472886 on Build GRCh38
rs952615626 Chr.19: 57472611-57472611 on Build GRCh38
rs910856950 Chr.19: 57472788-57472788 on Build GRCh38
rs10405925 Chr.19: 57472543-57472543 on Build GRCh38
rs10407734 Chr.19: 57472677-57472677 on Build GRCh38
rs118080625 Chr.19: 57472668-57472668 on Build GRCh38
rs558461811 Chr.19: 57472679-57472679 on Build GRCh38
rs370882367 Chr.19: 57472755-57472755 on Build GRCh38
rs12977381 Chr.19: 57472822-57472822 on Build GRCh38
rs12977532 Chr.19: 57472669-57472669 on Build GRCh38
rs757522761 Chr.19: 57472665-57472665 on Build GRCh38
rs897591729 Chr.19: 57472586-57472586 on Build GRCh38
rs576282864 Chr.19: 57472736-57472736 on Build GRCh38
rs956323517 Chr.19: 57472698-57472698 on Build GRCh38
rs754044898 Chr.19: 57472654-57472654 on Build GRCh38
rs148162742 Chr.19: 57472924-57472924 on Build GRCh38
rs764511070 Chr.19: 57472616-57472616 on Build GRCh38
rs10406026 Chr.19: 57472728-57472728 on Build GRCh38

View Full Product Details