Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26193485-26193982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
XM_011532567.2 NM_000182.4

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
UniGene
Hs.516032
Gene Chromosome Location
Chr.2: 26190635-26244726 on Build GRCh38
Transcripts
XM_011532567.2 NM_000182.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668766 c.1863G>T p.Q621H Chr.2: 26193599-26193599 on Build GRCh38
1494833 c.1795G>A p.V599M Chr.2: 26193667-26193667 on Build GRCh38
4389720 c.1787C>G p.A596G Chr.2: 26193675-26193675 on Build GRCh38
6677468 c.1800G>A p.A600A Chr.2: 26193662-26193662 on Build GRCh38
6091086 c.1799C>G p.A600G Chr.2: 26193663-26193663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749747433 Chr.2: 26193766-26193766 on Build GRCh38
rs61731155 Chr.2: 26193619-26193619 on Build GRCh38
rs747985669 Chr.2: 26193651-26193651 on Build GRCh38
rs763455069 Chr.2: 26193540-26193540 on Build GRCh38
rs142401180 Chr.2: 26193786-26193786 on Build GRCh38
rs1025776132 Chr.2: 26193825-26193825 on Build GRCh38
rs979861852 Chr.2: 26193791-26193791 on Build GRCh38
rs908843285 Chr.2: 26193697-26193697 on Build GRCh38
rs758983774 Chr.2: 26193825-26193828 on Build GRCh38
rs543883463 Chr.2: 26193758-26193758 on Build GRCh38
rs755208956 Chr.2: 26193718-26193718 on Build GRCh38
rs764942632 Chr.2: 26193714-26193714 on Build GRCh38
rs935397648 Chr.2: 26193759-26193759 on Build GRCh38
rs773865390 Chr.2: 26193823-26193824 on Build GRCh38
rs893811806 Chr.2: 26193775-26193775 on Build GRCh38
rs768093865 Chr.2: 26193812-26193812 on Build GRCh38
rs184904266 Chr.2: 26193911-26193911 on Build GRCh38
rs753722938 Chr.2: 26193709-26193709 on Build GRCh38
rs764067243 Chr.2: 26193565-26193565 on Build GRCh38
rs761728649 Chr.2: 26193823-26193823 on Build GRCh38
rs1012467825 Chr.2: 26193507-26193507 on Build GRCh38
rs371137138 Chr.2: 26193546-26193546 on Build GRCh38
rs1057516217 Chr.2: 26193774-26193774 on Build GRCh38
rs766881788 Chr.2: 26193544-26193544 on Build GRCh38
rs188167993 Chr.2: 26193938-26193938 on Build GRCh38
rs776121455 Chr.2: 26193820-26193820 on Build GRCh38
rs756064417 Chr.2: 26193563-26193563 on Build GRCh38
rs757001811 Chr.2: 26193595-26193595 on Build GRCh38
rs13432831 Chr.2: 26193529-26193529 on Build GRCh38
rs11552521 Chr.2: 26193696-26193696 on Build GRCh38
rs768566396 Chr.2: 26193884-26193884 on Build GRCh38
rs951632662 Chr.2: 26193905-26193905 on Build GRCh38
rs144391866 Chr.2: 26193615-26193615 on Build GRCh38
rs923784779 Chr.2: 26193744-26193744 on Build GRCh38
rs762714379 Chr.2: 26193807-26193807 on Build GRCh38
rs963053598 Chr.2: 26193700-26193700 on Build GRCh38
rs561523435 Chr.2: 26193679-26193679 on Build GRCh38
rs769580842 Chr.2: 26193668-26193669 on Build GRCh38
rs943412805 Chr.2: 26193509-26193509 on Build GRCh38
rs760394458 Chr.2: 26193793-26193793 on Build GRCh38
rs771364967 Chr.2: 26193662-26193662 on Build GRCh38
rs111662358 Chr.2: 26193778-26193778 on Build GRCh38
rs763015114 Chr.2: 26193800-26193801 on Build GRCh38
rs373966336 Chr.2: 26193633-26193633 on Build GRCh38
rs145512126 Chr.2: 26193550-26193550 on Build GRCh38
rs369641464 Chr.2: 26193784-26193784 on Build GRCh38
rs759125902 Chr.2: 26193551-26193551 on Build GRCh38
rs943465416 Chr.2: 26193889-26193889 on Build GRCh38
rs200602844 Chr.2: 26193738-26193738 on Build GRCh38
rs267599303 Chr.2: 26193634-26193634 on Build GRCh38
rs778014305 Chr.2: 26193761-26193761 on Build GRCh38
rs35630932 Chr.2: 26193744-26193745 on Build GRCh38
rs974621601 Chr.2: 26193704-26193704 on Build GRCh38
rs190409957 Chr.2: 26193674-26193674 on Build GRCh38
rs774863362 Chr.2: 26193663-26193663 on Build GRCh38
rs368455378 Chr.2: 26193673-26193673 on Build GRCh38
rs368557552 Chr.2: 26193785-26193785 on Build GRCh38
rs1006313329 Chr.2: 26193629-26193629 on Build GRCh38
rs752999034 Chr.2: 26193732-26193732 on Build GRCh38
rs779406286 Chr.2: 26193783-26193783 on Build GRCh38
rs752213744 Chr.2: 26193558-26193558 on Build GRCh38
rs149021400 Chr.2: 26193668-26193668 on Build GRCh38
rs35625271 Chr.2: 26193875-26193876 on Build GRCh38
rs753595181 Chr.2: 26193589-26193589 on Build GRCh38
rs756213834 Chr.2: 26193753-26193753 on Build GRCh38
rs781313065 Chr.2: 26193727-26193727 on Build GRCh38
rs772853038 Chr.2: 26193795-26193795 on Build GRCh38
rs764872153 Chr.2: 26193691-26193691 on Build GRCh38
rs773916672 Chr.2: 26193931-26193931 on Build GRCh38
rs1051580353 Chr.2: 26193733-26193733 on Build GRCh38
rs761749332 Chr.2: 26193710-26193710 on Build GRCh38
rs921898736 Chr.2: 26193707-26193707 on Build GRCh38
rs138584909 Chr.2: 26193685-26193685 on Build GRCh38
rs1004585840 Chr.2: 26193928-26193928 on Build GRCh38
rs962999828 Chr.2: 26193952-26193952 on Build GRCh38
rs761330765 Chr.2: 26193954-26193954 on Build GRCh38
rs755224363 Chr.2: 26193613-26193613 on Build GRCh38
rs778699495 Chr.2: 26193596-26193596 on Build GRCh38
rs201393639 Chr.2: 26193608-26193608 on Build GRCh38
rs1040477289 Chr.2: 26193899-26193899 on Build GRCh38
rs766984282 Chr.2: 26193552-26193552 on Build GRCh38
rs546433399 Chr.2: 26193555-26193556 on Build GRCh38

View Full Product Details