Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134953090-134953617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT2
Entrez Gene ID
905
Gene Name
cyclin T2
Gene Aliases
CYCT2
UniGene
Hs.744115
Gene Chromosome Location
Chr.2: 134918810-134959345 on Build GRCh38
Transcripts
NM_001241.3 NM_001320748.1 NM_001320749.1 NM_058241.2 NR_037649.1 NR_135468.1 XM_017005227.1 XM_017005228.1 XM_017005229.1 XM_017005230.1 XM_017005231.1 XM_017005232.1 XM_017005233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1006701 c.962C>A p.S321Y Chr.2: 134953417-134953417 on Build GRCh38
1399479 c.939A>G p.V313V Chr.2: 134953394-134953394 on Build GRCh38
1006703 c.1044C>T p.H348H Chr.2: 134953499-134953499 on Build GRCh38
3115974 c.913T>C p.S305P Chr.2: 134953368-134953368 on Build GRCh38
1006699 c.846G>T p.L282F Chr.2: 134953301-134953301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772559544 Chr.2: 134953258-134953258 on Build GRCh38
rs114529129 Chr.2: 134953204-134953204 on Build GRCh38
rs776568653 Chr.2: 134953546-134953546 on Build GRCh38
rs764081676 Chr.2: 134953451-134953451 on Build GRCh38
rs923278971 Chr.2: 134953328-134953328 on Build GRCh38
rs767422530 Chr.2: 134953201-134953201 on Build GRCh38
rs772869230 Chr.2: 134953215-134953216 on Build GRCh38
rs760465431 Chr.2: 134953534-134953534 on Build GRCh38
rs557451615 Chr.2: 134953335-134953335 on Build GRCh38
rs771733534 Chr.2: 134953434-134953434 on Build GRCh38
rs747491085 Chr.2: 134953384-134953384 on Build GRCh38
rs775871224 Chr.2: 134953281-134953281 on Build GRCh38
rs777364426 Chr.2: 134953207-134953207 on Build GRCh38
rs768892959 Chr.2: 134953283-134953283 on Build GRCh38
rs906889312 Chr.2: 134953156-134953156 on Build GRCh38
rs776893383 Chr.2: 134953295-134953295 on Build GRCh38
rs368278085 Chr.2: 134953331-134953331 on Build GRCh38
rs749449311 Chr.2: 134953505-134953505 on Build GRCh38
rs778010477 Chr.2: 134953306-134953306 on Build GRCh38
rs751362401 Chr.2: 134953194-134953194 on Build GRCh38
rs993754193 Chr.2: 134953168-134953168 on Build GRCh38
rs748906350 Chr.2: 134953220-134953220 on Build GRCh38
rs764625286 Chr.2: 134953573-134953573 on Build GRCh38
rs748181684 Chr.2: 134953486-134953486 on Build GRCh38
rs766205042 Chr.2: 134953590-134953590 on Build GRCh38
rs74513762 Chr.2: 134953422-134953422 on Build GRCh38
rs371316982 Chr.2: 134953497-134953497 on Build GRCh38
rs758870300 Chr.2: 134953251-134953251 on Build GRCh38
rs568369205 Chr.2: 134953134-134953134 on Build GRCh38
rs746282404 Chr.2: 134953249-134953249 on Build GRCh38
rs201915230 Chr.2: 134953196-134953196 on Build GRCh38
rs762108373 Chr.2: 134953299-134953299 on Build GRCh38
rs761343075 Chr.2: 134953567-134953567 on Build GRCh38
rs570939685 Chr.2: 134953586-134953586 on Build GRCh38
rs745511547 Chr.2: 134953588-134953588 on Build GRCh38
rs984032949 Chr.2: 134953261-134953261 on Build GRCh38
rs139813514 Chr.2: 134953485-134953485 on Build GRCh38
rs748306468 Chr.2: 134953395-134953395 on Build GRCh38
rs777019755 Chr.2: 134953180-134953180 on Build GRCh38
rs756895419 Chr.2: 134953349-134953349 on Build GRCh38
rs759328044 Chr.2: 134953418-134953418 on Build GRCh38
rs199605477 Chr.2: 134953185-134953185 on Build GRCh38
rs774193353 Chr.2: 134953307-134953307 on Build GRCh38
rs769338607 Chr.2: 134953387-134953387 on Build GRCh38
rs779063877 Chr.2: 134953224-134953231 on Build GRCh38
rs34015791 Chr.2: 134953241-134953241 on Build GRCh38
rs922720997 Chr.2: 134953545-134953545 on Build GRCh38
rs772831558 Chr.2: 134953576-134953576 on Build GRCh38
rs1024241311 Chr.2: 134953126-134953126 on Build GRCh38
rs767432151 Chr.2: 134953314-134953314 on Build GRCh38
rs758709203 Chr.2: 134953368-134953368 on Build GRCh38
rs750077587 Chr.2: 134953466-134953466 on Build GRCh38
rs747041013 Chr.2: 134953282-134953282 on Build GRCh38
rs780875456 Chr.2: 134953558-134953558 on Build GRCh38
rs377180297 Chr.2: 134953452-134953452 on Build GRCh38
rs977342136 Chr.2: 134953357-134953357 on Build GRCh38
rs775260481 Chr.2: 134953532-134953532 on Build GRCh38
rs769786913 Chr.2: 134953489-134953489 on Build GRCh38
rs764891019 Chr.2: 134953462-134953462 on Build GRCh38
rs142981962 Chr.2: 134953346-134953346 on Build GRCh38
rs572470846 Chr.2: 134953374-134953374 on Build GRCh38
rs766216424 Chr.2: 134953287-134953287 on Build GRCh38
rs370280587 Chr.2: 134953247-134953247 on Build GRCh38
rs758113255 Chr.2: 134953473-134953473 on Build GRCh38
rs763570996 Chr.2: 134953345-134953345 on Build GRCh38
rs775310444 Chr.2: 134953436-134953436 on Build GRCh38
rs79101220 Chr.2: 134953413-134953413 on Build GRCh38
rs865829499 Chr.2: 134953233-134953233 on Build GRCh38
rs781411502 Chr.2: 134953388-134953388 on Build GRCh38
rs969553136 Chr.2: 134953163-134953163 on Build GRCh38
rs145398488 Chr.2: 134953160-134953160 on Build GRCh38
rs911061111 Chr.2: 134953270-134953270 on Build GRCh38
rs146692051 Chr.2: 134953526-134953526 on Build GRCh38
rs761239145 Chr.2: 134953461-134953461 on Build GRCh38
rs759255002 Chr.2: 134953310-134953310 on Build GRCh38
rs750737788 Chr.2: 134953366-134953366 on Build GRCh38
rs765437605 Chr.2: 134953301-134953301 on Build GRCh38
rs768519724 Chr.2: 134953544-134953544 on Build GRCh38
rs755642476 Chr.2: 134953205-134953205 on Build GRCh38
rs1052458897 Chr.2: 134953583-134953583 on Build GRCh38
rs780261527 Chr.2: 134953483-134953483 on Build GRCh38
rs780574413 Chr.2: 134953383-134953383 on Build GRCh38
rs948434653 Chr.2: 134953559-134953559 on Build GRCh38
rs760592121 Chr.2: 134953439-134953439 on Build GRCh38
rs769994576 Chr.2: 134953411-134953411 on Build GRCh38
rs935396354 Chr.2: 134953565-134953565 on Build GRCh38
rs368857873 Chr.2: 134953484-134953484 on Build GRCh38

View Full Product Details