Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 213004961-213005503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
UniGene
Hs.604950
Gene Chromosome Location
Chr.2: 212999684-213152753 on Build GRCh38
Transcripts
NM_001079526.1 NM_016260.2 XM_005246384.4 XM_005246385.3 XM_005246386.4 XM_011510802.2 XM_011510803.2 XM_011510804.2 XM_011510805.2 XM_011510807.2 XM_011510808.2 XM_011510809.2 XM_011510810.2 XM_011510811.2 XM_011510812.2 XM_011510814.2 XM_011510815.2 XM_011510816.2 XM_011510817.2 XM_011510818.2 XM_011510819.2 XM_017003589.1 XM_017003590.1 XM_017003591.1 XM_017003592.1

SNP Detail

SNP ID Chromosome Location
rs982371193 Chr.2: 213005011-213005011 on Build GRCh38
rs865874438 Chr.2: 213005201-213005201 on Build GRCh38
rs916771345 Chr.2: 213005002-213005002 on Build GRCh38
rs954794944 Chr.2: 213004999-213004999 on Build GRCh38
rs889617077 Chr.2: 213005086-213005086 on Build GRCh38
rs757172122 Chr.2: 213005377-213005377 on Build GRCh38
rs71681011 Chr.2: 213005192-213005193 on Build GRCh38
rs970901655 Chr.2: 213005278-213005278 on Build GRCh38
rs899917344 Chr.2: 213005192-213005192 on Build GRCh38
rs1047012063 Chr.2: 213005069-213005072 on Build GRCh38
rs749923394 Chr.2: 213005004-213005004 on Build GRCh38
rs192107936 Chr.2: 213005043-213005043 on Build GRCh38
rs755700175 Chr.2: 213005054-213005054 on Build GRCh38
rs533158357 Chr.2: 213005383-213005383 on Build GRCh38
rs113896119 Chr.2: 213005195-213005195 on Build GRCh38
rs908091714 Chr.2: 213005299-213005299 on Build GRCh38
rs202244129 Chr.2: 213005268-213005269 on Build GRCh38
rs74811505 Chr.2: 213005189-213005190 on Build GRCh38
rs947792750 Chr.2: 213005066-213005066 on Build GRCh38
rs939660370 Chr.2: 213005021-213005021 on Build GRCh38
rs908143211 Chr.2: 213005019-213005019 on Build GRCh38
rs34696012 Chr.2: 213005321-213005322 on Build GRCh38
rs1040117951 Chr.2: 213005472-213005472 on Build GRCh38
rs981862729 Chr.2: 213005407-213005407 on Build GRCh38
rs977882544 Chr.2: 213005196-213005196 on Build GRCh38
rs752281851 Chr.2: 213005201-213005202 on Build GRCh38
rs1036708545 Chr.2: 213005030-213005030 on Build GRCh38
rs1047252301 Chr.2: 213005461-213005461 on Build GRCh38
rs71744347 Chr.2: 213005193-213005194 on Build GRCh38
rs940926957 Chr.2: 213005458-213005458 on Build GRCh38
rs1030591850 Chr.2: 213005193-213005193 on Build GRCh38
rs975020683 Chr.2: 213004992-213004993 on Build GRCh38
rs982726437 Chr.2: 213005297-213005297 on Build GRCh38
rs144911749 Chr.2: 213005150-213005150 on Build GRCh38
rs200916216 Chr.2: 213005267-213005268 on Build GRCh38
rs957946872 Chr.2: 213005198-213005198 on Build GRCh38
rs972368247 Chr.2: 213005462-213005462 on Build GRCh38
rs947811091 Chr.2: 213005281-213005281 on Build GRCh38
rs768788671 Chr.2: 213005197-213005198 on Build GRCh38
rs927683840 Chr.2: 213005414-213005414 on Build GRCh38
rs58270171 Chr.2: 213005200-213005201 on Build GRCh38
rs1032305023 Chr.2: 213005185-213005185 on Build GRCh38
rs1047365341 Chr.2: 213005149-213005150 on Build GRCh38
rs372338169 Chr.2: 213005191-213005191 on Build GRCh38
rs1049999306 Chr.2: 213005076-213005076 on Build GRCh38
rs1024167700 Chr.2: 213005253-213005253 on Build GRCh38
rs200090377 Chr.2: 213005198-213005199 on Build GRCh38
rs1019526382 Chr.2: 213005120-213005120 on Build GRCh38
rs151217930 Chr.2: 213005252-213005252 on Build GRCh38
rs770693428 Chr.2: 213005125-213005125 on Build GRCh38
rs999446525 Chr.2: 213005160-213005160 on Build GRCh38
rs1008502978 Chr.2: 213005101-213005101 on Build GRCh38
rs991354606 Chr.2: 213004993-213004993 on Build GRCh38
rs200427008 Chr.2: 213005269-213005270 on Build GRCh38
rs930237658 Chr.2: 213005143-213005143 on Build GRCh38
rs958089515 Chr.2: 213005189-213005189 on Build GRCh38
rs930155091 Chr.2: 213005415-213005415 on Build GRCh38
rs774979216 Chr.2: 213005191-213005192 on Build GRCh38
rs931068699 Chr.2: 213005058-213005058 on Build GRCh38
rs535182377 Chr.2: 213005468-213005468 on Build GRCh38

View Full Product Details