Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58992876-58993376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFCD
Entrez Gene ID
51497
Gene Name
negative elongation factor complex member C/D
Gene Aliases
HSPC130, NELF-C, NELF-D, TH1, TH1L
UniGene
Hs.517148
Gene Chromosome Location
Chr.20: 58981208-58995133 on Build GRCh38
Transcripts
NM_198976.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6467769 c.1355T>G p.L452R Chr.20: 58993096-58993096 on Build GRCh38
6258799 c.1371+1G>T p.? Chr.20: 58993113-58993113 on Build GRCh38
224058 c.1277G>A p.G426D Chr.20: 58993018-58993018 on Build GRCh38
4099821 c.1261C>G p.P421A Chr.20: 58993002-58993002 on Build GRCh38
1229120 c.1331A>C p.Q444P Chr.20: 58993072-58993072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754255816 Chr.20: 58993094-58993094 on Build GRCh38
rs1009731867 Chr.20: 58993271-58993271 on Build GRCh38
rs762028433 Chr.20: 58993095-58993095 on Build GRCh38
rs770638503 Chr.20: 58993004-58993004 on Build GRCh38
rs773815606 Chr.20: 58993014-58993014 on Build GRCh38
rs1001844087 Chr.20: 58993204-58993204 on Build GRCh38
rs545237353 Chr.20: 58993123-58993123 on Build GRCh38
rs368112048 Chr.20: 58993158-58993158 on Build GRCh38
rs1032517460 Chr.20: 58993224-58993224 on Build GRCh38
rs1005277718 Chr.20: 58992952-58992952 on Build GRCh38
rs538131830 Chr.20: 58993090-58993090 on Build GRCh38
rs776421925 Chr.20: 58992955-58992955 on Build GRCh38
rs767033397 Chr.20: 58993168-58993168 on Build GRCh38
rs6070686 Chr.20: 58993331-58993331 on Build GRCh38
rs368381606 Chr.20: 58993046-58993046 on Build GRCh38
rs752563181 Chr.20: 58993007-58993007 on Build GRCh38
rs374995793 Chr.20: 58992961-58992961 on Build GRCh38
rs7348127 Chr.20: 58993122-58993122 on Build GRCh38
rs907301245 Chr.20: 58993019-58993019 on Build GRCh38
rs758561018 Chr.20: 58993115-58993115 on Build GRCh38
rs759154065 Chr.20: 58993024-58993024 on Build GRCh38
rs912192974 Chr.20: 58993169-58993169 on Build GRCh38
rs141996345 Chr.20: 58993100-58993100 on Build GRCh38
rs560747085 Chr.20: 58993292-58993292 on Build GRCh38
rs975151329 Chr.20: 58993212-58993212 on Build GRCh38
rs6092717 Chr.20: 58993140-58993140 on Build GRCh38
rs367761562 Chr.20: 58992927-58992928 on Build GRCh38
rs756152151 Chr.20: 58993138-58993138 on Build GRCh38
rs111726128 Chr.20: 58993334-58993334 on Build GRCh38
rs200807638 Chr.20: 58993067-58993067 on Build GRCh38
rs112186327 Chr.20: 58993113-58993113 on Build GRCh38
rs6070687 Chr.20: 58993338-58993338 on Build GRCh38
rs759257423 Chr.20: 58992974-58992975 on Build GRCh38
rs941834040 Chr.20: 58993053-58993053 on Build GRCh38
rs1047418996 Chr.20: 58992902-58992902 on Build GRCh38
rs771559225 Chr.20: 58993025-58993025 on Build GRCh38
rs765221552 Chr.20: 58992994-58992994 on Build GRCh38
rs777803092 Chr.20: 58993155-58993155 on Build GRCh38
rs189355 Chr.20: 58993179-58993179 on Build GRCh38
rs761140078 Chr.20: 58993082-58993082 on Build GRCh38
rs193192 Chr.20: 58993093-58993093 on Build GRCh38
rs894071388 Chr.20: 58993238-58993238 on Build GRCh38
rs772548407 Chr.20: 58993302-58993302 on Build GRCh38
rs1034515524 Chr.20: 58993088-58993088 on Build GRCh38
rs762571207 Chr.20: 58993110-58993116 on Build GRCh38
rs6092716 Chr.20: 58992927-58992927 on Build GRCh38
rs747576059 Chr.20: 58993271-58993274 on Build GRCh38
rs189567482 Chr.20: 58993058-58993058 on Build GRCh38
rs886463827 Chr.20: 58992903-58992903 on Build GRCh38
rs762674747 Chr.20: 58992979-58992979 on Build GRCh38
rs1024278535 Chr.20: 58992958-58992958 on Build GRCh38
rs371514895 Chr.20: 58993161-58993161 on Build GRCh38
rs371084351 Chr.20: 58993099-58993099 on Build GRCh38
rs986015289 Chr.20: 58992945-58992945 on Build GRCh38
rs963796969 Chr.20: 58993185-58993185 on Build GRCh38

View Full Product Details