Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30546507-30547031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L6
Entrez Gene ID
730005
Gene Name
SEC14 like lipid binding 6
Gene Aliases
-
UniGene
Hs.505605
Gene Chromosome Location
Chr.22: 30522797-30546717 on Build GRCh38
Transcripts
NM_001193336.2 XM_017028931.1 XM_017028933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6468111 c.54+1G>A p.? Chr.22: 30546628-30546628 on Build GRCh38
5663769 c.13G>T p.V5L Chr.22: 30546670-30546670 on Build GRCh38
7193792 c.14T>C p.V5A Chr.22: 30546669-30546669 on Build GRCh38
3693907 c.45G>A p.S15S Chr.22: 30546638-30546638 on Build GRCh38
5707629 c.49G>A p.A17T Chr.22: 30546634-30546634 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs900009180 Chr.22: 30546713-30546714 on Build GRCh38
rs770742615 Chr.22: 30546870-30546870 on Build GRCh38
rs942164343 Chr.22: 30546720-30546720 on Build GRCh38
rs751528720 Chr.22: 30546626-30546626 on Build GRCh38
rs894426637 Chr.22: 30546768-30546768 on Build GRCh38
rs185816926 Chr.22: 30546837-30546837 on Build GRCh38
rs962656648 Chr.22: 30546599-30546599 on Build GRCh38
rs998622975 Chr.22: 30546788-30546788 on Build GRCh38
rs755461080 Chr.22: 30546963-30546963 on Build GRCh38
rs113555233 Chr.22: 30546550-30546550 on Build GRCh38
rs563075588 Chr.22: 30546703-30546703 on Build GRCh38
rs868708736 Chr.22: 30546533-30546533 on Build GRCh38
rs974300926 Chr.22: 30546629-30546629 on Build GRCh38
rs148882530 Chr.22: 30546758-30546758 on Build GRCh38
rs986376928 Chr.22: 30546849-30546849 on Build GRCh38
rs1042921043 Chr.22: 30546934-30546934 on Build GRCh38
rs1031809516 Chr.22: 30546796-30546796 on Build GRCh38
rs914255628 Chr.22: 30546907-30546907 on Build GRCh38
rs529705414 Chr.22: 30546622-30546622 on Build GRCh38
rs915770677 Chr.22: 30546631-30546631 on Build GRCh38
rs573400448 Chr.22: 30546978-30546978 on Build GRCh38
rs5997679 Chr.22: 30546711-30546711 on Build GRCh38
rs992823416 Chr.22: 30546915-30546915 on Build GRCh38
rs948642201 Chr.22: 30546919-30546919 on Build GRCh38
rs972193866 Chr.22: 30546873-30546873 on Build GRCh38
rs542508696 Chr.22: 30546982-30546982 on Build GRCh38
rs76415233 Chr.22: 30546552-30546552 on Build GRCh38
rs751135423 Chr.22: 30546760-30546760 on Build GRCh38
rs150121401 Chr.22: 30546696-30546696 on Build GRCh38
rs758272261 Chr.22: 30546604-30546604 on Build GRCh38
rs988415947 Chr.22: 30546885-30546885 on Build GRCh38
rs946991017 Chr.22: 30546708-30546708 on Build GRCh38
rs938768131 Chr.22: 30546908-30546908 on Build GRCh38
rs1036813556 Chr.22: 30546728-30546728 on Build GRCh38
rs568437548 Chr.22: 30546803-30546803 on Build GRCh38
rs557457201 Chr.22: 30546836-30546836 on Build GRCh38
rs750664669 Chr.22: 30546705-30546706 on Build GRCh38
rs900854837 Chr.22: 30546814-30546814 on Build GRCh38
rs982658565 Chr.22: 30546577-30546577 on Build GRCh38
rs1042909667 Chr.22: 30546780-30546780 on Build GRCh38
rs758034059 Chr.22: 30546726-30546726 on Build GRCh38
rs923492605 Chr.22: 30546686-30546686 on Build GRCh38
rs112701289 Chr.22: 30546551-30546551 on Build GRCh38
rs553543783 Chr.22: 30546858-30546858 on Build GRCh38
rs1052753276 Chr.22: 30546689-30546689 on Build GRCh38
rs904316528 Chr.22: 30546783-30546783 on Build GRCh38
rs780227887 Chr.22: 30546932-30546932 on Build GRCh38
rs780656771 Chr.22: 30546651-30546651 on Build GRCh38
rs386820637 Chr.22: 30546550-30546552 on Build GRCh38
rs5997680 Chr.22: 30546901-30546901 on Build GRCh38
rs149457357 Chr.22: 30546542-30546542 on Build GRCh38
rs182764828 Chr.22: 30546555-30546555 on Build GRCh38
rs749660936 Chr.22: 30546956-30546956 on Build GRCh38
rs537478335 Chr.22: 30546824-30546824 on Build GRCh38
rs745421224 Chr.22: 30546694-30546694 on Build GRCh38
rs112506192 Chr.22: 30546774-30546774 on Build GRCh38
rs997925500 Chr.22: 30546979-30546979 on Build GRCh38
rs1015519262 Chr.22: 30546583-30546585 on Build GRCh38
rs9608979 Chr.22: 30546714-30546714 on Build GRCh38
rs981229020 Chr.22: 30546669-30546669 on Build GRCh38
rs938659566 Chr.22: 30546759-30546759 on Build GRCh38
rs955367950 Chr.22: 30546565-30546568 on Build GRCh38
rs900946795 Chr.22: 30546970-30546970 on Build GRCh38
rs756705918 Chr.22: 30546638-30546638 on Build GRCh38
rs1004211011 Chr.22: 30546749-30546749 on Build GRCh38
rs915909503 Chr.22: 30546918-30546918 on Build GRCh38
rs976374679 Chr.22: 30546639-30546639 on Build GRCh38
rs919362885 Chr.22: 30546883-30546883 on Build GRCh38
rs539993343 Chr.22: 30546848-30546848 on Build GRCh38
rs928248834 Chr.22: 30546676-30546676 on Build GRCh38
rs565785957 Chr.22: 30546746-30546746 on Build GRCh38
rs904368519 Chr.22: 30546944-30546944 on Build GRCh38
rs752431968 Chr.22: 30546588-30546588 on Build GRCh38
rs932834289 Chr.22: 30546937-30546937 on Build GRCh38
rs953642646 Chr.22: 30546841-30546841 on Build GRCh38

View Full Product Details