Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14111940-14112444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD4
Entrez Gene ID
131474
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 4
Gene Aliases
MIA40, TIMM40
UniGene
Hs.517815
Gene Chromosome Location
Chr.3: 14112077-14124871 on Build GRCh38
Transcripts
NM_001098502.1 NM_144636.2

SNP Detail

SNP ID Chromosome Location
rs550991970 Chr.3: 14112392-14112392 on Build GRCh38
rs1133118 Chr.3: 14112277-14112277 on Build GRCh38
rs543362188 Chr.3: 14111967-14111967 on Build GRCh38
rs886227611 Chr.3: 14112124-14112124 on Build GRCh38
rs964621091 Chr.3: 14111993-14111993 on Build GRCh38
rs978295268 Chr.3: 14112335-14112335 on Build GRCh38
rs148875919 Chr.3: 14112318-14112326 on Build GRCh38
rs748126400 Chr.3: 14112383-14112383 on Build GRCh38
rs1021946174 Chr.3: 14112246-14112246 on Build GRCh38
rs571547196 Chr.3: 14112341-14112341 on Build GRCh38
rs945136703 Chr.3: 14112075-14112075 on Build GRCh38
rs528903061 Chr.3: 14111979-14111979 on Build GRCh38
rs987385379 Chr.3: 14111978-14111978 on Build GRCh38
rs186779064 Chr.3: 14111976-14111976 on Build GRCh38
rs955652020 Chr.3: 14112359-14112359 on Build GRCh38
rs768544579 Chr.3: 14112275-14112275 on Build GRCh38
rs779819139 Chr.3: 14112179-14112179 on Build GRCh38
rs73140595 Chr.3: 14112033-14112033 on Build GRCh38
rs549028507 Chr.3: 14112001-14112001 on Build GRCh38
rs1003284531 Chr.3: 14112131-14112131 on Build GRCh38
rs894770523 Chr.3: 14112234-14112234 on Build GRCh38
rs3814412 Chr.3: 14112344-14112344 on Build GRCh38
rs751939809 Chr.3: 14111982-14111985 on Build GRCh38
rs968271345 Chr.3: 14112331-14112331 on Build GRCh38
rs551555872 Chr.3: 14112231-14112231 on Build GRCh38
rs1017474595 Chr.3: 14111987-14111987 on Build GRCh38
rs548367077 Chr.3: 14112213-14112213 on Build GRCh38
rs915049266 Chr.3: 14112069-14112069 on Build GRCh38
rs528199764 Chr.3: 14112185-14112185 on Build GRCh38

View Full Product Details