Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110605312-110605819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA2
Entrez Gene ID
3737
Gene Name
potassium voltage-gated channel subfamily A member 2
Gene Aliases
EIEE32, HBK5, HK4, HUKIV, KV1.2, MK2, NGK1, RBK2
UniGene
Hs.248139
Gene Chromosome Location
Chr.1: 110593580-110631536 on Build GRCh38
Transcripts
NM_001204269.1 NM_004974.3 XM_011541396.2 XM_011541397.2 XM_011541398.2 XM_011541399.2 XM_011541400.2 XM_017001213.1

SNP Detail

SNP ID Chromosome Location
rs538479001 Chr.1: 110605474-110605474 on Build GRCh38
rs983530990 Chr.1: 110605475-110605475 on Build GRCh38
rs865950565 Chr.1: 110605411-110605411 on Build GRCh38
rs556602844 Chr.1: 110605421-110605421 on Build GRCh38
rs372237953 Chr.1: 110605393-110605393 on Build GRCh38
rs933067015 Chr.1: 110605693-110605693 on Build GRCh38
rs948235557 Chr.1: 110605564-110605564 on Build GRCh38
rs769040141 Chr.1: 110605637-110605637 on Build GRCh38
rs1006475290 Chr.1: 110605721-110605721 on Build GRCh38
rs964653792 Chr.1: 110605753-110605753 on Build GRCh38
rs888927103 Chr.1: 110605718-110605718 on Build GRCh38
rs369570792 Chr.1: 110605496-110605496 on Build GRCh38
rs1050619219 Chr.1: 110605717-110605717 on Build GRCh38
rs966591886 Chr.1: 110605379-110605379 on Build GRCh38
rs555095737 Chr.1: 110605764-110605764 on Build GRCh38
rs77148768 Chr.1: 110605794-110605794 on Build GRCh38
rs538932653 Chr.1: 110605445-110605445 on Build GRCh38
rs746368068 Chr.1: 110605436-110605436 on Build GRCh38
rs916732658 Chr.1: 110605495-110605495 on Build GRCh38
rs979734392 Chr.1: 110605398-110605398 on Build GRCh38
rs896182685 Chr.1: 110605638-110605638 on Build GRCh38
rs908033639 Chr.1: 110605484-110605484 on Build GRCh38
rs925543719 Chr.1: 110605587-110605587 on Build GRCh38
rs1016607293 Chr.1: 110605382-110605382 on Build GRCh38
rs542452769 Chr.1: 110605585-110605585 on Build GRCh38
rs1052044545 Chr.1: 110605629-110605629 on Build GRCh38
rs755271769 Chr.1: 110605680-110605680 on Build GRCh38
rs747232096 Chr.1: 110605586-110605586 on Build GRCh38
rs1016137356 Chr.1: 110605747-110605747 on Build GRCh38
rs3887820 Chr.1: 110605458-110605458 on Build GRCh38
rs188801376 Chr.1: 110605461-110605461 on Build GRCh38
rs1000711542 Chr.1: 110605744-110605744 on Build GRCh38
rs1054787749 Chr.1: 110605628-110605628 on Build GRCh38
rs560563440 Chr.1: 110605612-110605612 on Build GRCh38
rs935626511 Chr.1: 110605618-110605618 on Build GRCh38
rs373015164 Chr.1: 110605606-110605606 on Build GRCh38
rs972727180 Chr.1: 110605426-110605426 on Build GRCh38
rs773135682 Chr.1: 110605702-110605702 on Build GRCh38

View Full Product Details