Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127795688-127796202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA4L
Entrez Gene ID
22824
Gene Name
heat shock protein family A (Hsp70) member 4 like
Gene Aliases
APG-1, HSPH3, Osp94
UniGene
Hs.135554
Gene Chromosome Location
Chr.4: 127781821-127834073 on Build GRCh38
Transcripts
NM_001317381.1 NM_001317382.1 NM_001317383.1 NM_014278.3 XM_011531745.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938612 c.259A>G p.I87V Chr.4: 127795768-127795768 on Build GRCh38
3600178 c.229G>A p.D77N Chr.4: 127795831-127795831 on Build GRCh38
4880735 c.194A>T p.H65L Chr.4: 127795796-127795796 on Build GRCh38
3600180 c.285T>G p.P95P Chr.4: 127795887-127795887 on Build GRCh38
5532449 c.355C>T p.P119S Chr.4: 127795864-127795864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770770339 Chr.4: 127795805-127795805 on Build GRCh38
rs760675719 Chr.4: 127795834-127795834 on Build GRCh38
rs974437426 Chr.4: 127796123-127796123 on Build GRCh38
rs765686750 Chr.4: 127795754-127795754 on Build GRCh38
rs947572773 Chr.4: 127795890-127795890 on Build GRCh38
rs944036112 Chr.4: 127795962-127795962 on Build GRCh38
rs953201670 Chr.4: 127796144-127796144 on Build GRCh38
rs552203306 Chr.4: 127795991-127795991 on Build GRCh38
rs915022818 Chr.4: 127796124-127796124 on Build GRCh38
rs755630197 Chr.4: 127795883-127795883 on Build GRCh38
rs375704600 Chr.4: 127795743-127795743 on Build GRCh38
rs144848136 Chr.4: 127796101-127796101 on Build GRCh38
rs201383324 Chr.4: 127795888-127795888 on Build GRCh38
rs189118675 Chr.4: 127796023-127796023 on Build GRCh38
rs535724502 Chr.4: 127796041-127796041 on Build GRCh38
rs533738475 Chr.4: 127795975-127795975 on Build GRCh38
rs202140695 Chr.4: 127795718-127795718 on Build GRCh38
rs911638580 Chr.4: 127795792-127795792 on Build GRCh38
rs767103769 Chr.4: 127795868-127795868 on Build GRCh38
rs754330651 Chr.4: 127795744-127795744 on Build GRCh38
rs768798898 Chr.4: 127795835-127795835 on Build GRCh38
rs138445498 Chr.4: 127795865-127795865 on Build GRCh38
rs537978257 Chr.4: 127796022-127796022 on Build GRCh38
rs760761507 Chr.4: 127795740-127795740 on Build GRCh38
rs775399179 Chr.4: 127795739-127795739 on Build GRCh38
rs780801336 Chr.4: 127795776-127795776 on Build GRCh38
rs757154644 Chr.4: 127795895-127795895 on Build GRCh38
rs75776460 Chr.4: 127795779-127795779 on Build GRCh38
rs1023074520 Chr.4: 127796001-127796001 on Build GRCh38
rs200089402 Chr.4: 127795840-127795840 on Build GRCh38
rs367736477 Chr.4: 127795780-127795780 on Build GRCh38
rs752324370 Chr.4: 127795870-127795870 on Build GRCh38
rs745683811 Chr.4: 127795926-127795926 on Build GRCh38
rs967733445 Chr.4: 127796028-127796028 on Build GRCh38
rs200226261 Chr.4: 127795775-127795775 on Build GRCh38
rs867936873 Chr.4: 127795784-127795784 on Build GRCh38
rs750906753 Chr.4: 127795781-127795781 on Build GRCh38
rs567172230 Chr.4: 127795959-127795959 on Build GRCh38
rs749165073 Chr.4: 127795790-127795790 on Build GRCh38
rs369705910 Chr.4: 127795855-127795855 on Build GRCh38
rs911508869 Chr.4: 127796094-127796094 on Build GRCh38
rs762282282 Chr.4: 127795748-127795748 on Build GRCh38
rs1041297620 Chr.4: 127795976-127795976 on Build GRCh38
rs1012964487 Chr.4: 127795997-127795997 on Build GRCh38
rs547178659 Chr.4: 127795987-127795987 on Build GRCh38
rs372033195 Chr.4: 127795978-127795978 on Build GRCh38
rs193060211 Chr.4: 127795791-127795791 on Build GRCh38
rs762409065 Chr.4: 127795838-127795838 on Build GRCh38
rs113694613 Chr.4: 127795902-127795902 on Build GRCh38
rs574573840 Chr.4: 127796030-127796030 on Build GRCh38
rs943058139 Chr.4: 127796115-127796115 on Build GRCh38
rs772207913 Chr.4: 127795820-127795820 on Build GRCh38
rs13119199 Chr.4: 127796007-127796007 on Build GRCh38
rs746140197 Chr.4: 127795814-127795814 on Build GRCh38
rs778673195 Chr.4: 127795811-127795811 on Build GRCh38
rs1001199763 Chr.4: 127795999-127795999 on Build GRCh38
rs750734128 Chr.4: 127795853-127795853 on Build GRCh38
rs950727966 Chr.4: 127796032-127796032 on Build GRCh38
rs751327696 Chr.4: 127795766-127795766 on Build GRCh38
rs775742078 Chr.4: 127795823-127795823 on Build GRCh38
rs375802667 Chr.4: 127795854-127795854 on Build GRCh38
rs758942751 Chr.4: 127795837-127795837 on Build GRCh38
rs1022768589 Chr.4: 127795745-127795745 on Build GRCh38
rs572012802 Chr.4: 127796164-127796164 on Build GRCh38
rs895822750 Chr.4: 127795992-127795992 on Build GRCh38
rs41278095 Chr.4: 127796017-127796017 on Build GRCh38

View Full Product Details