Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 204623-205119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC127
Entrez Gene ID
133957
Gene Name
coiled-coil domain containing 127
Gene Aliases
-
UniGene
Hs.294145
Gene Chromosome Location
Chr.5: 204760-218182 on Build GRCh38
Transcripts
NM_145265.2

SNP Detail

SNP ID Chromosome Location
rs1006647000 Chr.5: 204722-204722 on Build GRCh38
rs997767239 Chr.5: 204755-204755 on Build GRCh38
rs372576973 Chr.5: 204928-204928 on Build GRCh38
rs984669013 Chr.5: 204667-204667 on Build GRCh38
rs1031024232 Chr.5: 204770-204770 on Build GRCh38
rs965387000 Chr.5: 204937-204937 on Build GRCh38
rs868693703 Chr.5: 205093-205093 on Build GRCh38
rs966732265 Chr.5: 204801-204801 on Build GRCh38
rs3180962 Chr.5: 204960-204960 on Build GRCh38
rs536489427 Chr.5: 204973-204973 on Build GRCh38
rs1048498860 Chr.5: 205007-205007 on Build GRCh38
rs894982901 Chr.5: 204677-204677 on Build GRCh38
rs1006618415 Chr.5: 205051-205051 on Build GRCh38
rs576157983 Chr.5: 204989-204989 on Build GRCh38
rs114552951 Chr.5: 204997-204997 on Build GRCh38
rs952753457 Chr.5: 204657-204657 on Build GRCh38
rs564904685 Chr.5: 205001-205001 on Build GRCh38
rs144626085 Chr.5: 205094-205094 on Build GRCh38
rs549771776 Chr.5: 204767-204767 on Build GRCh38
rs376714314 Chr.5: 204719-204719 on Build GRCh38
rs573309925 Chr.5: 204918-204919 on Build GRCh38
rs978531742 Chr.5: 204814-204814 on Build GRCh38
rs900653062 Chr.5: 205079-205079 on Build GRCh38
rs541317705 Chr.5: 205023-205023 on Build GRCh38
rs1036650178 Chr.5: 204698-204698 on Build GRCh38
rs753063641 Chr.5: 205068-205068 on Build GRCh38
rs940657643 Chr.5: 204693-204693 on Build GRCh38
rs189237288 Chr.5: 205008-205008 on Build GRCh38
rs949279299 Chr.5: 204689-204689 on Build GRCh38
rs765629214 Chr.5: 205066-205066 on Build GRCh38
rs999122076 Chr.5: 204922-204922 on Build GRCh38
rs909050283 Chr.5: 204983-204983 on Build GRCh38
rs938322086 Chr.5: 204737-204737 on Build GRCh38
rs1017763373 Chr.5: 204729-204729 on Build GRCh38
rs111436099 Chr.5: 204975-204975 on Build GRCh38
rs537817535 Chr.5: 204692-204692 on Build GRCh38
rs760705160 Chr.5: 204851-204851 on Build GRCh38
rs759849566 Chr.5: 205052-205052 on Build GRCh38
rs184476278 Chr.5: 204841-204841 on Build GRCh38
rs894253386 Chr.5: 204827-204827 on Build GRCh38
rs776944124 Chr.5: 204969-204969 on Build GRCh38
rs928313313 Chr.5: 204708-204708 on Build GRCh38
rs888165901 Chr.5: 204716-204716 on Build GRCh38
rs1054174 Chr.5: 204837-204837 on Build GRCh38
rs998131246 Chr.5: 205082-205082 on Build GRCh38
rs13474 Chr.5: 204962-204962 on Build GRCh38
rs74475573 Chr.5: 204871-204871 on Build GRCh38
rs996872616 Chr.5: 204948-204948 on Build GRCh38
rs921324457 Chr.5: 204651-204651 on Build GRCh38
rs903309899 Chr.5: 204916-204916 on Build GRCh38
rs994279611 Chr.5: 204696-204696 on Build GRCh38
rs554037289 Chr.5: 204899-204899 on Build GRCh38
rs764549515 Chr.5: 205098-205098 on Build GRCh38
rs55661820 Chr.5: 204650-204650 on Build GRCh38

View Full Product Details