Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150227975-150228469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2A
Entrez Gene ID
815
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Gene Aliases
CAMKA
UniGene
Hs.743976
Gene Chromosome Location
Chr.5: 150219491-150289840 on Build GRCh38
Transcripts
NM_015981.3 NM_171825.2 XM_017009898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523236 c.1204G>A p.G402S Chr.5: 150228225-150228225 on Build GRCh38
1064315 c.1168G>T p.A390S Chr.5: 150228261-150228261 on Build GRCh38
1672036 c.1153G>T p.D385Y Chr.5: 150228276-150228276 on Build GRCh38
1240963 c.1186C>A p.L396M Chr.5: 150228243-150228243 on Build GRCh38
3852755 c.1153G>A p.D385N Chr.5: 150228276-150228276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199731337 Chr.5: 150228323-150228323 on Build GRCh38
rs763794315 Chr.5: 150228194-150228194 on Build GRCh38
rs755573671 Chr.5: 150228291-150228291 on Build GRCh38
rs748310058 Chr.5: 150228158-150228158 on Build GRCh38
rs773681241 Chr.5: 150228356-150228356 on Build GRCh38
rs1047209344 Chr.5: 150228081-150228081 on Build GRCh38
rs559912434 Chr.5: 150228256-150228256 on Build GRCh38
rs371819590 Chr.5: 150228332-150228332 on Build GRCh38
rs747326987 Chr.5: 150228394-150228394 on Build GRCh38
rs1013571163 Chr.5: 150228419-150228419 on Build GRCh38
rs1029154818 Chr.5: 150228126-150228126 on Build GRCh38
rs115695646 Chr.5: 150228020-150228020 on Build GRCh38
rs962547326 Chr.5: 150228017-150228017 on Build GRCh38
rs931240153 Chr.5: 150228076-150228076 on Build GRCh38
rs199818897 Chr.5: 150228324-150228324 on Build GRCh38
rs781376173 Chr.5: 150228293-150228293 on Build GRCh38
rs899562609 Chr.5: 150228105-150228105 on Build GRCh38
rs947900009 Chr.5: 150228054-150228054 on Build GRCh38
rs752096411 Chr.5: 150228290-150228290 on Build GRCh38
rs773120233 Chr.5: 150228165-150228165 on Build GRCh38
rs375087065 Chr.5: 150228277-150228277 on Build GRCh38
rs373861512 Chr.5: 150228159-150228159 on Build GRCh38
rs918026134 Chr.5: 150228193-150228193 on Build GRCh38
rs771933373 Chr.5: 150228177-150228177 on Build GRCh38
rs968744576 Chr.5: 150228133-150228133 on Build GRCh38
rs113291724 Chr.5: 150228190-150228190 on Build GRCh38
rs574279988 Chr.5: 150228098-150228098 on Build GRCh38
rs541226321 Chr.5: 150228114-150228114 on Build GRCh38
rs886779989 Chr.5: 150228086-150228086 on Build GRCh38
rs996777266 Chr.5: 150228115-150228115 on Build GRCh38
rs371839355 Chr.5: 150228238-150228238 on Build GRCh38
rs1044889162 Chr.5: 150228055-150228055 on Build GRCh38
rs113331868 Chr.5: 150228191-150228191 on Build GRCh38
rs545265319 Chr.5: 150228019-150228019 on Build GRCh38
rs770091073 Chr.5: 150228161-150228161 on Build GRCh38
rs987328181 Chr.5: 150228417-150228417 on Build GRCh38
rs769870566 Chr.5: 150228312-150228312 on Build GRCh38
rs753708011 Chr.5: 150228210-150228210 on Build GRCh38
rs535748217 Chr.5: 150228407-150228407 on Build GRCh38
rs753832630 Chr.5: 150228152-150228153 on Build GRCh38
rs956936329 Chr.5: 150228416-150228416 on Build GRCh38
rs917971866 Chr.5: 150228011-150228011 on Build GRCh38
rs749310158 Chr.5: 150228317-150228317 on Build GRCh38
rs368362858 Chr.5: 150228298-150228298 on Build GRCh38
rs547567479 Chr.5: 150228349-150228349 on Build GRCh38
rs996607072 Chr.5: 150228347-150228347 on Build GRCh38
rs764964453 Chr.5: 150228236-150228236 on Build GRCh38
rs898438034 Chr.5: 150228066-150228066 on Build GRCh38
rs562358127 Chr.5: 150228097-150228097 on Build GRCh38
rs769152779 Chr.5: 150228149-150228149 on Build GRCh38
rs367981515 Chr.5: 150228175-150228175 on Build GRCh38
rs867691670 Chr.5: 150228289-150228290 on Build GRCh38
rs771256162 Chr.5: 150228431-150228431 on Build GRCh38
rs979709716 Chr.5: 150228137-150228137 on Build GRCh38
rs748703129 Chr.5: 150228333-150228333 on Build GRCh38
rs761351468 Chr.5: 150228217-150228217 on Build GRCh38
rs116674406 Chr.5: 150228337-150228337 on Build GRCh38
rs921314926 Chr.5: 150228362-150228362 on Build GRCh38
rs567965985 Chr.5: 150228393-150228393 on Build GRCh38
rs370156881 Chr.5: 150228150-150228150 on Build GRCh38
rs547238138 Chr.5: 150228441-150228441 on Build GRCh38
rs778064967 Chr.5: 150228313-150228313 on Build GRCh38
rs376699738 Chr.5: 150228192-150228192 on Build GRCh38
rs904027965 Chr.5: 150228373-150228373 on Build GRCh38
rs372610352 Chr.5: 150228322-150228322 on Build GRCh38

View Full Product Details