Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178916477-178916910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC107984119
Entrez Gene ID
107984119
Gene Name
zinc finger protein 181-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 178895637-178916670 on Build GRCh38
Transcripts
XM_017010140.1 NM_030613.3

Gene Symbol
ZFP2
Entrez Gene ID
80108
Gene Name
ZFP2 zinc finger protein
Gene Aliases
ZNF751, zfp-2
UniGene
Hs.654533
Gene Chromosome Location
Chr.5: 178895894-178933212 on Build GRCh38
Transcripts
XM_017010140.1 NM_030613.3

SNP Detail

SNP ID Chromosome Location
rs139618439 Chr.5: 178916705-178916706 on Build GRCh38
rs34040460 Chr.5: 178916869-178916870 on Build GRCh38
rs527558467 Chr.5: 178916771-178916771 on Build GRCh38
rs1004890418 Chr.5: 178916560-178916560 on Build GRCh38
rs916720550 Chr.5: 178916745-178916745 on Build GRCh38
rs141251285 Chr.5: 178916531-178916531 on Build GRCh38
rs564059667 Chr.5: 178916581-178916581 on Build GRCh38
rs11409413 Chr.5: 178916636-178916637 on Build GRCh38
rs397688243 Chr.5: 178916639-178916640 on Build GRCh38
rs906522089 Chr.5: 178916530-178916530 on Build GRCh38
rs1036387143 Chr.5: 178916836-178916836 on Build GRCh38
rs968009574 Chr.5: 178916861-178916861 on Build GRCh38
rs895216819 Chr.5: 178916515-178916515 on Build GRCh38
rs1011864383 Chr.5: 178916851-178916851 on Build GRCh38
rs1015222825 Chr.5: 178916564-178916564 on Build GRCh38
rs767590688 Chr.5: 178916835-178916835 on Build GRCh38
rs397723305 Chr.5: 178916638-178916639 on Build GRCh38
rs995058385 Chr.5: 178916582-178916582 on Build GRCh38
rs953204909 Chr.5: 178916603-178916603 on Build GRCh38
rs866437005 Chr.5: 178916744-178916744 on Build GRCh38
rs906958223 Chr.5: 178916820-178916820 on Build GRCh38
rs1047142376 Chr.5: 178916815-178916815 on Build GRCh38
rs12654454 Chr.5: 178916790-178916790 on Build GRCh38
rs570691024 Chr.5: 178916834-178916834 on Build GRCh38
rs911645851 Chr.5: 178916626-178916626 on Build GRCh38
rs966999967 Chr.5: 178916677-178916677 on Build GRCh38
rs1021858841 Chr.5: 178916856-178916856 on Build GRCh38
rs764933533 Chr.5: 178916528-178916528 on Build GRCh38
rs75814988 Chr.5: 178916684-178916684 on Build GRCh38
rs894637254 Chr.5: 178916845-178916845 on Build GRCh38
rs898181255 Chr.5: 178916770-178916770 on Build GRCh38
rs899062564 Chr.5: 178916874-178916874 on Build GRCh38
rs560404971 Chr.5: 178916709-178916709 on Build GRCh38
rs936963317 Chr.5: 178916725-178916725 on Build GRCh38
rs1046481290 Chr.5: 178916527-178916527 on Build GRCh38
rs1012314802 Chr.5: 178916521-178916521 on Build GRCh38
rs987195074 Chr.5: 178916611-178916611 on Build GRCh38
rs1028857480 Chr.5: 178916583-178916583 on Build GRCh38

View Full Product Details