Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39313980-39314471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK17
Entrez Gene ID
89822
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
K2p17.1, TALK-2, TALK2, TASK-4, TASK4
UniGene
Hs.162282
Gene Chromosome Location
Chr.6: 39299001-39314461 on Build GRCh38
Transcripts
NM_001135111.1 NM_031460.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359690 c.233T>A p.I78N Chr.6: 39314088-39314088 on Build GRCh38
3697852 c.67G>A p.V23M Chr.6: 39314254-39314254 on Build GRCh38
5815595 c.77T>A p.L26Q Chr.6: 39314244-39314244 on Build GRCh38
5787070 c.81C>T p.L27L Chr.6: 39314240-39314240 on Build GRCh38
6230579 c.82G>A p.A28T Chr.6: 39314239-39314239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769190726 Chr.6: 39314078-39314078 on Build GRCh38
rs1021970270 Chr.6: 39314059-39314059 on Build GRCh38
rs752761152 Chr.6: 39314240-39314240 on Build GRCh38
rs774012245 Chr.6: 39314183-39314183 on Build GRCh38
rs906031976 Chr.6: 39314029-39314029 on Build GRCh38
rs185410067 Chr.6: 39314389-39314389 on Build GRCh38
rs762943020 Chr.6: 39314145-39314145 on Build GRCh38
rs374734238 Chr.6: 39314436-39314436 on Build GRCh38
rs770254034 Chr.6: 39314320-39314320 on Build GRCh38
rs746939056 Chr.6: 39314072-39314072 on Build GRCh38
rs77147819 Chr.6: 39314391-39314391 on Build GRCh38
rs745806536 Chr.6: 39314151-39314151 on Build GRCh38
rs1056913670 Chr.6: 39314026-39314026 on Build GRCh38
rs373663901 Chr.6: 39314127-39314127 on Build GRCh38
rs776089414 Chr.6: 39314334-39314334 on Build GRCh38
rs568314591 Chr.6: 39314332-39314332 on Build GRCh38
rs751109636 Chr.6: 39314280-39314280 on Build GRCh38
rs748326556 Chr.6: 39314055-39314055 on Build GRCh38
rs986572279 Chr.6: 39314440-39314440 on Build GRCh38
rs947712221 Chr.6: 39314357-39314357 on Build GRCh38
rs760057589 Chr.6: 39314317-39314317 on Build GRCh38
rs192466468 Chr.6: 39314173-39314173 on Build GRCh38
rs756042230 Chr.6: 39314270-39314270 on Build GRCh38
rs764280915 Chr.6: 39314347-39314347 on Build GRCh38
rs936039113 Chr.6: 39314277-39314277 on Build GRCh38
rs771293542 Chr.6: 39314187-39314187 on Build GRCh38
rs559262761 Chr.6: 39314286-39314286 on Build GRCh38
rs146171979 Chr.6: 39314409-39314409 on Build GRCh38
rs761146240 Chr.6: 39314085-39314085 on Build GRCh38
rs757059294 Chr.6: 39314261-39314261 on Build GRCh38
rs966229395 Chr.6: 39314358-39314358 on Build GRCh38
rs28422024 Chr.6: 39314016-39314016 on Build GRCh38
rs766819933 Chr.6: 39314098-39314098 on Build GRCh38
rs755038296 Chr.6: 39314047-39314047 on Build GRCh38
rs771033826 Chr.6: 39314074-39314074 on Build GRCh38
rs35830268 Chr.6: 39314374-39314375 on Build GRCh38
rs766412671 Chr.6: 39314035-39314035 on Build GRCh38
rs139959421 Chr.6: 39314307-39314307 on Build GRCh38
rs557095645 Chr.6: 39314007-39314007 on Build GRCh38
rs765231421 Chr.6: 39314086-39314086 on Build GRCh38
rs891950807 Chr.6: 39314397-39314397 on Build GRCh38
rs778472867 Chr.6: 39314288-39314288 on Build GRCh38
rs542153673 Chr.6: 39314158-39314158 on Build GRCh38
rs747814509 Chr.6: 39314298-39314298 on Build GRCh38
rs372753787 Chr.6: 39314052-39314052 on Build GRCh38
rs370525658 Chr.6: 39314115-39314115 on Build GRCh38
rs371385851 Chr.6: 39314291-39314291 on Build GRCh38
rs753996296 Chr.6: 39314042-39314042 on Build GRCh38
rs575249030 Chr.6: 39314209-39314209 on Build GRCh38
rs912408544 Chr.6: 39314443-39314443 on Build GRCh38
rs1024498718 Chr.6: 39314314-39314314 on Build GRCh38
rs553576430 Chr.6: 39314429-39314429 on Build GRCh38
rs201418585 Chr.6: 39314131-39314131 on Build GRCh38
rs956253428 Chr.6: 39314045-39314045 on Build GRCh38
rs540036217 Chr.6: 39314076-39314076 on Build GRCh38
rs887999961 Chr.6: 39314004-39314004 on Build GRCh38
rs760990793 Chr.6: 39314186-39314186 on Build GRCh38
rs1021660157 Chr.6: 39314245-39314245 on Build GRCh38
rs779589903 Chr.6: 39314272-39314272 on Build GRCh38
rs34279441 Chr.6: 39314269-39314269 on Build GRCh38
rs748744661 Chr.6: 39314283-39314283 on Build GRCh38
rs927526508 Chr.6: 39314378-39314378 on Build GRCh38
rs62397171 Chr.6: 39314343-39314343 on Build GRCh38
rs765250531 Chr.6: 39314121-39314121 on Build GRCh38
rs897918400 Chr.6: 39314041-39314041 on Build GRCh38
rs946191112 Chr.6: 39314400-39314400 on Build GRCh38
rs375910334 Chr.6: 39314079-39314079 on Build GRCh38
rs1017539387 Chr.6: 39314355-39314355 on Build GRCh38
rs536415573 Chr.6: 39314386-39314386 on Build GRCh38
rs755021136 Chr.6: 39314110-39314110 on Build GRCh38
rs1044641038 Chr.6: 39314366-39314366 on Build GRCh38
rs1002973113 Chr.6: 39314206-39314206 on Build GRCh38
rs764874589 Chr.6: 39314011-39314011 on Build GRCh38
rs1040806698 Chr.6: 39314160-39314173 on Build GRCh38
rs771317153 Chr.6: 39314300-39314300 on Build GRCh38
rs779494063 Chr.6: 39314163-39314163 on Build GRCh38
rs968701926 Chr.6: 39314254-39314254 on Build GRCh38
rs1007453180 Chr.6: 39314337-39314337 on Build GRCh38
rs951228771 Chr.6: 39314439-39314439 on Build GRCh38
rs368194171 Chr.6: 39314165-39314165 on Build GRCh38
rs201819169 Chr.6: 39314146-39314146 on Build GRCh38
rs61748648 Chr.6: 39314140-39314140 on Build GRCh38
rs909535328 Chr.6: 39314147-39314147 on Build GRCh38
rs762570124 Chr.6: 39314080-39314080 on Build GRCh38
rs10947804 Chr.6: 39314260-39314260 on Build GRCh38
rs781115980 Chr.6: 39314142-39314142 on Build GRCh38
rs1002941876 Chr.6: 39314037-39314037 on Build GRCh38
rs765355370 Chr.6: 39314223-39314223 on Build GRCh38
rs757147464 Chr.6: 39314141-39314141 on Build GRCh38
rs773603729 Chr.6: 39314081-39314081 on Build GRCh38
rs754336373 Chr.6: 39314106-39314106 on Build GRCh38
rs763247377 Chr.6: 39314249-39314249 on Build GRCh38
rs780907022 Chr.6: 39314268-39314268 on Build GRCh38
rs370961739 Chr.6: 39314138-39314138 on Build GRCh38
rs201082190 Chr.6: 39314216-39314216 on Build GRCh38
rs564264745 Chr.6: 39314236-39314236 on Build GRCh38

View Full Product Details