Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83592866-83593373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP91
Entrez Gene ID
9892
Gene Name
synaptosome associated protein 91
Gene Aliases
AP180, CALM
UniGene
Hs.368046
Gene Chromosome Location
Chr.6: 83552885-83709408 on Build GRCh38
Transcripts
NM_001242792.1 NM_001242793.1 NM_001242794.1 NM_001256717.1 NM_001256718.1 NM_014841.2 XM_005248770.4 XM_006715615.1 XM_011536265.1 XM_011536266.1 XM_011536269.1 XM_011536271.1 XM_011536273.2 XM_011536275.2 XM_011536276.2 XM_011536277.1 XM_017011554.1 XM_017011555.1 XM_017011556.1 XM_017011557.1 XM_017011558.1 XM_017011559.1 XM_017011560.1 XM_017011561.1 XM_017011562.1 XM_017011563.1 XM_017011564.1 XM_017011565.1 XM_017011566.1 XM_017011567.1 XM_017011568.1 XM_017011569.1 XM_017011570.1 XM_017011571.1 XM_017011572.1 XM_017011573.1 XM_017011574.1 XM_017011575.1 XM_017011576.1 XM_017011577.1 XM_017011578.1 XM_017011579.1 XM_017011580.1 XM_017011581.1 XM_017011582.1 XM_017011583.1 XM_017011584.1 XM_017011585.1 XM_017011586.1 XM_017011587.1 XM_017011588.1 XM_017011589.1 XM_017011590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
484401 c.1730C>T p.A577V Chr.6: 83593226-83593226 on Build GRCh38
356514 c.1838T>A p.L613H Chr.6: 83592954-83592954 on Build GRCh38
4472525 c.1789C>T p.P597S Chr.6: 83593003-83593003 on Build GRCh38
3875855 c.1728A>C p.A576A Chr.6: 83593228-83593228 on Build GRCh38
4472524 c.1789C>T p.P597S Chr.6: 83593003-83593003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751328589 Chr.6: 83593052-83593052 on Build GRCh38
rs780080267 Chr.6: 83593156-83593156 on Build GRCh38
rs767230616 Chr.6: 83593273-83593273 on Build GRCh38
rs747462420 Chr.6: 83593231-83593231 on Build GRCh38
rs577695989 Chr.6: 83592938-83592938 on Build GRCh38
rs771212412 Chr.6: 83592903-83592903 on Build GRCh38
rs564502734 Chr.6: 83593301-83593301 on Build GRCh38
rs1047627270 Chr.6: 83593140-83593140 on Build GRCh38
rs548007011 Chr.6: 83593281-83593281 on Build GRCh38
rs911272474 Chr.6: 83592964-83592964 on Build GRCh38
rs929985041 Chr.6: 83593118-83593118 on Build GRCh38
rs1041039545 Chr.6: 83593100-83593100 on Build GRCh38
rs370917546 Chr.6: 83593114-83593114 on Build GRCh38
rs776735874 Chr.6: 83593242-83593242 on Build GRCh38
rs1003709549 Chr.6: 83593307-83593307 on Build GRCh38
rs1004066935 Chr.6: 83592973-83592973 on Build GRCh38
rs543135489 Chr.6: 83593001-83593001 on Build GRCh38
rs746221657 Chr.6: 83593203-83593203 on Build GRCh38
rs978676301 Chr.6: 83593126-83593126 on Build GRCh38
rs768643258 Chr.6: 83593235-83593235 on Build GRCh38
rs759318879 Chr.6: 83593269-83593269 on Build GRCh38
rs1021878212 Chr.6: 83593305-83593305 on Build GRCh38
rs1033654 Chr.6: 83593323-83593323 on Build GRCh38
rs760151771 Chr.6: 83592948-83592948 on Build GRCh38
rs909526839 Chr.6: 83593246-83593246 on Build GRCh38
rs562452559 Chr.6: 83593225-83593225 on Build GRCh38
rs897294162 Chr.6: 83592947-83592947 on Build GRCh38
rs774727476 Chr.6: 83592931-83592931 on Build GRCh38
rs766120982 Chr.6: 83593047-83593047 on Build GRCh38
rs754962003 Chr.6: 83593053-83593053 on Build GRCh38
rs756919437 Chr.6: 83593158-83593158 on Build GRCh38
rs765479981 Chr.6: 83593247-83593247 on Build GRCh38
rs374139901 Chr.6: 83593143-83593143 on Build GRCh38
rs545689640 Chr.6: 83593098-83593098 on Build GRCh38
rs755582333 Chr.6: 83593159-83593159 on Build GRCh38
rs781090309 Chr.6: 83593061-83593061 on Build GRCh38
rs944113261 Chr.6: 83593044-83593044 on Build GRCh38
rs756813838 Chr.6: 83593173-83593173 on Build GRCh38
rs755061472 Chr.6: 83593285-83593289 on Build GRCh38
rs61741864 Chr.6: 83592950-83592950 on Build GRCh38
rs370500053 Chr.6: 83592949-83592949 on Build GRCh38
rs867746242 Chr.6: 83593239-83593239 on Build GRCh38
rs922338558 Chr.6: 83593201-83593201 on Build GRCh38
rs575505443 Chr.6: 83593002-83593002 on Build GRCh38
rs753356649 Chr.6: 83593297-83593297 on Build GRCh38
rs914835109 Chr.6: 83592930-83592930 on Build GRCh38
rs1002594858 Chr.6: 83593202-83593202 on Build GRCh38
rs763542020 Chr.6: 83593292-83593292 on Build GRCh38
rs748901622 Chr.6: 83592897-83592897 on Build GRCh38
rs377348069 Chr.6: 83592998-83592998 on Build GRCh38
rs542171338 Chr.6: 83593119-83593119 on Build GRCh38
rs368460682 Chr.6: 83593182-83593182 on Build GRCh38
rs775771617 Chr.6: 83592978-83592978 on Build GRCh38
rs895240952 Chr.6: 83593343-83593343 on Build GRCh38
rs868610985 Chr.6: 83592994-83592994 on Build GRCh38
rs760832767 Chr.6: 83592980-83592980 on Build GRCh38
rs774185978 Chr.6: 83593258-83593258 on Build GRCh38
rs989001838 Chr.6: 83592914-83592914 on Build GRCh38
rs748913020 Chr.6: 83593181-83593181 on Build GRCh38
rs761985315 Chr.6: 83593245-83593245 on Build GRCh38
rs778703447 Chr.6: 83593309-83593309 on Build GRCh38
rs932357258 Chr.6: 83593213-83593213 on Build GRCh38
rs1006013657 Chr.6: 83593178-83593178 on Build GRCh38
rs754021482 Chr.6: 83593014-83593014 on Build GRCh38
rs756414542 Chr.6: 83593138-83593139 on Build GRCh38
rs201713356 Chr.6: 83593279-83593279 on Build GRCh38
rs762050063 Chr.6: 83593022-83593022 on Build GRCh38
rs779460520 Chr.6: 83593192-83593192 on Build GRCh38
rs1012192134 Chr.6: 83593074-83593074 on Build GRCh38
rs976947838 Chr.6: 83593197-83593197 on Build GRCh38
rs754003599 Chr.6: 83593155-83593156 on Build GRCh38
rs370989881 Chr.6: 83593226-83593226 on Build GRCh38
rs779114030 Chr.6: 83593285-83593286 on Build GRCh38
rs941622692 Chr.6: 83593259-83593259 on Build GRCh38

View Full Product Details